Selecting the correct reverse transcriptase (RT) for cDNA synthesis is critical to obtaining high yields of high-quality, full-length cDNA. We offer a range of stand-alone reverse transcriptases, optimized to generate cDNA for a variety of research needs.
Which stand-alone reverse transcriptase is right for you?
Value RT for commonly used RNA templates Effective & versatile RT for wide range of RNA templates Most effective RT for all RNAs, including difficult templates
Order now Order now Order now
Learn more >
SuperScript® III
Learn more >
SuperScript® IV
Learn more >
Optimal reaction temperature 37°C 50°C 50–55°C
Speed 60 min 50 min 10 min
Sensitivity Low High Up to 12.3 kb
cDNA yield Low High Superior
Max cDNA length Up to 7 kb Up to 12 kb Up to 20 kb
Resistance to reaction inhibitors Poor Good High
Protocol Download protocol Download protocol Download protocol