Plant Science News
Register for upcoming live events and listen to recorded past webinars here   View All
Selected Plant Research Publications featuring Life Technologies Products
Large-Scale Reverse Genetics in Arabidopsis: Case Studies from the Chloroplast 2010 Project
Agricultural Related Publications
Article link: Plant Phys 152:529 (2010)
Researchers at Michigan State University took part in the Chloroplast 2010 Project, designed to screen thousands of sequence-indexed Arabidopsis thaliana T-DNA insertion mutants for various phenotypes. In order to have confidence that a given mutant phenotype is associated with an alteration to a specific gene, the team looked for two or more independent homozygous mutants for each target gene. A high-throughput method was essential for this number of queries. The team employed a first-pass SYBR® Green method to eliminate homozygous samples. The remaining samples were subjected to further analysis to distinguish between heterozygous and wild-type genotypes using the two-reaction method and agarose gel electrophoresis involving Applied Biosystems® SYBR® Green PCR Master Mix and AmpliTaq Gold® with PCR on the 7900 HT Sequence Detection System.
Sulfur Transfer through an Arbuscular Mycorrhiza
Plant Science Related Publications
Article link: Plant Phys 149:549 (2008)
In an investigation aimed at the role of arbuscular mycorrhizal symbiosis in sulfur (S) uptake in plants was undertaken by a team at Michigan State University, USA. The researchers sought to determine which forms of S were taken up, metabolized, and transferred to the plant host roots by the fungal partner and to identify fungal genes were responsible for handling S. To examine fungal genes potentially involved in S transport, total RNA was isolated from frozen germinating spore tissue, and cDNA was synthesized using SuperScript® II reverse transcriptase. Gene fragments were generated using PCR and gene-specific primers, and were sequenced using an Applied Biosystems® 3130 Genetic Analyzer. Real-time PCR (using the Applied Biosystems® 7900 HT Sequence Detection System and Power SYBR® Green 2-Step Master Mix) revealed changes in a gene involved in sulfate transport and assimilation, which indicates a role for transcriptional regulation of sulfate permease in the reduction in sulfate transport in the presence of cysteine.
Agricultural Biotechnology Resources
Scientific Talks
  • Juan Enriquez, a founding director of the Harvard Business School Life Sciences Project and a former member of the Celera Genomics marine-based team to collect genetic data from the world's oceans speaks about biofuels.
    Watch the video
  • Dr Cary Fowler, Executive Director for the Global Crop Diversity Trust, discusses efforts to preserve the diversity of food crop traits through the maintenance of a global seed bank.
     Watch the video
  • Dr Jim Haseloff from the Department of Plant Sciences of the University of Cambridge discusses the use of Synthetic Biology approaches to Plant Research at the Synthetic Biology 4.0 Conference.
    Watch the video
Short Videos
  • Feeding the world: conversations with farmers at the Global Farmer-to-Farmer Roundtable and World Food Prize Symposium (October, 2010).
    Watch the video
  • Dr Pamela Roland at the University of California, Davis talks about genetic engineering approaches to stress tolerance in food crops.
    Watch the video
  • Harrison Dillon, CTO of Solazyme is growing strains of algae to produce renewable oils for Biofuels.
    Watch the video
  • Dr Sue Gardiner, Scientist in the Breeding & Genomics Group at Plant & Food Research in New Zealand, talks about her career in plant breeding technology.
    Watch the video
  • Estimating allele effects in Plant Breeding.
    Watch the video