Identify and measure antibodies
ELISA provides a fast, sensitive and convenient method to identify and quantify antibody classes, subclasses, and isotypes during and after hybridoma development to assess immune response.
Invitrogen immunoglobulin-specific ELISA kits and isotyping ELISA kits enable accurate quantitation and high throughput for testing more samples at a time. Select single-isotype or multi-isotype (isotyping) kits involving IgG, IgA, IgE, and IgM subclasses and isotypes from human, mouse and rat species.
Choose a kit featured on this page, or select an immunoglobulin ELISA kit from among all target-specific ELISA kits using the following selection guide.
Find ELISA kits by target
Antibody isotype determination
Invitrogen ELISA kits for isotyping are available for mouse and rat in two different formats:
  Ig Isotyping Instant ELISA kits Ig Isotyping Uncoated ELISA kits
Format Coated ELISA kit (Instant ELISA format) Uncoated ELISA kit
Features Addition of sample is all that is required, because coating & detector antibodies are already lyophilized on the plate. Antibodies come in vials and are not pre-coated onto the well plate, allowing you to modify the assay and protocol.
Features Measure 10 -12 samples per plate
Easy: Only 1 wash step
Fast: 2 hours total incubation
Size: 2 x 96 tests
Measure 10 -12 samples per plate
Less expensive
Customizable well pattern
Size: 10 x 96 tests
Isotypes IgG1, IgG2a, IgG2b, IgG3, IgA, IgM and kappa and lambda IgG1, IgG2a, IgG2b, IgG3, IgA, IgM and kappa and lambda
Mouse kits 88-50660-22 88-50630-86
88-50630-88 (with plates)
Rat kits 88-50650-22 88-50640-86
88-50640-88
Learn about other products for Antibody Isotyping and Characterization
Individual antibody isotype quantitation
Immunoglobulin-specific ELISA kits are available in two different formats:
  • Coated ELISA kits
    • Pre-coated plates
    • Serum and plasma samples
    • Comprehensive validated kits
  • Uncoated ELISA kits
    • Ready-to-use reagents included
    • Serum, plasma, and cell culture supernatants
    • 2, 10, or 20 plate kit size to suit your experiment size
View and select products
Select an immunoglobulin ELISA kit below or from among all target-specific ELISA kits using the following
selection guide: Find ELISA Kits by Target
Resources
Learning
Support
Related resources