Uncoated ELISA Kits
Affordable, coat-it-yourself ELISA kits
Invitrogen™ uncoated ELISA kits have added reagents needed to prepare and run the ELISA, including ELISA-optimized antibody matched pairs, standards, detection reagents, and coating buffers (plates are optional with 10 plate and 20 plate sizes).
Uncoated ELISAs are ideal for experienced ELISA users on a budget, with less stringent requirements for inter- and intra-assay variance.
Read on to learn more, or select uncoated ELISA kits from among all targets-specific ELISA kits using the following selection guide.
Key features of Invitrogen uncoated ELISA kits
  • Flexible—Purchase based on the volume of your research through a choice of package sizes, including 2-, 10-, and 20-plate kits.
  • Easy to use—Includes optimized antibody pairs and recombinant standards. Unlike ELISA sets from other suppliers, our uncoated ELISA kits also include TMB substrate and added essential reagents and buffers to perform your assay.
  • Affordable—Priced to accommodate the most demanding budgets and to maximize your research.
Simple kit structure of Invitrogen uncoated ELISA kits. Most of these “ready-set-go” kits are available in 2-, 10-, and 20-plate (96-well) kit sizes. Kits include all reagents necessary to perform the assay (with the exception of acid stop solution and wash buffer). Plates (96-well) are optional with the 10- and 20-plate kit sizes.
Procedure summary for “ready-set-go” Invitrogen uncoated ELISA kits.
Find your uncoated ELISA kit now
Select uncoated ELISA kits from among all targets-specific ELISA kits using the following selection guide.
Find ELISA kits by target
Visistat reference component