Invitrogen™ DynaMag™ magnets isolate any target in combination with Invitrogen™ Dynabeads™ magnetic beads. Your waiting time is minimized as these powerful magnets quickly pull your Dynabeads-bound target to the tube wall. The DynaMag magnets help ensure optimal working positions and are functionally adapted to suit your workflow.
General features
  • The strongest magnets we offer, these magnets are designed and certified for use with Dynabeads
  • Designed with ergonomics in mind
  • Choices to accommodate most working volumes
For cellular applications
1–5 mL tubes 15 mL conicals 50 mL conicals
For molecular applications
1.5–2 mL tubes >1.5–2 mL tubes 1–5 mL tubes
Magnet selection guide for Dynabeads products
Magnet Optimal working volume Description
 DynaMag™-2
Removable rack for vortexing, rotation, or manual sample shaking
10 µL–2,000 µL
(This magnet replaces the Dynal® MPC-S magnet)
  • Holds 16 (1.5–2 mL) microfuge tubes
  • 8 positions on each side of the rack
  • Top rack can be quickly removed from the magnet in the base, ready for vortexing, rotation or manual sample shaking. A center pin in the rack ensures equal vortexing of all tubes.
  • Excellent control and visibility of your isolations
SampleRack for DynaMag™-2
Removable plastic rack for handling and storage of sample tubes
 (see above)
  • The removable tube-holder plastic rack for the DynaMag™-2 is also available as a separate item
  • An additional SampleRack minimizes the need to move tubes between racks
  • The SampleRack can be placed on the bench, in the fridge, or in the incubator
DynaMag™-Spin
10 µL–1,500 µL
  • Holds 6 (1.5 mL) microfuge tubes
  • Circular top rack can be quickly removed from the magnet in the base, ready for vortexing or manual sample shaking
  • Good control of your isolations
DynaMag™-5
DynaMag-5
1 mL–5 mL
  • Flexible magnet: Handles many different tube sizes (11–16 mm in diameter, e.g. flow tubes and blood collection tubes) in a more functional way
  • More samples: Takes 8 tubes at a time of 1 mL to 5 mL volumes
  • Removable SampleRack: The tubes can be kept in the same space during the whole isolation process and beyond, just lift the whole SampleRack on/off the magnet base during handling. Buy an extra SampleRack and double the capacity
SampleRack for DynaMag™-5
(see above) 
  • The removable tube-holder plastic rack for the DynaMag™-5 is also available as a separate item
  • Additional SampleRacks may be useful when there are multiple users of the DynaMag™-5 magnet or when processing more than 8 samples at a time
  • The SampleRack can be placed on the bench, in the fridge, or in the incubator
 DynaMag™-15
Flip and use for 5 mL tubes!
1 mL–15 mL
  • Holds 4 standard 15 mL tubes
  • Can be used for any starting sample in combination with all types of Dynabeads®
  • Good control and visibility of your isolations
  • We recommend DynaMag™-5 for FACS tubes or blood collection tubes
 DynaMag™-50
 5 mL–50 mL
(This magnet replaces the Dynal MPC-50 magnet)
  • Holds 2 x 50 mL tubes
  • Can be used for any starting sample in combination with all types of Dynabeads®
  • Good control and visibility of your isolations
DynaMag™-96 Side
       (magnified below)
5 µL–200 µL
(This magnet replaces the Dynal MPC®-9600 magnet)
  • Holds PCR-Style Strips & Plates: 
    • PCR strips (8 & 12 well)
    • non-skirted 96-well plates
    • ½-skirted (200 µL) 96-well plates 
    • 48 & 96 microtubes for the Applied Biosystems® GeneAmp® PCR System
  • Pulls beads to the side of each well
  • For manual & automated work
  • Footprint same as a 96-well plate
  • Features 13 columns
  • In conjunction with an automated platform, the 13th column enables sample mixing without physically interacting with the sample.This is performed by repeatedly shifting the plate from the left-most to the right-most position & vice-versa.
 DynaMag™-96 Bottom
       (magnified below)
5 µL–200 µL
(This magnet replaces the Dynal MPC®-96B magnet)
  • Holds PCR-Style Strips & Plates: 
    • PCR strips (8 & 12 well)
    • non-skirted 96-well plates
    • ½-skirted (200 µL) 96-well plates 
    • 48 & 96 microtubes for the Applied Biosystems® GeneAmp® PCR System
  • Pulls beads to the bottom of each well
  • For manual & automated work
  • Footprint same as a 96-well plate
 DynaMag™-96 Side Skirted
       (magnified below)
5 µL–200 µL
  •  Holds skirted 96-well PCR-Style plates.
  •  Pulls beads to the side of each well.
  • For automated work.
  • Footprint size same as a 96-well plate.
Also works with the following cell culture plates:
  • 96-well round bottom plates
  • 96-well flat bottom plates
  • 24-well plates
  • 12-well plates
  • 6-well plates
  • not recommended for use with 48-well plates
 DynaMag™ CTS™
  • For closed sterile blood bags
  • Scalable volumes: 50 mL to 330 mL in static separations. 
  • >10 L in continuous flow separations following T cell expansion protocols.
  • Positively isolate bead-bound cells e.g. for subsequent stimulation/ expansion of T cells with Dynabeads® CD3/CD28 CTS™, and for removal of the beads following the expansion protocol.
Hulamixer sample mixer promo