Novex Assay Kits for Luminex Assays
Invitrogen™ multiplex assay kits are manufactured in our own ISO 13485 certified facility. This ensures reliable and quality products with rigorous validation.
Our target specific assays are based on Luminex® xMAP® technology and they are designed to simultaneously measure multiple specific protein targets in a single sample. Our assays can be used for either single or multiplexed quantitation of cytokines and signal transduction proteins using as little as 50 µL of sample. Our portfolio of standard and custom assays include both polystyrene (nonmagnetic) and magnetic reagent kits, which offer consistent performance with a broad range of sample types and species. We also offer complimentary in-person consulting through our Field Application Specialists. Contact us at LuminexFAS@thermofisher.com for personalized assistance.
Download our guide
Find out about the latest tools available for protein detection and quantitation. Download our free immunoassay guide to protein quantitiation
Need help?
We offer complimentary technical consulting with a field application specialist. Contact us
Popular products
Cytokine 30-Plex Panel, Human Magnetic Luminex Assay
Cytokine 20-Plex Panel, Mouse Magnetic Luminex Assay
Magnetic 96-Well Separator for Luminex Assays
Featured categories
Multiplex assays for the Luminex platform
Assay kits using magnetic and nonmagnetic polystyrene beads for simultaneous measurement of 3 to 30 analytes (cytokines, growth factors, etc.) using Luminex® xMAP® (multi-analyte profiling) technology.
Singleplex assays for the Luminex platform
Reagent sets of antibody-coated polystyrene (non-magnetic) beads, detector antibody, standards for measuring individual analytes using Luminex® xMAP® technology. Create and validate your own multiplexed combinations.
Custom assays for the Luminex platform
Select from a large portfolio of available targets to design your own multiplex assay and allow us to blend, optimize, and test it for you for use with Luminex® 100/200™, FLEXMAP 3D®, and MAGPIX® systems.
Magnetic singleplex assays for the Luminex platform
Reagent sets of antibody-coated magnetic beads, detector antibody, standards for measuring individual analytes using Luminex® xMAP® technology. Create and validate your own multiplexed combinations.
Singleplex, multiplex and custom assays for the Luminex platform
Our Invitrogen assays, available in polystyrene and magnetic bead formats, are an integral part of cytokine, pathway analysis, and biomarker discovery screening workflows.
  • Singleplex assays can be used to analyze individual protein targets—mix your own custom panels or try premixed multiplex assays
  • Premixed multiplex assays are available for the most widely-studied cytokines, adhesion proteins, and growth factors for human, primate (monkey), mouse, rat, and swine research samples
  • Custom luminex assays from a range of available protein targets, blended for you by our team to optimize for low background, high signal-to-noise ratios, and linearity of dilution
Our Invitrogen assays for the Luminex platform are easy to set up and run, using a procedure that is very similar to an ELISA (Figure 1). If you are new to Luminex® xMAP® technology or are using assays for the Luminex platform for the first time, we have a team of dedicated Field Application Scientists (FASs) ready to help. Our FASs come to your lab to guide your efforts running Luminex® assays and using Luminex® instruments and software. Contact LuminexFAS@thermofisher.com  to request your personal consultation today.
Luminex® xMAP® technology is based on polystyrene and paramagnetic microspheres (beads) that are internally dyed with red and infrared fluorophores of differing intensities.  Each dyed bead is given a unique number, or bead region, allowing the differentiation of individual beads.  Next, individual bead sets are coated with a capture antibody qualified for a specific analyte. Multiple analyte-specific beads can then be combined in a single well of a 96-well microplate assay format to detect and quantify up to 100 targets simultaneously using a Luminex® instrument for analysis.
Tables 1, 2, and 3 provide examples of the results achieved with our Invitrogen assays and protein standards (included in the assays). Unlike other assays, we use the maximum number of beads in each assay, so that data are collected from at least 100 events per bead region per target. Measurement of a minimum of 100 events, compared to the 50 recommended by some manufacturers, enables lower %CVs and more precise results.
Our Invitrogen assays are subject to rigorous quality control and validation prior to release; we extensively test cross-reactivity to help ensure the specificity of each assay for its intended target. In addition, the protein standards we supply are precisely calibrated to reference materials for maximum consistency.
Figure 1. Fast and easy Invitrogen assay protocol. Beads of defined spectral properties are conjugated to protein-specific capture antibodies and added along with samples (including standards of known protein concentration, control samples, and test samples), into the wells of a filter-bottom microplate. Target protein binds to the capture antibodies over the course of a two hour incubation. After washing the beads, protein-specific, biotinylated detector antibodies are added and incubated with the beads for 1 hr. Next, excess biotinylated detector antibodies are removed, and streptavidin-conjugated fluorescent protein, R-Phycoerythrin (SAV-R-PE), is added and incubated for 30 min. SAV-R-PE binds to the biotinylated detector antibodies, forming a four-member, solid-phase sandwich. After washing to remove unbound SAV-RPE, the beads are analyzed with a Luminex® detection system. By monitoring the spectral properties of the beads and the amount of associated R-Phycoerythrin (R-PE) fluorescence, the concentration of one or more proteins can be determined.
Table 1. More reliable data
These data indicate that %CVs and assay precision are improved by collecting data from 100 bead events compared to collecting data from only 50 bead events.
Sample Description
%CV (MFI)
50 Bead events
(0.5X Beads/ well)
100 Bead events
(1X Beads/ well)
Average (Serum)8.712.0
Average (Plasma)14.8 7.3
Average (TCS)8.55.2
Average (All)10.56.9
Table 2. Better-calibrated standards
Our Invitrogen assays are calibrated to National Institute of Biological Standards and Controls (NIBSC), a set of international reference standards, when available, or to published intracellular cell models for phosphoprotein activity to help ensure that measurements are relevant. Protein standards are tested before and after lyophilization, in single as well as multiplexed assays, for consistency in standard integrity and performance.
Standards for Invitrogen Multiplex Assays for the Luminex platform
  Expected
(pg/ml)
Calc.
(pg/ml)
%
Differ
GM-CSF1,9641,9881
IFN-γ1,078948-12
IL-21,4321,4894
IL-49821,08911
IL-61,0401,1026
IL-81,3571,266-7
IL-10954910-5
TNF-α2,3752,278-4
Competitor Standards
  Expected
(pg/ml)
Calc.
(pg/ml)
%
Differ
GM-CSF500271-46
IFN-γ500445-11
IL-2500484-3
IL-4500237-53
IL-6500301-40
IL-8500306-39
IL-1050076854
TNF-α5003,620624
Table 3. Greater assay specificity
We measured assay response from dozens of like-target proteins to help ensure that the antibodies selected for use in our Invitrogen assays are highly specific for their desired targets. In this table, we compare results (fluorescence units shown) for similar target proteins using our Invitrogen assays or assays from another manufacturer. None of the Invitrogen assays showed significant signal from similar, non-target proteins, whereas a few of the assays from the other supplier showed significant cross-reactivity with non-target proteins.
Invitrogen Multiplex Assays for the Luminex platform
  IL-6  IL-10  TNF-
α
IFN-
γ
IL-66,827312212
IL-10128,1512312
TNF-
α
7306,64210
IFN-
γ
835253,540
Zero1110137
Competitor Assay Fluorescence Units
  IL-6  IL-10  TNF-
α
IFN-
γ
IL-68,878153240
IL-1057,090812
TNF-
α
132159,999330
IFN-
γ
902020614,248
Zero31659
Free Protein Expression Handbook
Resources
Design Your Own Invitrogen Assay Maven Multimodule project for Thermo Fisher Scientific AEM functional modules
Support
Luminex® Instrument Support Maven Multimodule project for Thermo Fisher Scientific AEM functional modules