PB4500_Luminex_Instrument_Support
Luminex® xMAP® technology combines advanced fluidics, optics, and digital signal processing with proprietary microsphere technology to deliver multiplexed assay capabilities. This information is presented to support researchers who use MAGPIX®, Luminex 100/200™, and FlexMAP 3D® Luminex® systems.
If your Luminex® instrument needs attention or you require technical assistance with your Luminex® assays or xPONENT® software for data acquisition and analysis, contact our Technical Support team or email LuminexFAS@lifetech.com.
Luminex® instrument information and support
If your Luminex® instrument needs attention or you require technical assistance with your Luminex® assays, contact our Technical Support team or email LuminexFAS@lifetech.com.
  Description: https://www.lifetechnologies.com/content/dam/LifeTech/migration/images/cell-analysis/other.par.35996.image.100.70.1.thu-magpix-gif.gif 
Efficient and compact in size
Description: https://www.lifetechnologies.com/content/dam/LifeTech/migration/images/cell-analysis/other.par.65736.image.100.70.1.thu-lum-200-gif.gif 
The versatile, efficient  standard in multiplexing
Description: https://www.lifetechnologies.com/content/dam/LifeTech/migration/images/cell-analysis/other.par.93767.image.100.70.1.thu-flexmap-3d-gif.gif 
High-throughput, multiplex, automation compatible
  MAGPIX® System Luminex® 200™ System FLEXMAP 3D® System 
Optics LED/CCD camera Lasers/APDs/PMTs Lasers/APDs/PMTs
Hardware Fluorescent imager Flow cytometry based Flow cytometry based
Bead compatibility Magnetic Magnetic/polystyrene Magnetic/polystyrene
Multiplex capacity 50 100 500
Read time (96-well plate) ~ 60 min ~ 40 min ~ 20 min
Dynamic range 3.5 logs 3.5 logs 4.5 logs
Microtiter plate 96 well 96 well 96 and 384 well
Magnetic 96-well Separator Catalog #A14179 (compatible with MAGPIX®, Luminex® 200™, and FLEXMAP 3D® Systems)
xPONENT® Software
xPONENT® software is a modular, flexible, software package for control of the MAGPIX®, Luminex®200™, and FLEXMAP 3D® systems, offering:
  • Intuitive graphical user interface for simplified workflow
  • Automated startup, shutdown, and routine maintenance operations
  • Enhanced data analysis and report generation capabilities
  • 21 CFR Part 11 ready
  • Automation and LIS connectivity
xPONENT® Software benefits
  • Modular and flexible—Designed for ease of use, xPONENT® software allows novice, occasional, and advanced users set up and run assays in minimal amounts of time.
  • User customization—The default graphical user interface follows a typical assay workflow, and it can be customized to meet the needs of individual laboratories.
  • Powerful data analysis—Industry-standard curve fitting models for quantitative analysis, allelic ratios for genotyping, and qualitative algorithms for immunology assays in addition to data visualization tools.
xPONENT® Software support
If you require technical assistance with your xPONENT® software for data acquisition and analysis, contact our Technical Support team or email LuminexFAS@lifetech.com.
Luminex® Assays
See our selection of Singleplex Luminex® Assays and Multiplex Luminex® Assays. Or search our catalog for immunoassays using the Immunoassay Selection Guide.
If you can't find the assay that matches your experiment, contact us about a custom Luminex® assay.
Free white paper