Robust multiplex panels and combinable simplex kits for the Luminex platform
Invitrogen ProcartaPlex multiplex immunoassays consist of pre-configured multi-analyte reagent panels and single-analyte kits of prepared magnetic beads for quantitative analysis of protein targets in a single sample using Luminex technology. “Convenience” multiplex panels are pre-mixed and optimized bead sets for use without modification. “Combinable” multiplex panels are target sets which can be combined or supplemented by addition of beads from simplex kits.
Multiplex panels
ProcartaPlex multiplex panels are pre-configured mixtures of target-specific magnetic beads for quantitative analysis of multiple targets in a single sample. Panels are available for samples from six species (human, mouse, rat, non-human primate, porcine, and canine).
Panel features
  • Choose from a variety of commonly investigated biologically- and disease-defined analyte panels.
  • Quantitatively measure secreted biomarkers in plasma, serum, other bodily fluids, and cell culture supernatants.
  • Select “convenience” panels to use the assay as configured, or select “combinable” panels to customize the assay by mixing with another panel or adding compatible ProcartaPlex simplex beads.
  • Create unique panels by combining individual simplex kits can be purchased individually with a ProcartaPlex basic Kit (includes all non-analyte specific reagents needed to run the ProcartaPlex assay) to create your unique customized multiplex panel.
  • Alternatively, consider ProcartaPlex High Sensitivity Assays or ProcartaPlex Platinum Assays.
Combinable and convenience multiplex panels by species
Human combinable panels
Products can be combined with other combinable panels and simplex kits. Detection antibody (50X) needs to be diluted prior usage. Assays may contain more than one vial of beads and detection antibody.
Human Panel Name Cat. No. Antigens
Cytokine/Chemokine/Growth Factor 45-Plex Human Panel 1 EPX450-12171-901 BDNF, EGF, Eotaxin, FGF-basic, GM-CSF, GROα, HGF, IFNγ, IFNα, IL-1RA, IL-1β, IL-1α, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-12 p70, IL-13, IL-15, IL-17A, IL-18, IL-21, IL-22, IL-23, IL-27, IL-31, IP-10, LIF, MCP-1, MIP-1α, MIP-1β, NGFβ, PDGF-BB, PLGF, RANTES, SCF, SDF1α, TNFα, TNFβ, VEGF-A, VEGF-D
Cytokine & Chemokine 34-Plex Human Panel 1A EPX340-12167-901 Eotaxin, GM-CSF, GROα, IFNα, IFNγ, IL-1β, IL-1α, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-12 p70, IL-13, IL-15, IL-17A, IL-18, IL-21, IL-22, IL-23, IL-27, IL-31, IP-10, MCP-1, MIP-1α, MIP-1β, RANTES, SDF1α, TNFα, TNFβ
Cytokine 25-Plex Human Panel 1B EPX250-12166-901 GM-CSF, IFNα, IFNγ, IL-1α, IL-1β, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-9, IL-10, IL-12 p70, IL-13, IL-15, IL-17A, IL-18, IL-21, IL-22, IL-23, IL-27, IL-31, TNFα, TNFβ
Inflammation 20-Plex Human Panel EPX200-12185-901 E-Selectin, GM-CSF, ICAM-1, IFNα, IFNγ, IL-1α, IL-1β, IL-4, IL-6, IL-8, IL-10, IL-12p70, IL-13, IL-17A, IP-10, MCP-1, MIP-1α, MIP-1β, P-Selectin, TNFα
Th1/Th2 & Chemokine 20-Plex Human Panel 1 EPX200-12173-901 Eotaxin, GM-CSF, GROα, IFNγ, IL-1β, IL-2, IL-4, IL-5, IL-6, IL-8, IL-12 p70, IL-13, IL-18, IP-10, MCP-1, MIP-1α, MIP-1β, RANTES, SDF1α, TNFα
Th1/Th2 & Cytokine 18-Plex Human Panel 1C EPX180-12172-901 GM-CSF, IFNα, IFNγ, IL-1α, IL-1β, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-12 p70, IL-13, IL-15, IL-18, IL-31, TNFα, TNFβ
Th1/Th2/Th9/Th17/Th22/Treg Cytokine 18-Plex Human Panel EPX180-12165-901 GM-CSF, IFNγ, IL-1β, IL-2, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12 p70, IL-13, IL-17A, IL-18, IL-21, IL-22, IL-23, IL-27, TNFα
Th9/Th17/Th22/Treg & Chemokine 16-Plex Human Panel 1 EPX160-12175-901 Eotaxin, GROα, IL-8, IL-9, IL-10, IL-17A, IL-21, IL-22, IL-23, IL-27,IP-10, MCP-1, MIP-1α, MIP-1β, RANTES, SDF1α
Cytokine 1C & Chemokine 16-Plex Human Panel 1 EPX160-12176-901 Eotaxin, GROα, IFNα, IL-1α, IL-1RA, IL-7, IL-8, IL-15, IL-31, IP-10, MCP-1, MIP-1α, MIP-1β, RANTES, SDF1α, TNFβ
Th9/Th17/Th22/Treg & Cytokine 14-Plex Human Panel 1C EPX140-12174-901 Eotaxin, GROα, IL-8, IL-9, IL-10, IL-17A, IL-21, IL-22, IL-23, IL-27,IP-10, MCP-1, MIP-1α, MIP-1β, RANTES, SDF1α
Immuno-Oncology Checkpoint 14-Plex Human Panel 1 EPX14A-15803-901 BTLA, CD28, CD80, CD137, CD27, CD152, GITR, HVEM, IDO, LAG-3, PD-1, PD-L1, PD-L2, TIM-3
Immuno-Oncology Checkpoint 14-Plex Human Panel 2 EPX140-15815-901 MICA, MICB, Nectin-2, NT5E, Perforin, PVR, Siglec-7, Siglec-9, ULBP-1, ULBP-3, Tactile, E-Cadherin, Arginase-1, ULBP-4
Growth Factor 11-Plex Human Panel EPX110-12170-901 BDNF,NGFβ, EGF, FGF-basic, HGF, LIF, PDGF-BB, PlGF, SCF, VEGF-A, VEGF-D
Th1/Th2 Cytokine 11-Plex Human Panel EPX110-10810-901 GM-CSF, IFNγ, IL-1β, IL-2, IL-4, IL-5, IL-6, IL-12 p70, IL-13, IL-18, TNFα
Chemokine 9-Plex Human Panel 1 EPX090-12187-901 Eotaxin, GROα, RANTES, IL-8, IP-10, MCP-1, MIP-1α, MIP-1β, SDF1α
Th9/Th17/Th22/Treg Cytokine 7-Plex Human Panel EPX070-10817-901 IL-9, IL-10, IL-17A, IL-21, IL-22, IL-23, IL-27
Cytokine 7-Plex Human Panel 1C EPX070-10010-901 IFNα, IL-1α, IL-1RA, IL-7, IL-15, IL-31, TNFβ
Essential Th1/Th2 Cytokine 6-Plex Human Panel EPX060-10009-901 IFNγ, IL-4, IL-5, IL-6, IL-12 p70, TNFα
MMP 5-Plex Human Panel 1 EPX050-10015-901 MMP-1, MMP-7, MMP-8, MMP-12, MMP-13
Essential 4-Plex Human Panel 2 EPX040-10008-901 IL-8, IL-1β, IL-10, IL-17A
MMP 3-Plex Human Panel 2 EPX030-10829-901 MMP-2, MMP-3, MMP-9
Human convenience panels
Beads and detection antibodies are premixed in one vial each. Panels are not combinable with other ProcartaPlex assays.
Human Panel Name Cat. No. Antigens
Immune Monitoring 65-Plex Human Panel EPX650-10065-901 APRIL, BAFF, BLC, CD30, CD40L, ENA-78, Eotaxin, Eotaxin-2, Eotaxin-3, FGF-2, Fractalkine, G-CSF, GM-CSF, Gro a, HGF, IFN-a, IFN-g, IL-10, IL-12p70, IL-13, IL-15, IL-16, IL-17A, IL-18, IL-1a,IL-1b, IL-2, IL-20, IL-21, IL-22, IL-23, IL-27, IL-2R, IL-3, IL-31, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IP-10, I-TAC, LIF, MCP-1, MCP-2, MCP-3, M-CSF, MDC, MIF, MIG, MIP1a, MIP-1b, MIP-3a, MMP-1,NGF beta, SCF, SDF-1a, TNF b, TNF-a, TNF-R2, TRAIL, TSLP, TWEAK, VEGF-A
Cytokine/Chemokine/Growth Factor Convenience 45-Plex Human Panel 1 EPXR450-12171-901 BDNF, EGF, Eotaxin, FGF-basic, GM-CSF, GROα, HGF, IFNγ, IFNα, IL-1RA, IL-1β, IL-1α, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-12 p70, IL-13, IL-15, IL-17A, IL-18, IL-21, IL-22, IL-23, IL-27, IL-31, IP-10, LIF, MCP-1, MIP-1α, MIP-1β, NGFβ, PDGF-BB, PLGF, RANTES, SCF, SDF1α, TNFα, TNFβ, VEGF-A, VEGF-D
Cytokine & Chemokine Convenience 34-Plex Human Panel 1A EPXR340-12167-901 Eotaxin, GM-CSF, GROα, IFNα, IFNγ, IL-1β, IL-1α, IL-1RA, IL-2, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-12 p70, IL-13, IL-15, IL-17A, IL-18, IL-21, IL-22, IL-23, IL-27, IL-31, IP-10, MCP-1, MIP-1α, MIP-1β, RANTES, SDF1α, TNFα, TNFβ
TNF Superfamily 15-Plex Human Panel EPX150-15801-901 4-1BBL, APRIL, BAFF, CD30L, EDA-1, FasL, GITRL, CD40L, NGFβ, TNFα, TNFβ, LIGHT, RANKL, TRAIL, TWEAK
TNF Receptor Superfamily 12-Plex Human Panel EPX120-15802-901 Fas (APO), CD27, CD30, CD40, 4-1BB , DR6, GITR, HVEM, TNF-R1, TNF-R2, TRAIL-R1,TRAIL-R2
Metabolism/Obesity 9-Plex Human Panel 2 EPX09A-15804-901 C-peptide, Cortisol, FGF-23, GLP-1, Ghrelin, Insulin, PYY, Leptin, Resisitin
Graft Versus Host Disease 8-Plex Human Panel EPX080-15814-901 CD30, Granzyme A, HGF, IL-2R (CD25), KRT18, ST2/IL33R, Syndecan, TNF RI
Myokine 8-Plex Human Panel EPX080-12186-901 BDNF, IL-1RA, IL-10, IL-15, IL-6, IL-8, LIF, TNFα
Antibody Isotyping 7-Plex Human Panel EPX070-10818-901 IgG1, IgG2, IgG3, IgG4, IgE, IgA, IgM
Coagulation 6-Plex Human Panel 1 EPX060-10824-901 Factor XI, Factor XII, Factor XIII, Antithrombin, Prothrombin, CRP
Metabolism/Obesity 5-Plex Human Panel 1 EPX050-15805-901 Adiponectin, ICAM-1, NGAL, ZAG, RBP4
Coagulation 4-Plex Human Panel 3 EPX040-10825-901 Factor IX, Protein S, Protein C, vWF
Coagulation 3-Plex Human Panel 2 EPX030-10823-901 Factor V, Factor VII, Factor VIII
Mouse combinable panels
Products can be combined with other combinable panels and simplex kits. Detection antibody (50X) needs to be diluted prior usage. Assays may contain more than one vial of beads and detection antibody.
Mouse Panel Name Cat. No. Antigens
Cytokine & Chemokine 36-Plex Mouse Panel 1A EPX360-26092-901 ENA-78, Eotaxin, G-CSF, GM-CSF, GROα, IFNα, IFNγ, IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12 p70, IL-13, IL-15/IL-15R, IL-17A, IL-18, IL-22, IL-23, IL-27, IL-28, IL-31, IP-10, LIF, MCP-1, M-CSF, MCP-3, MIP-1α, MIP-1β, MIP-2, RANTES, TNFα
Cytokine & Chemokine 26-Plex Mouse Panel 1 EPX260-26088-901 Eotaxin, GM-CSF, GROα, IFNγ, IL-1β, IL-2, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12 p70, IL-13, IL-17A, IL-18, IL-22, IL-23, IL-27, IP-10, MCP-1, MCP-3, MIP-1α, MIP-1β, MIP-2, RANTES, TNFα
Th1/Th2 & Chemokine 20-Plex Mouse Panel 1 EPX200-26090-901 Eotaxin, GROα, GM-CSF, IFNγ, IL-1β, IL-2, IL-4, IL-5, IL-6, IL-12 p70, IL-13, IL-18, IP-10, MCP-1, MCP-3, MIP-1α, MIP-1β, MIP-2, RANTES, TNFα
Th1/Th2/Th9/Th17/Th22/Treg Cytokine 17-Plex Mouse Panel EPX170-26087-901 GM-CSF, IFNγ, IL-1β, IL-2, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12 p70, IL-13, IL-17A, IL-18, IL-22, IL-23, IL-27, TNFα
Th9/Th17/Th22/Treg & Chemokine 15-Plex Mouse Panel EPX150-26089-901 IL-10, IL-17A, IL-22, IL-23, IL-27, IL-9, GROα, IP-10, MCP-1, MCP-3, MIP-1α, MIP-1β, MIP-2, RANTES, Eotaxin
Th1/Th2 Cytokine 11-Plex Mouse Panel EPX110-20820-901 GM-CSF, IFNγ, IL-1β, IL-2, IL-4, IL-5, IL-6, IL-12 p70, IL-13, IL-18, TNFα
Cytokine 10-Plex Mouse Panel 1B EPX100-26091-901 ENA-78, G-CSF, IFNα, IL-1α, IL-3, IL-15/IL-15R, IL-28, IL-31, LIF, M-CSF
Chemokine 9-Plex Mouse Panel 1 EPX090-20821-901 GROα, IP-10, MCP-1, MCP-3, MIP-1α, MIP-1β, MIP-2, RANTES, Eotaxin
Essential 8-Plex Mouse Panel 2 EPX080-20832-901 IL-1β, IL-10, IL-17A, GROα, MCP-1, MIP-2, MIP-1α, MIP-1β
Th9/Th17/Th22/Treg Cytokine 6-Plex Mouse Panel EPX060-20822-901 IL-9, IL-10, IL-17A, IL-22, IL-23, IL-27
Essential Th1/Th2 Cytokine 6-Plex Mouse Panel EPX060-20831-901 IFNγ, IL-4, IL-5, IL-6, IL-12 p70, TNFα
Mouse convenience panels
Beads and detection antibodies are premixed in one vial each. Panels are not combinable with other ProcartaPlex assays.
Mouse Panel Name Cat. No. Antigens
Immune Monitoring 48-Plex Mouse ProcartaPlex Panel EPX480-20834-901 BAFF, BTC, ENA-78, Eotaxin, G-CSF, G-CSF, GRO-a, IFN-a, IFN-g, IL-10, IL-12p70, IL-13, IL-15, IL-17A, IL-18, IL-19, IL-1a, IL1-b, IL-2, IL-22, IL-23, IL-25, IL-27, IL-28, IL-2Ra, IL-3, IL-31, IL-33, IL-33R, IL-4, IL-5, IL-6, IL-7, IL-7Ra, IL-9, IP-10, Leptin, LIF, MCP-1, MCP-3, M-CSF, MIP-1a, MIP-1b, MIP-2, RANKL, RANTES, TNF-a, VEGF-A
Cytokine & Chemokine Convenience 36-Plex Mouse Panel 1A EPXR360-26092-901 ENA-78, Eotaxin, G-CSF, GM-CSF, GROα, IFNα, IFNγ, IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12 p70, IL-13, IL-15/IL-15R, IL-17A, IL-18, IL-22, IL-23, IL-27, IL-28, IL-31, IP-10, LIF, MCP-1, M-CSF, MCP-3, MIP-1α, MIP-1β, MIP-2, RANTES, TNFα
Cytokine & Chemokine Convenience 26-Plex Mouse Panel 1 EPXR260-26088-901 Eotaxin, GM-CSF, GROα, IFNγ, IL-1β, IL-2, IL-4, IL-5, IL-6, IL-9, IL-10, IL-12 p70, IL-13, IL-17A, IL-18, IL-22, IL-23, IL-27, IP-10, MCP-1, MCP-3, MIP-1α, MIP-1β, MIP-2, RANTES, TNFα
Antibody Isotyping 7-Plex Mouse Panel 2 EPX070-20816-901 IgG1, IgG2b, IgG2c, IgG3, IgA, IgE, IgM
Antibody Isotyping 7-Plex Mouse Panel EPX070-20815-901 IgG1, IgG2a, IgG2b, IgG3, IgA, IgE, IgM
Myokine 5-Plex Mouse Panel EPX050-20833-901 IL-10, IL-15/IL-15R, IL-6, LIF, TNFα
IFN alpha/IFN beta 2-Plex Mouse Panel EPX020-22187-901 IFNα, IFN β
Canine Panel Name Cat. No. Antigens
Cytokine/Chemokine/Growth Factor 11-Plex Canine Panel 1 EPX11A-50511-901 IFNγ, IL-2, IL-6, IL-8, IL-10, IL-12 p40, MCP-1, NGFβ, SCF, TNFα, VEGF-A
Porcine Panel Name Cat. No. Antigens
Cytokine & Chemokine 9-Plex Porcine Panel 1 EPX090-60829-901 IL-1 β, IL-4, IL-6, IL-8, IL-10, IL-12p40, IFNα, IFNγ, TNFα
Rat combinable panels
Products can be combined with other combinable panels and simplex kits. Detection antibody (50X) needs to be diluted prior usage. Assays may contain more than one vial of beads and detection antibody.
Rat Panel Name Cat. No. Antigens
Cytokine & Chemokine 22-Plex Rat Panel EPX220-30122-901 IL-1α, G-CSF, IL-10, IL-17A, IL-1β, IL-6, TNFα, IL-4, GM-CSF, IFNγ, IL-2, IL-5, IL-13, IL-12p70, Eotaxin, GROα, IP-10, MCP-1, MCP-3, MIP-1α, MIP-2, RANTES
Th Complete 14-Plex Rat Panel EPX140-30120-901 L-1 α, IL-1 β, GM-CSF, G-CSF, IFNγ, IL-2, IL-4, IL-5, IL-6, IL-10, IL-12p70, IL-13, IL-17A/CTLA-8, TNFα
Chemokine 8-Plex Rat Panel EPX080-30121-901 GROα, MCP-1, MIP-1α, RANTES, Eotaxin, IP-10, MCP-3, MIP-2
Rat convenience panels
Beads and detection antibodies are premixed in one vial each. Panels are not combinable with other ProcartaPlex Assays.
Rat Panel Name Cat. No. Antigens
Antibody Isotyping 6-Plex Rat Panel EPX060-30123-901 IgG1, IgG2a, IgG2b, IgG2c, IgA, IgM
Kidney Toxicity 5-Plex Rat Panel 1 EPX050-30124-901 KIM-1, osteopontin, TFF3, VEGF-A, calbindin
Kidney Toxicity 5-Plex Rat Panel 2 EPX050-30125-901 Albumin, clusterin, cystatin, NGAL, TIMP-1
Non-human primate combinable panels
Products can be combined with other combinable panels and simplex kits. Detection antibody (50X) needs to be diluted prior usage. Assays may contain more than one vial of beads and detection antibody.
Non-Human Primate Panel Name Cat. No. Antigens
Cytokine/Chemokine/Growth Factor 37-Plex NHP Panel EPX370-40045-901 BDNF, BLC, Eotaxin, FGF-2, G-CSF, GM-CSF, IFNα, IFNγ, IL-1β, IL-10, IL-12p70, IL-13, IL-15, IL-17A, IL-18, IL-1RA, IL-2, IL-23, IL-4, IL-5, IL-6, IL-7, IL-8, IP-10, I-TAC, MCP-1, MIG, MIP-1α, MIP-1β, NGF β, PDGF-BB, sCD40L, SCF, SDF-1α, TNFα, VEGF-A, VEGF-D
Cytokine & Chemokine 30-Plex NHP Panel EPX300-40044-901 BLC, Eotaxin, G-CSF , GM-CSF, IFNα, IFNγ, IL-1β, IL-10, IL-12p70, IL-13, IL-15, IL-17A /CTLA-8, IL-18, IL-1RA, IL-2, IL-23, IL-4, IL-5, IL-6, IL-7, IL-8, IP-10, I-TAC, MCP-1, MIG, MIP-1α, MIP-1β, sCD40L, SDF-1α, TNFα
Th 14-Plex NHP Panel EPX140-40040-901 IFNγ, IL-12p70, IL-13, IL-1β, IL-2, IL-4, IL-5, IL-6, TNFα, GM-CSF, IL-18, IL-10, IL-17A, IL-23
Chemokine 10-Plex NHP Panel EPX100-40041-901 IL-8, IP-10, MCP-1, MIP-1α, MIP-1β, Eotaxin, SDF-1α, BLC, I-TAC, MIG
Growth Factor 7-Plex NHP Panel EPX070-40043-901 BDNF, FGF-basic, PDGF-BB, SCF, VEGF-A, VEGF-D, NGFβ
Cytokine 6-Plex NHP Panel 1B EPX060-40042-901 G-CSF, IFNα, IL-1RA, IL-15, IL-7, CD40L
Cross-reactivity chart for NHP ProcartaPlex simplex kits
Cat. No. Analyte Chim-
panzee
Afr. gr.
monkey
Ba-boon Cyno-
molgus
Pigtail
macaque
Rhesus
monkey
Mar-
moset
Squirrel
monkey
EPX01A-42032-901 Adiponectin +++ N.T. + + N.T. ++ - -
EPX01A-42116-901 BDNF  +++ ++ + + + ++ + +
EPX01A-42147-901 BLC/CXCL13 +++ +++ +++ +++ ++ +++ ++ +
EPX01A-40239-901 CD40L +++ + +++ - - +++ +++ +
EPX01A-42120-901 Eotaxin +++ + +++ +++ +++ +++ +++ ++
EPX01A-42074-901 FGF-2 +++ +++ ++ +++ ++ +++ +++ +++
EPX01A-42001-901 G-CSF +++ + +++ +++ +++ +++ ++ +
EPX01A-40283-901 GM-CSF +++ +++ + +++ + + + +
EPX01A-42027-901 Granzyme B +++ - - - - - - -
EPX01A-40201-901 ICAM-1 +++ +++ +++ +++ +++ +++ ++ ++
EPX01A-40216-901 IFN alpha ++ - + ++ - ++ ++ -
EPX01A-40228-901 IFN gamma +++ +++ +++ +++ ++ +++ +++ +++
EPX01A-40224-901 IL-1 beta +++ + +++ +++ ++ +++ ++ ++
EPX01A-40642-901 IL-10 +++ + + + - + ++ +
EPX01A-42090-901 IL-12p40 +++ +++ +++ +++ ++ +++ +++ +++
EPX01A-40238-901 IL-12p70 +++ ++ ++ + + + +++ +++
EPX01A-40231-901 IL-13 +++ + +++ + +/- ++ +/- +/-
EPX01A-42089-901 IL-15 +++ ++ ++ ++ + +++ +++ +
EPX01A-42017-901 IL-17A +++ +++ ++ ++ ++ +++ +++ +
EPX01A-42160-901 IL-17F +++ ++ ++ ++ ++ ++ + +
EPX01A-40267-901 IL-18 +++ +++ ++ ++ ++ ++ ++ +
EPX01A-42080-901 IL-1RA +++ ++ +++ +++ +++ +++ + +/-
EPX01A-40221-901 IL-2 +++ - - - - - + +
EPX01A-42023-901 IL-23 +++ ++ ++ ++ ++ +++ ++ ++
EPX01A-40225-901 IL-4 +++ +++ +++ +++ ++ +++ ++ ++
EPX01A-40278-901 IL-5 +++ ++ +++ ++ + ++ ++ -
EPX01A-40213-901 IL-6 +++ + ++ +++ +++ +++ +/- +/-
EPX01A-40237-901 IL-7 +++ - ++ ++ - +++ +++ +
EPX01A-40204-901 IL-8 +++ ++ +++ +++ +++ +++ +/- +/-
EPX01A-40284-901 IP-10 +++ +/- +/- +/- +/- +/- +/- +/-
EPX01A-42124-901 I-TAC +++ +++ +++ +++ ++ +++ ++ ++
EPX01B-40281-901 MCP-1 +++ +++ +++ +++ +++ +++ +/- +/-
EPX01A-42125-901 MCP-3 + + + + + + +++ +
EPX01A-42127-901 MIF +++ +++ +++ +++ +++ +++ +++ +++
EPX01A-40285-901 MIG +++ + ++ +++ ++ +++ ++ ++
EPX01A-42029-901 MIP-1 alpha +++ +++ +++ +++ ++ +++ ++ +++
EPX01A-42030-901 MIP-1 beta +++ +++ +++ +++ +++ +++ +++ +++
EPX01A-42117-901 NGF beta +++ + ++ - - +++ +++ +++
EPX01A-42033-901 PAI-1 +++ ++ + + + +++ +++ ++
EPX01A-42071-901 PDGF-BB +++ ++ +++ +++ +/- +++ +++ +++
EPX01A-40287-901 RANTES +++ +++ +++ +++ +++ +++ +++ +++
EPX01A-42040-901 Resistin +++ +++ +++ ++ +++ + +++ +/-
EPX01A-42137-901 SCF + N.T. N.T. N.T. N.T. + ++ +
EPX01A-42138-901 SDF-1 alpha +++ +++ +++ +++ +++ +++ + +/-
EPX01A-42139-901 TGF alpha ++ + + + + - - +
EPX01A-40249-901 TGF beta 1 +++ +++ ++ ++ ++ +++ +++ ++
EPX01A-40223-901 TNF alpha +++ ++ +++ +++ +++ +++ +++ +
EPX01A-40202-901 TNF beta +++ ++ ++ +++ ++ +++ - -
EPX01A-42004-901 TRAIL +++ ++ +++ ++ ++ ++ ++ ++
EPX01A-40232-901 VCAM-1 +++ +/- +/- - +/- - - -
EPX01A-40277-901 VEGF A +++ +++ +++ +++ +++ +++ +++ ++
EPX01A-42076-901 VEGF D N.T. N.T. N.T. N.T. N.T. N.T. ++ N.T.
N.T. = Not tested
View full listing of multiplex panels
Simplex kits
ProcartaPlex simplex kits are magnetic bead sets used to detect individual analytes. They are designed to enable the addition of targets to multiplex panels or the combination of individual simplex kits to produce custom panels. A corresponding basic kit is required when combining simplex kits separate from any multiplex panel.
View full listing of simplex kits
Resources
Learning resources
Support resources
Related resources