Which iBright Imaging System is right for you?
Whether you seek ultimate performance or the best value, we have a solution for you.
Specifications of iBright Imaging Systems (CL1000 and FL1000 models)
CL1000
FL1000
Applications
Chemiluminescence western blots Yes
Protein gels Yes
DNA gels Yes
Multiplex western blots No Yes
RGB fluorescence western blots No Yes
Near-IR western blots No Yes
Camera
Detector Cooled 16-bit CCD
Lens 25 mm, f/0.95
Resolution 9.1 megapixels
Field of view 22.5 x 18.0 cm
Binning modes 1 x 1, 2 x 2, 3 x 3, 4 x 4, 5 x 5, 6 x 6, 8 x 8
System interface
Touch screen display 12.1 inch, capacitive touch screen
Computer Internal
Storage and connectivity
Integrated hard drive 64 GB (additional 10 GB of expandable cloud storage included)
USB Yes
Network Yes
Image file format Proprietary image format (G2I), TIFF, JPG, PNG, PDF
System hardware
Excitation source Green LED transillumination Epi-LEDs, green LED transillumination
Filter wheel 2 filter positions, motorized 7 filter positions, motorized
Shipping
Dimensions (L x W x H) 68 x 38 x 60 cm
Weight 50 kg (110 lbs)
Visistat reference component