File Name: SetupiBlot3.0.0.exe
Instument:
Original iBlot® Gel Transfer Device
Version: 3.0.0
Date Published:
1/17/2012
Language:
English
Download Size:
30 MB
Download
Updated Programs
Program Number Volt Default Run Time Run Time Limit
P0 20 V for 1 min
23 V for 4 min
25 V for remainder
7 min 13 min
P1 25 6 min 10 min
P2 23 6 min 11 min
P3 20 7 min 13 min
P4 15 7 min 16 min
P5 10 7 min 25 min
P6 7.5 3 min 25 min
P7 5 3 min 25 min
P8 20 V for 2 min
23 V for 2 min
25 V for remainder
7 min 13 min
P9 20 V for 2 min
5 V for 3 min (x2)
8 min 8 min
System Requirements
  • Supported Operating Systems: Windows 7, Windows XP and Windows Vista
  • PC with 600 MHz equivalent or higher processor
  • 128 MB of hard drive space
Additional Requirements
Instructions
  1. Click the download button and save the file SetupiBlot3.0.0.exe to your computer.
  2. Click SetupiBlot3.0.0.exe to install the iBlot Updater (note where the program is installed so you can run it after installation)
  3. Make sure that the iBlot power switch is in the OFF position, and the device is NOT connected to a PC or other instruments before starting.
  4. Run iblotUpdater.exe and press next.
  5. Press and hold the "Select" button (black button closest to the LCD screen).
  6. Continue holding the "Select" button and turn the iBlot power on. Random symbols will be displayed on the LCD screen.
  7. Release the "Select" button and connect the iBlot to the computer with a USB A to B cable (A end into the computer, B end into the iBlot). The computer should now begin to search for the iBlot. This step may take several minutes. The PC will locate your iBlot and inform you if it is found.
  8. Press 'Next' to begin the iBlot Firmware Update. Do not disconnect or use device until iBlot updater is complete.
  9. After the update is complete, the program displays a message indicating that the update was successful.
  10. Disconnect the USB cable from the iBlot device.
Troubleshooting
  1. If the update was unsuccessful, the message "The Update failed" will appear.
  2. Please retry running the program, and if the problem persists, contact Invitrogen's Technical Support.
  3. Do NOT use a USB hub. The iBlot device must be connected directly to the computer for the firmware upgrade to work.