Protein Interaction Crosslinking for Mass Spectrometry
Thermo Scientific™ MS-grade crosslinkers are high-quality reagents that are available in multiple packaging options and sizes. We offer extensive technical expertise and support for various applications, as well as validation of these products in workflows using Thermo Scientific mass spectrometers.
  • High quality—products manufactured in ISO 9001–certified facilities
  • Convenience—products available in multiple package sizes, including “no-weigh” format
  • Wide variety—available with different linker lengths, MS-cleavability and deuterium isotope labels
  • Technical support—web resources and technical service to help ensure successful results
Choose the right crosslinker for your mass spec application
  DSG (disuccinimidyl glutarate) BS2G-d0 DSSO DSBU DSS BS3
Crosslinker DSG BS2G-d0 DSSO
DSBU
DSS BS3
Spacer arm 7.7 Å 7.7 Å 10.3 Å
12.5  Å
11.4 Å 11.4 Å
Water soluble No Yes No
No
No Yes
MS cleavable No No Yes
Yes
No No
  Order now Order now Order now Order now Order now Order now
  BS2G-d4 BS3-d4
Crosslinker BS2G-d4 BS3-d4
Spacer arm 7.7 Å 11.4 Å
Water soluble Yes Yes
MS cleavable No No
  Order now Order now
  DSG (disuccinimidyl glutarate) BS2G-d0 DSSO DSBU DSS BS3
Crosslinker DSG BS2G-d0 DSSO
DSBU
DSS BS3
Spacer arm 7.7 Å 7.7 Å 10.3 Å
12.5  Å
11.4 Å 11.4 Å
Water soluble No Yes No
No
No Yes
MS cleavable No No Yes
Yes
No No
  Order now Order now Order now Order now Order now Order now
  BS2G-d4 BS3-d4
Crosslinker BS2G-d4 BS3-d4
Spacer arm 7.7 Å 11.4 Å
Water soluble Yes Yes
MS cleavable No No
  Order now Order now
Featured product data
Comparison of BSA crosslinking efficiency by SDS-PAGE
Comparison of BSA crosslinking efficiency by SDS-PAGE. Different crosslinkers were incubated with BSA at molar excess of crosslinker to protein (e.g., 20- 100-, or 500-fold). Crosslinking efficiency is shown by decreased mobility by SDS-PAGE and varied by crosslinker type, solubility, and concentration.
BSA crosslinked peptide
BSA crosslinked peptide spectra identified by MS2-MS3 method and XLinkX using DSSO crosslinker. XlinkX software uses unique fragment ion patterns of MS-cleavable crosslinkers (purple annotation) to detect and filter crosslinked peptides for a crosslinked database search.
Graph showing number of BSA crosslinked peptides
Graph showing number of BSA crosslinked peptides identified using different noncleavable (BS3, DSS) and cleavable (DSSO) crosslinkers for various MSn methods. Both BS3 and DSS had similar numbers of crosslinked peptides identified for collision-induced fragmentation (CID) and high-energy CID (HCD) methods, but DSSO identified fewer crosslinked peptides. Because DSSO is MS-cleavable, the MS2-MS3 method can be used to identify more crosslinks when a linear-peptide database search is used. All crosslinkers showed similar numbers of identified crosslinked peptides by electron-transfer and higher-energy collision dissociation (EThcD).
Resources