Protect your precious samples
Our cryogenic storage tubes and accessories are designed as a system to protect and organize your valuable, irreplaceable samples. Scientists worldwide have been storing samples in Thermo Scientific Nalgene and Nunc cryo tubes for more than 40 years—whether you're storing hundreds of samples or millions, we have you covered.
Cryotube Selection Tool
Most Popular Cryotube Products
Nalgene Standard 1.8mL externally threaded
Nunc Universal 1.8mL externally threaded with 2D barcodes
Nunc Internally threaded 1.8mL round bottom non-starfoot tubes
Featured Cryotube Categories
Nunc Universal Tubes
Universal cap, universal latch rack, and free-standing tube. Available racked or in bulk, with 2D barcodes, non-coded or both 1D (linear) plus 2D barcodes to serve every need.
Nunc Biobanking & Cell Culture Tubes
Non-cytotoxic, non-mutagenic and pyrogen-free IVD compatible tubes. Internal and external thread options from 1-4.5mL working volumes. Racked, bulk, uncapped, and barcoded options available.
Nalgene General Long-Term Storage
Externally threaded tubes available in 1-15mL working volumes with and without barcoding options.
Nunc Coded Cryobank Vials
Available 2D barcoded and microplate racked or bulk formats supporting capping/decapping equipment and labeling options. Automation compatible and available in a volume range of 0.5-5.0mL.
Nunc General Stor-It Tubes
Contain samples in cost-efficient round-bottom, externally-threaded Thermo Scientific Nunc General Long Term Storage Tubes.
Linear Bar-coded Tubes
Nalgene and Nunc tubes with side printed linear barcodes available in 1-5mL working volumes. Available internal and externally threaded as well as capless options.
Nunc Bank-It Vials
Internally threaded and pre-racked tubes available in 0.5-1mL working volume. Available with 2D barcodes and both CE and IATA certified.
CryoBox Freezer Racks
Durable stainless steel and aluminum racks that fit most common upright and chest freezers.
Complete the Package!
Streamline the process of sample identification with Thermo Scientific Cryo Vial Closure Color Coders for Nalgene and Nunc cryovials.
Protect tubes submerged in LN2 and prevent tube explosion due to LN2 contamination with Nunc Cryoflex tubing or Cryosleeves.
Protect and organize your samples in materials ranging from economical cardboard to break-resistant, durable polycarbonate to meet any need and budget.