matrix-2d-storage-tubes
When your workflow includes semi-automated and fully-automated platforms for high throughput, Thermo Scientific™ tubes allow robust storage and tracking of sample volumes ranging from 7μL to 12mL in 384-, 96-, 48- and 24-place arrays. We have storage formats available for temperatures down to VPLN, and multiple sealing options including ScrewTop caps and septa seals. Solid, one-piece construction means there are no labels to fall off and tubes made from medical-grade resins are supplied free of RNAse, DNAse, endotoxins and cytotoxins and SAL down to 10-6.
Matrix Tube Selection Tool
Matrix Tube Formats and Caps
Matrix ScrewTop Tubes
Matrix 2D Tubes
  • Open top and screw top formats
  • Enable small volume storage
  • 384-, 96-, 48-, and 24-tube arrays ideal for single use
  • Color cap options
  • -196°C to 121°C
Matrix Duraseal
Matrix Duraseal
  • Permanently installed septa for long-term storage
  • Store down to -150°C
  • Can be pierced multiple times and maintain seals
Matrix SepraSeal
Matrix SepraSeal
  • Individual tube capping
  • Up to 96 septa at once
  • Store down to -20°C
  • Pre-split septa for multiple piercing
  • Mats for long term storage, pierceable,removable
100% leak tested to ensure tube integrity and sample security
  • Designed for automated systems including liquid handling, decappers and other automated platforms
  • Rack base and tube design prevents the tube from spinning in the rack when decapping of single tube hand decapping or, multi channel automated decapping
  • Laser-etched 2D barcode on the base of every tube ensures secure sample tracking with highest contrast available for readability even with some frost on the tube
  • The linear and human readable codes match this unique 2D barcode, allowing 1D scanning or visual identification of samples across satellite or collaborative sites without 2D readers
Colored Capping Options
RED = red
PUR = purple
BLU = blue
GRE = green
YEL = yellow
GRY = gray (septa only)
WHI = white (caps only)
* Not including 3747, 3748
Colored options are available for Matrix ScrewTop caps* and SepraSeal septa. Simply add the following 3 characters to each catalog number for the colored version equivalent. (e.g. 4465RED = red SepraSeal septa in 10 mats of 96/case).
Matrix Storage Tube Selection Guide
Filter Results
Recommended product(s)
Related Products
SuperSealer Instruments
SampleSeal Heat Sealer             
DuraSeal and SepraSeal Capping
Resources
Support
Need technical support? Contact our expert team for technical and application support of laboratory products.