Accurate heterozygous indel mutation detection
on an Applied Biosystems Genetic Analyzer.
Identifying heterozygous base positions or small insertions or deletions in genomic DNA is often employed to locate mutations or polymorphisms in diploid organisms, detect genetic rearrangements, and uncover rare variants. The combination of the new Primer Designer™ Tool, BigDye® Direct Cycle Sequencing Kit and the Applied Biosystems® 3500 Series Genetic Analyzers for comprehensive, reliable sequencing data on targeted DNA regions and in heterogeneous mixtures, is critical for countless life science and research applications.
Step-by-Step Guide to Resequencing for Heterozygote Detection
DNA Purification kit
DNA extraction is a critical first step in the experimental workflow of DNA Sequencing and Fragment analysis. The overall quality, accuracy and length of the DNA sequence read can be significantly affected by characteristics of the sample itself, and the method chosen for nucleic acid extraction. Ideal methods will vary depending on the source or tissue type, how it was obtained from its source, and how the sample was handled or stored prior to extraction.
Recommended Products: DNA Isolation
PCR Kit
Select and order pre-designed PCR and Sanger sequencing primer pairs using the Primer Designer™ Tool.
BigDye® Direct Cycle Sequencing Kit contains reagents to perform the PCR, PCR clean up and sequencing reactions.
Recommended Products: Perform PCR
DNA strands
BigDye® Direct Cycle Sequencing Kit contains reagents to perform the PCR, PCR clean up and sequencing reactions. PCR Clean up and sequencing reaction are perform in a single step.
BigDye® Direct cycle sequencing kits
Sequencing Reaction Cleanup
After the sequencing reaction, it is important to remove unincorporated dye terminators and salts that may compete for capillary electrophoretic injection. Unincorporated terminators can co-migrate with the sequencing template, resulting in basecalling.
Recommended Products: Purifying Sequencing Reaction
Which Sample Cleanup Product Is Right for You?
Cleanup Method Description & Application Product
Reagent-Based Sequencing Template Cleanup Scavenges all unincorporated BigDye® terminators. Stabilizes samples before analysis. Best for long and short fragment recovery.
Capillary Electrophoresis
During capillary electrophoresis, the products of the PCR are injected electrokinetically into capillaries filled with polymer. High voltage is applied so that the fluorescent DNA fragments are separated by size and are detected by a laser/camera system.
Which Electrophoresis Instrument (Genetic Analyzer) Is Right for You?
  Instrument
  3730xl
3730xL
3730
3730
3500xl
3500xL
3500
3500
3130xl
3130xL
3130
3130
310
310
Number of Capillaries 96 48 24 8 16 4 1
Compatible Applications:
(S) Supported; (A) AB Demonstrated; (C) Customer Demonstrated; (N) Not Supported
Heterozygote Detection & Resequenicng (SNP-based) S S S S S S A
After electrophoresis, Data Collection software creates a Sample File of the raw data. Using downstream software applications, further data analysis is required to translate the raw data into the corresponding electropherogram.
Which Data Analysis Software Is Right for You?
DNA Purification kit
DNA extraction is a critical first step in the experimental workflow of DNA Sequencing and Fragment analysis. The overall quality, accuracy and length of the DNA sequence read can be significantly affected by characteristics of the sample itself, and the method chosen for nucleic acid extraction. Ideal methods will vary depending on the source or tissue type, how it was obtained from its source, and how the sample was handled or stored prior to extraction.
Recommended Products: DNA Isolation
PCR Kit
Select and order pre-designed PCR and Sanger sequencing primer pairs using the Primer Designer™ Tool.
BigDye® Direct Cycle Sequencing Kit contains reagents to perform the PCR, PCR clean up and sequencing reactions.
Recommended Products: Perform PCR
DNA strands
BigDye® Direct Cycle Sequencing Kit contains reagents to perform the PCR, PCR clean up and sequencing reactions. PCR Clean up and sequencing reaction are perform in a single step.
BigDye® Direct cycle sequencing kits
Sequencing Reaction Cleanup
After the sequencing reaction, it is important to remove unincorporated dye terminators and salts that may compete for capillary electrophoretic injection. Unincorporated terminators can co-migrate with the sequencing template, resulting in basecalling.
Recommended Products: Purifying Sequencing Reaction
Which Sample Cleanup Product Is Right for You?
Cleanup Method Description & Application Product
Reagent-Based Sequencing Template Cleanup Scavenges all unincorporated BigDye® terminators. Stabilizes samples before analysis. Best for long and short fragment recovery.
Capillary Electrophoresis
During capillary electrophoresis, the products of the PCR are injected electrokinetically into capillaries filled with polymer. High voltage is applied so that the fluorescent DNA fragments are separated by size and are detected by a laser/camera system.
Which Electrophoresis Instrument (Genetic Analyzer) Is Right for You?
  Instrument
  3730xl
3730xL
3730
3730
3500xl
3500xL
3500
3500
3130xl
3130xL
3130
3130
310
310
Number of Capillaries 96 48 24 8 16 4 1
Compatible Applications:
(S) Supported; (A) AB Demonstrated; (C) Customer Demonstrated; (N) Not Supported
Heterozygote Detection & Resequenicng (SNP-based) S S S S S S A
After electrophoresis, Data Collection software creates a Sample File of the raw data. Using downstream software applications, further data analysis is required to translate the raw data into the corresponding electropherogram.
Which Data Analysis Software Is Right for You?
Learn More
Application Notes
Protocol Summary
Poster
Step-by-Step Guide to Resequencing for Heterozygote Detection
DNA Purification kit
DNA extraction is a critical first step in the experimental workflow of DNA Sequencing and Fragment analysis. The overall quality, accuracy and length of the DNA sequence read can be significantly affected by characteristics of the sample itself, and the method chosen for nucleic acid extraction. Ideal methods will vary depending on the source or tissue type, how it was obtained from its source, and how the sample was handled or stored prior to extraction.
Recommended Products: DNA Isolation
PCR Kit
Select and order pre-designed PCR and Sanger sequencing primer pairs using the Primer Designer™ Tool.
BigDye® Direct Cycle Sequencing Kit contains reagents to perform the PCR, PCR clean up and sequencing reactions.
Recommended Products: Perform PCR
DNA strands
BigDye® Direct Cycle Sequencing Kit contains reagents to perform the PCR, PCR clean up and sequencing reactions. PCR Clean up and sequencing reaction are perform in a single step.
BigDye® Direct cycle sequencing kits
Sequencing Reaction Cleanup
After the sequencing reaction, it is important to remove unincorporated dye terminators and salts that may compete for capillary electrophoretic injection. Unincorporated terminators can co-migrate with the sequencing template, resulting in basecalling.
Recommended Products: Purifying Sequencing Reaction
Which Sample Cleanup Product Is Right for You?
Cleanup Method Description & Application Product
Reagent-Based Sequencing Template Cleanup Scavenges all unincorporated BigDye® terminators. Stabilizes samples before analysis. Best for long and short fragment recovery.
Capillary Electrophoresis
During capillary electrophoresis, the products of the PCR are injected electrokinetically into capillaries filled with polymer. High voltage is applied so that the fluorescent DNA fragments are separated by size and are detected by a laser/camera system.
Which Electrophoresis Instrument (Genetic Analyzer) Is Right for You?
  Instrument
  3730xl
3730xL
3730
3730
3500xl
3500xL
3500
3500
3130xl
3130xL
3130
3130
310
310
Number of Capillaries 96 48 24 8 16 4 1
Compatible Applications:
(S) Supported; (A) AB Demonstrated; (C) Customer Demonstrated; (N) Not Supported
Heterozygote Detection & Resequenicng (SNP-based) S S S S S S A
After electrophoresis, Data Collection software creates a Sample File of the raw data. Using downstream software applications, further data analysis is required to translate the raw data into the corresponding electropherogram.
Which Data Analysis Software Is Right for You?
DNA Purification kit
DNA extraction is a critical first step in the experimental workflow of DNA Sequencing and Fragment analysis. The overall quality, accuracy and length of the DNA sequence read can be significantly affected by characteristics of the sample itself, and the method chosen for nucleic acid extraction. Ideal methods will vary depending on the source or tissue type, how it was obtained from its source, and how the sample was handled or stored prior to extraction.
Recommended Products: DNA Isolation
PCR Kit
Select and order pre-designed PCR and Sanger sequencing primer pairs using the Primer Designer™ Tool.
BigDye® Direct Cycle Sequencing Kit contains reagents to perform the PCR, PCR clean up and sequencing reactions.
Recommended Products: Perform PCR
DNA strands
BigDye® Direct Cycle Sequencing Kit contains reagents to perform the PCR, PCR clean up and sequencing reactions. PCR Clean up and sequencing reaction are perform in a single step.
BigDye® Direct cycle sequencing kits
Sequencing Reaction Cleanup
After the sequencing reaction, it is important to remove unincorporated dye terminators and salts that may compete for capillary electrophoretic injection. Unincorporated terminators can co-migrate with the sequencing template, resulting in basecalling.
Recommended Products: Purifying Sequencing Reaction
Which Sample Cleanup Product Is Right for You?
Cleanup Method Description & Application Product
Reagent-Based Sequencing Template Cleanup Scavenges all unincorporated BigDye® terminators. Stabilizes samples before analysis. Best for long and short fragment recovery.
Capillary Electrophoresis
During capillary electrophoresis, the products of the PCR are injected electrokinetically into capillaries filled with polymer. High voltage is applied so that the fluorescent DNA fragments are separated by size and are detected by a laser/camera system.
Which Electrophoresis Instrument (Genetic Analyzer) Is Right for You?
  Instrument
  3730xl
3730xL
3730
3730
3500xl
3500xL
3500
3500
3130xl
3130xL
3130
3130
310
310
Number of Capillaries 96 48 24 8 16 4 1
Compatible Applications:
(S) Supported; (A) AB Demonstrated; (C) Customer Demonstrated; (N) Not Supported
Heterozygote Detection & Resequenicng (SNP-based) S S S S S S A
After electrophoresis, Data Collection software creates a Sample File of the raw data. Using downstream software applications, further data analysis is required to translate the raw data into the corresponding electropherogram.
Which Data Analysis Software Is Right for You?
Learn More
Application Notes
Protocol Summary
Poster
Visistat reference component
Visistat reference component