The development and progression of cancer is a complex process, and many nucleic acid modifications are believed to play significant roles. These include single mutations in the DNA, RNA or mitochondrial DNA, deletions, insertions, and duplications of a gene (or part of a gene), DNA methylation, alternative splicing events, and more. Applied Biosystems® genetic analyzers and associated kits offer the flexibility you need to get the answers you’re looking for.
Screening Prior to Sequencing
category-70x75.par.87731.image.70.75.1.sscp
SSCP is an electrophoretic mobility assay used to find variants in a large number of samples so that fewer DNA sequencing runs are needed.
Learn more about:
Mutation Detection
category-70x75.par.13415.image.70.75.1.krass-braff
DNA mutations are the most common events driving the development of cancer. Our kits and genetic analysis instruments help simplify your application development process—for example SNP detection (a single SNP or up to 10 SNPs located on different DNA segments) and sequencing complete genes.
Learn more about:
Genetic Rearrangement
category-70x75.par.7942.image.70.75.1.rfq
Genetic rearrangements—including duplications, inversions, and deletions of DNA fragments—are known to contribute to many genetic diseases.
Learn more about:
Methylation Analysis
category-70x75.par.82216.image.70.75.1.msmsa
DNA methylation plays an important role in the regulation of gene expression and, consequently, in the appearance of various disease states. Using our methylation detection solutions, you can determine the overall level of methylation or identify the methylation status of a specific CpG.
Learn more about:
Visistat reference component
Resources
Interested in commercializing your assay?
We can help ›
Support
New Capillary Electrophoresis Instruments Support Center
Find tips, troubleshooting help, and resources for support related to your capillary electrophoresis instruments.
Fragment Analysis Software Support Center