Chimerism is a term used to describe the occurrence of genetically distinct cell types in a single organism, which can result from transfusion or transplantation or can be inherited (e.g., in plants). In the case of a bone marrow transplant, the relative amounts of donor and recipient cells can be used to determine if engraftment has taken place.
Chimerisms can be monitored via sensitive PCR-based methods and analysis of STR markers. The sensitivity, improved data quality, and precision of the 3500 Series Genetic Analyzers combined with our five-color chemistry and pre-optimized run conditions create the ideal solution for STR analysis.
Step-by-Step Guide to Chimerism
DNA Purification kit
DNA extraction is a critical first step in the experimental workflow of DNA Sequencing and Fragment analysis. The overall quality, accuracy and length of the DNA sequence read can be significantly affected by characteristics of the sample itself, and the method chosen for nucleic acid extraction. Ideal methods will vary depending on the source or tissue type, how it was obtained from its source, and how the sample was handled or stored prior to extraction.
Recommended Products: DNA Isolation
PCR Kit
Amplify microsatellite loci of interest by PCR using fluorescent labeled primer.
Recommended Products: Design Primer & Perform PCR
Fragment Analysis sizing graph image.
Add aliquot of PCR product to size standard and Hi-Di Formamide mix; heat denature and proceed with electrophoresis.
Recommended Products: Prepare Sample for Analysis
Sizing Information
To learn more about our GeneScan Size standards used in sizing experiments please see our table below.
When DNA fragments are labeled with: Choose a size standards labeled with: Recommended Size Standard Possible Applications Other Kits and Products
dR110, dR6G, dTAMRA™, dROX (Dye Set DS-02 - Filter E5) LIZ®
SNaPshot®
Microsatellite, LOH, ISSR, Chimerism, ISSR, RFLP, T-RFLP, MSMSA
5-FAM™, HEX™, NED™ (Dye Set DS-30 - Filter D) ROX™ GeneScan™ 500 ROX™ Size Standard Custom Fragment Analysis Custom Labeled Primer
6-FAM™, VIC®, NED™ (Dye Set DS-31 - Filter D**) ROX™ GeneScan™ 500 ROX™ Size Standard
Custom Fragment Analysis
Custom Labeled Primer
5-FAM™, JOE, NED™ (Dye Set DS-32 - Filter F) ROX™ GeneScan™ 500 ROX™ Size Standard
Microsatellite (Forensic)
AFLP
6-FAM™, VIC®, NED™, PET® (Dye Set DS-33 - Filter G5) LIZ® GeneScan™ 600 LIZ Size Standard v2.0
Microsatellite (Forensic)
Pathogen Detection
Custom Fragment Analysis
AmpFℓSTR® Kits,
TrueScience™ RespiFinder®
Custom Labeled Primer
Capillary Electrophoresis
During capillary electrophoresis, the products of the PCR are injected electrokinetically into capillaries filled with polymer. High voltage is applied so that the fluorescent DNA fragments are separated by size and are detected by a laser/camera system.
Which Electrophoresis Instrument (Genetic Analyzer) Is Right for You?
  Instrument
  3730xl
3730xL
3730
3730
3500xl
3500xL
3500
3500
3130xl
3130xL
3130
3130
310
310
Number of Capillaries 96 48 24 8 16 4 1
Compatible Applications:
(S) Supported; (A) AB Demonstrated; (C) Customer Demonstrated; (N) Not Supported
Microsatellite
Chimerism Studies C C C C C C C
Analyze the microsatellite profile. Appearance of numerous extra alleles in the microsatellite profile indicates a genome instability.
Which Data Analysis Software Is Right for You?
DNA Purification kit
DNA extraction is a critical first step in the experimental workflow of DNA Sequencing and Fragment analysis. The overall quality, accuracy and length of the DNA sequence read can be significantly affected by characteristics of the sample itself, and the method chosen for nucleic acid extraction. Ideal methods will vary depending on the source or tissue type, how it was obtained from its source, and how the sample was handled or stored prior to extraction.
Recommended Products: DNA Isolation
PCR Kit
Amplify microsatellite loci of interest by PCR using fluorescent labeled primer.
Recommended Products: Design Primer & Perform PCR
Fragment Analysis sizing graph image.
Add aliquot of PCR product to size standard and Hi-Di Formamide mix; heat denature and proceed with electrophoresis.
Recommended Products: Prepare Sample for Analysis
Sizing Information
To learn more about our GeneScan Size standards used in sizing experiments please see our table below.
When DNA fragments are labeled with: Choose a size standards labeled with: Recommended Size Standard Possible Applications Other Kits and Products
dR110, dR6G, dTAMRA™, dROX (Dye Set DS-02 - Filter E5) LIZ®
SNaPshot®
Microsatellite, LOH, ISSR, Chimerism, ISSR, RFLP, T-RFLP, MSMSA
5-FAM™, HEX™, NED™ (Dye Set DS-30 - Filter D) ROX™ GeneScan™ 500 ROX™ Size Standard Custom Fragment Analysis Custom Labeled Primer
6-FAM™, VIC®, NED™ (Dye Set DS-31 - Filter D**) ROX™ GeneScan™ 500 ROX™ Size Standard
Custom Fragment Analysis
Custom Labeled Primer
5-FAM™, JOE, NED™ (Dye Set DS-32 - Filter F) ROX™ GeneScan™ 500 ROX™ Size Standard
Microsatellite (Forensic)
AFLP
6-FAM™, VIC®, NED™, PET® (Dye Set DS-33 - Filter G5) LIZ® GeneScan™ 600 LIZ Size Standard v2.0
Microsatellite (Forensic)
Pathogen Detection
Custom Fragment Analysis
AmpFℓSTR® Kits,
TrueScience™ RespiFinder®
Custom Labeled Primer
Capillary Electrophoresis
During capillary electrophoresis, the products of the PCR are injected electrokinetically into capillaries filled with polymer. High voltage is applied so that the fluorescent DNA fragments are separated by size and are detected by a laser/camera system.
Which Electrophoresis Instrument (Genetic Analyzer) Is Right for You?
  Instrument
  3730xl
3730xL
3730
3730
3500xl
3500xL
3500
3500
3130xl
3130xL
3130
3130
310
310
Number of Capillaries 96 48 24 8 16 4 1
Compatible Applications:
(S) Supported; (A) AB Demonstrated; (C) Customer Demonstrated; (N) Not Supported
Microsatellite
Chimerism Studies C C C C C C C
Analyze the microsatellite profile. Appearance of numerous extra alleles in the microsatellite profile indicates a genome instability.
Which Data Analysis Software Is Right for You?
Step-by-Step Guide to Chimerism
DNA Purification kit
DNA extraction is a critical first step in the experimental workflow of DNA Sequencing and Fragment analysis. The overall quality, accuracy and length of the DNA sequence read can be significantly affected by characteristics of the sample itself, and the method chosen for nucleic acid extraction. Ideal methods will vary depending on the source or tissue type, how it was obtained from its source, and how the sample was handled or stored prior to extraction.
Recommended Products: DNA Isolation
PCR Kit
Amplify microsatellite loci of interest by PCR using fluorescent labeled primer.
Recommended Products: Design Primer & Perform PCR
Fragment Analysis sizing graph image.
Add aliquot of PCR product to size standard and Hi-Di Formamide mix; heat denature and proceed with electrophoresis.
Recommended Products: Prepare Sample for Analysis
Sizing Information
To learn more about our GeneScan Size standards used in sizing experiments please see our table below.
When DNA fragments are labeled with: Choose a size standards labeled with: Recommended Size Standard Possible Applications Other Kits and Products
dR110, dR6G, dTAMRA™, dROX (Dye Set DS-02 - Filter E5) LIZ®
SNaPshot®
Microsatellite, LOH, ISSR, Chimerism, ISSR, RFLP, T-RFLP, MSMSA
5-FAM™, HEX™, NED™ (Dye Set DS-30 - Filter D) ROX™ GeneScan™ 500 ROX™ Size Standard Custom Fragment Analysis Custom Labeled Primer
6-FAM™, VIC®, NED™ (Dye Set DS-31 - Filter D**) ROX™ GeneScan™ 500 ROX™ Size Standard
Custom Fragment Analysis
Custom Labeled Primer
5-FAM™, JOE, NED™ (Dye Set DS-32 - Filter F) ROX™ GeneScan™ 500 ROX™ Size Standard
Microsatellite (Forensic)
AFLP
6-FAM™, VIC®, NED™, PET® (Dye Set DS-33 - Filter G5) LIZ® GeneScan™ 600 LIZ Size Standard v2.0
Microsatellite (Forensic)
Pathogen Detection
Custom Fragment Analysis
AmpFℓSTR® Kits,
TrueScience™ RespiFinder®
Custom Labeled Primer
Capillary Electrophoresis
During capillary electrophoresis, the products of the PCR are injected electrokinetically into capillaries filled with polymer. High voltage is applied so that the fluorescent DNA fragments are separated by size and are detected by a laser/camera system.
Which Electrophoresis Instrument (Genetic Analyzer) Is Right for You?
  Instrument
  3730xl
3730xL
3730
3730
3500xl
3500xL
3500
3500
3130xl
3130xL
3130
3130
310
310
Number of Capillaries 96 48 24 8 16 4 1
Compatible Applications:
(S) Supported; (A) AB Demonstrated; (C) Customer Demonstrated; (N) Not Supported
Microsatellite
Chimerism Studies C C C C C C C
Analyze the microsatellite profile. Appearance of numerous extra alleles in the microsatellite profile indicates a genome instability.
Which Data Analysis Software Is Right for You?
DNA Purification kit
DNA extraction is a critical first step in the experimental workflow of DNA Sequencing and Fragment analysis. The overall quality, accuracy and length of the DNA sequence read can be significantly affected by characteristics of the sample itself, and the method chosen for nucleic acid extraction. Ideal methods will vary depending on the source or tissue type, how it was obtained from its source, and how the sample was handled or stored prior to extraction.
Recommended Products: DNA Isolation
PCR Kit
Amplify microsatellite loci of interest by PCR using fluorescent labeled primer.
Recommended Products: Design Primer & Perform PCR
Fragment Analysis sizing graph image.
Add aliquot of PCR product to size standard and Hi-Di Formamide mix; heat denature and proceed with electrophoresis.
Recommended Products: Prepare Sample for Analysis
Sizing Information
To learn more about our GeneScan Size standards used in sizing experiments please see our table below.
When DNA fragments are labeled with: Choose a size standards labeled with: Recommended Size Standard Possible Applications Other Kits and Products
dR110, dR6G, dTAMRA™, dROX (Dye Set DS-02 - Filter E5) LIZ®
SNaPshot®
Microsatellite, LOH, ISSR, Chimerism, ISSR, RFLP, T-RFLP, MSMSA
5-FAM™, HEX™, NED™ (Dye Set DS-30 - Filter D) ROX™ GeneScan™ 500 ROX™ Size Standard Custom Fragment Analysis Custom Labeled Primer
6-FAM™, VIC®, NED™ (Dye Set DS-31 - Filter D**) ROX™ GeneScan™ 500 ROX™ Size Standard
Custom Fragment Analysis
Custom Labeled Primer
5-FAM™, JOE, NED™ (Dye Set DS-32 - Filter F) ROX™ GeneScan™ 500 ROX™ Size Standard
Microsatellite (Forensic)
AFLP
6-FAM™, VIC®, NED™, PET® (Dye Set DS-33 - Filter G5) LIZ® GeneScan™ 600 LIZ Size Standard v2.0
Microsatellite (Forensic)
Pathogen Detection
Custom Fragment Analysis
AmpFℓSTR® Kits,
TrueScience™ RespiFinder®
Custom Labeled Primer
Capillary Electrophoresis
During capillary electrophoresis, the products of the PCR are injected electrokinetically into capillaries filled with polymer. High voltage is applied so that the fluorescent DNA fragments are separated by size and are detected by a laser/camera system.
Which Electrophoresis Instrument (Genetic Analyzer) Is Right for You?
  Instrument
  3730xl
3730xL
3730
3730
3500xl
3500xL
3500
3500
3130xl
3130xL
3130
3130
310
310
Number of Capillaries 96 48 24 8 16 4 1
Compatible Applications:
(S) Supported; (A) AB Demonstrated; (C) Customer Demonstrated; (N) Not Supported
Microsatellite
Chimerism Studies C C C C C C C
Analyze the microsatellite profile. Appearance of numerous extra alleles in the microsatellite profile indicates a genome instability.
Which Data Analysis Software Is Right for You?
Highlighted products for Chimerism Analysis:
GeneMapper® Software
Verti Thermal Cycler
Visistat reference component
Resources
Interested in commercializing your assay?
We can help ›
Support
New Capillary Electrophoresis Instruments Support Center
Find tips, troubleshooting help, and resources for support related to your capillary electrophoresis instruments.
Fragment Analysis Software Support Center
Visistat reference component