Chip Expected Sequencing Run Time Expected Output* Expected Reads
  200-base reads 400-base reads 200-base reads 400-base reads  
Ion 314™ Chip v2
2.3 hr 3.7 hr 30-50 Mb 60-100 Mb 400-550 thousand
Ion 316™ Chip v2
3.0 hr 4.9 hr 300-600 Mb 600 Mb-1 Gb 2-3 million
Ion 318™ Chip v2
4.4 hr 7.3 hr 600 Mb-1 Gb 1.2-2 Gb 4-5.5 million
*Expected output with >99% aligned/measured accuracy.  Output is dependent on read length and application.
Specifications for the Ion PGM™ Sequencer
Working environment [for indoor use only] Temperature: 68–77°F (20–25°C)
Humidity: 40–60% non-condensing
Altitude: <6,500 ft (2,000 m)
Clearances:
12 in (30.5 cm) in rear
4 in (10 cm) on left side
4 in (10 cm) on right side
4 in (10 cm) from front edge of bench to sequencer bezel
8 in (20 cm) from front edge of bench to the conical tubes
36 in (90 cm) aisle in front of bench for operator access
Gas supply Connection: 0.25 in push-to-connect fitting
Pressure: 35–45 psi
Composition: Nitrogen (grade 4.5, 99.995% or better)
Other connections Ethernet: 1GigE
USB: 2x USB 2.0
Power Voltage: 100 V (min) to 240 V (max)
Current: 9 A (max)
Frequency: 50/60 Hz
Power draw: 200-300 W
Dimensions Width: 24 in (61 cm)
Depth: 20 in (51 cm)
Height: 21 in (53 cm)
Weight Crated for shipment: 85 lb (39 kg)
Free-standing: 65 lb (30 kg)
Additional Equipment PGM™ Torrent Server
Ion Chef™ System
Ion OneTouch™ System
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
Visistat reference component