icon-reproductive-genomics-pgs-showcase-image-230x195
Aneuploidy can now be detected via next-generation sequencing using DNA from a single or multiple cells isolated from preimplantation embryos. The Ion S5™ and S5™ XL Systems offer a simple and scalable workflow for aneuploidy analysis. Through the use of whole genome amplification (WGA), genomic DNA from a small number of cells—even a single cell— can be used for low-pass sequencing of the entire genome. Following that, aneuploidy status may be determined using Ion Reporter™ Software. To get started, download the appropriate user guides from the table below.
Sequencing workflow for aneuploidy analysis
Application
1. Prepare library
2. Prepare template
3. Run sequence
4. Analyze data
Aneuploidy analysis
Whole-genome amplification and fragment library construction with Ion SingleSeq kits included in Ion ReproSeq PGS kits.
Ion ReproSeq PGS kits user guide
Isothermal amplification with Ion Chef System.
Ion 520 & 530 Kit-Chef user guide
Sequence up to 96 pre-implantation embryo biopsy samples with Ion 530 Chip.
Ion S5 and Ion S5 XL Systems user guide
Data analysis and storage with Torrent Suite Software and Ion Reporter Software and Server.
Torrent Suite Software 5.4 help guide
Ion Reporter Software help guide
Documents and resources
Literature
User guides and FAQs
Support
Learn more
Training, events, and webinars