HiCEP workflowEnlarge Image
Figure 1:HiCEP workflow, click to enlarge.
The HiCEP method is an amplified fragment length polymorphism (AFLP)-based gene expression profiling method that requires no prior sequence information and has a reduced rate of false positives and a high degree of detection of both coding and non-coding transcripts.
Whole transcriptome expression profiling is historically performed using hybridization-based microarray methods. There are a number of limitations to microarray-based approaches such as low sensitivity and specificity, poor dynamic range, as well as a need for prior sequence information. The HiCEP method was developed to address the shortcomings in gene expression profiling and provide a sensitive method for detecting a large proportion of transcripts in both known and unknown genes, with a low false positive rate.
Step-by-Step Guide to High Coverage Gene Expression Profiling
PCR Kit
Amplify the DNA region of interest.
Recommended Products:
Fragment Analysis sizing graph image.
Add aliquot of PCR product to size standard and Hi-Di Formamide mix; heat denature and proceed with electrophoresis.
Recommended Products: Prepare Sample for Analysis
Sizing Information
To learn more about our GeneScan Size standards used in sizing experiments please see our table below.
When DNA fragments are labeled with: Choose a size standards labeled with: Recommended Size Standard Possible Applications Other Kits and Products
dR110, dR6G, dTAMRA™, dROX (Dye Set DS-02 - Filter E5) LIZ®
SNaPshot®
Microsatellite, LOH, ISSR, Chimerism, ISSR, RFLP, T-RFLP, MSMSA
5-FAM™, HEX™, NED™ (Dye Set DS-30 - Filter D) ROX™ GeneScan™ 500 ROX™ Size Standard Custom Fragment Analysis Custom Labeled Primer
6-FAM™, VIC®, NED™ (Dye Set DS-31 - Filter D**) ROX™ GeneScan™ 500 ROX™ Size Standard
Custom Fragment Analysis
Custom Labeled Primer
5-FAM™, JOE, NED™ (Dye Set DS-32 - Filter F) ROX™ GeneScan™ 500 ROX™ Size Standard
Microsatellite (Forensic)
AFLP
6-FAM™, VIC®, NED™, PET® (Dye Set DS-33 - Filter G5) LIZ® GeneScan™ 600 LIZ Size Standard v2.0
Microsatellite (Forensic)
Custom Fragment Analysis
Analyze the HiCEP profile.
Which Data Analysis Software Is Right for You?
Fragment Analysis sizing graph image.
Add aliquot of PCR product to size standard and Hi-Di Formamide mix; heat denature and proceed with electrophoresis.
Recommended Products: Prepare Sample for Analysis
Sizing Information
To learn more about our GeneScan Size standards used in sizing experiments please see our table below.
When DNA fragments are labeled with: Choose a size standards labeled with: Recommended Size Standard Possible Applications Other Kits and Products
dR110, dR6G, dTAMRA™, dROX (Dye Set DS-02 - Filter E5) LIZ®
SNaPshot®
Microsatellite, LOH, ISSR, Chimerism, ISSR, RFLP, T-RFLP, MSMSA
5-FAM™, HEX™, NED™ (Dye Set DS-30 - Filter D) ROX™ GeneScan™ 500 ROX™ Size Standard Custom Fragment Analysis Custom Labeled Primer
6-FAM™, VIC®, NED™ (Dye Set DS-31 - Filter D**) ROX™ GeneScan™ 500 ROX™ Size Standard
Custom Fragment Analysis
Custom Labeled Primer
5-FAM™, JOE, NED™ (Dye Set DS-32 - Filter F) ROX™ GeneScan™ 500 ROX™ Size Standard
Microsatellite (Forensic)
AFLP
6-FAM™, VIC®, NED™, PET® (Dye Set DS-33 - Filter G5) LIZ® GeneScan™ 600 LIZ Size Standard v2.0
Microsatellite (Forensic)
Custom Fragment Analysis
Analyze the HiCEP profile.
Which Data Analysis Software Is Right for You?
Step-by-Step Guide to High Coverage Gene Expression Profiling
PCR Kit
Amplify the DNA region of interest.
Recommended Products:
Fragment Analysis sizing graph image.
Add aliquot of PCR product to size standard and Hi-Di Formamide mix; heat denature and proceed with electrophoresis.
Recommended Products: Prepare Sample for Analysis
Sizing Information
To learn more about our GeneScan Size standards used in sizing experiments please see our table below.
When DNA fragments are labeled with: Choose a size standards labeled with: Recommended Size Standard Possible Applications Other Kits and Products
dR110, dR6G, dTAMRA™, dROX (Dye Set DS-02 - Filter E5) LIZ®
SNaPshot®
Microsatellite, LOH, ISSR, Chimerism, ISSR, RFLP, T-RFLP, MSMSA
5-FAM™, HEX™, NED™ (Dye Set DS-30 - Filter D) ROX™ GeneScan™ 500 ROX™ Size Standard Custom Fragment Analysis Custom Labeled Primer
6-FAM™, VIC®, NED™ (Dye Set DS-31 - Filter D**) ROX™ GeneScan™ 500 ROX™ Size Standard
Custom Fragment Analysis
Custom Labeled Primer
5-FAM™, JOE, NED™ (Dye Set DS-32 - Filter F) ROX™ GeneScan™ 500 ROX™ Size Standard
Microsatellite (Forensic)
AFLP
6-FAM™, VIC®, NED™, PET® (Dye Set DS-33 - Filter G5) LIZ® GeneScan™ 600 LIZ Size Standard v2.0
Microsatellite (Forensic)
Custom Fragment Analysis
Analyze the HiCEP profile.
Which Data Analysis Software Is Right for You?
Fragment Analysis sizing graph image.
Add aliquot of PCR product to size standard and Hi-Di Formamide mix; heat denature and proceed with electrophoresis.
Recommended Products: Prepare Sample for Analysis
Sizing Information
To learn more about our GeneScan Size standards used in sizing experiments please see our table below.
When DNA fragments are labeled with: Choose a size standards labeled with: Recommended Size Standard Possible Applications Other Kits and Products
dR110, dR6G, dTAMRA™, dROX (Dye Set DS-02 - Filter E5) LIZ®
SNaPshot®
Microsatellite, LOH, ISSR, Chimerism, ISSR, RFLP, T-RFLP, MSMSA
5-FAM™, HEX™, NED™ (Dye Set DS-30 - Filter D) ROX™ GeneScan™ 500 ROX™ Size Standard Custom Fragment Analysis Custom Labeled Primer
6-FAM™, VIC®, NED™ (Dye Set DS-31 - Filter D**) ROX™ GeneScan™ 500 ROX™ Size Standard
Custom Fragment Analysis
Custom Labeled Primer
5-FAM™, JOE, NED™ (Dye Set DS-32 - Filter F) ROX™ GeneScan™ 500 ROX™ Size Standard
Microsatellite (Forensic)
AFLP
6-FAM™, VIC®, NED™, PET® (Dye Set DS-33 - Filter G5) LIZ® GeneScan™ 600 LIZ Size Standard v2.0
Microsatellite (Forensic)
Custom Fragment Analysis
Analyze the HiCEP profile.
Which Data Analysis Software Is Right for You?
Highlighted products for High Coverage Gene Expression Profiling:
Verti Thermal Cycler
3500 Series Genetic Analyzers
GeneMapper® Software
Visistat reference component