Trade in your 310 or 3100 Series genetic analyzer for a new SeqStudio Genetic Analyzer System and simplify your workflow.*
Same workflow, same trusted technology—now with an innovative all-in-one cartridge that reduces setup time from hours to minutes. The Applied Biosystems SeqStudio Genetic Analyzer System is a low-throughput, easy-to-use, convenient benchtop system that is designed to deliver gold-standard Sanger sequencing technology and fragment analysis with just a simple click. It is easily used across a broad range of applications as well as multiple users.
Just click. That's it.
Load up your samples, click in the cartridge, and go—no need for technical experts for setup or maintenance, no calibration before each run, and no installation of capillaries, polymer, and anode buffer. The cartridge is stable for four months on the instrument.
Access your data anytime, anywhere**
This cloud-enabled system is integrated with Thermo Fisher Connect so you can set up your run, check its progress, and access your data from wherever you have Internet access, all without the need to stay in the lab.** Cloud-based Applied Biosystems Sanger Analysis modules are available for secondary analysis. Get the same data quality, service, and support you’ve come to expect from Applied Biosystems technology with a modernized experience at a price any lab can afford.
Convenient and easy-to-use cartridge system includes the capillaries, polymer, and buffers required for each run
Combine Sanger sequencing and fragment analysis reactions in the same instrument run
Fast turnaround with a run time in as little as 30 minutes
Set up and monitor your run. View, manage, and share your data from anywhere, using your phone, tablet, or computer (Apple™ computer or PC).**
1-year instrument warranty included, and in-lab application training will get you up and running quickly
Secondary analysis software package included with system purchase
*Offer valid for end-user customers in the US (excluding Puerto Rico) and Canada. Suppliers of Life Science products are not eligible to participate in this promotion. Determination of eligibility is at the sole discretion of Life Technologies. Trade-in any 310, 3100, or 3130 Series genetic analyzer for a discount off the list price of a new Applied Biosystems SeqStudio Genetic Analyzer System. Discount will apply to the list price in effect at the time the order is received by Life Technologies. Discount will vary based on the type of instrument traded in. Offer valid on qualifying orders received no later than October 31, 2018.  Customer may trade-in and purchase at a discount a maximum of 10 instruments during the promotion period.  Trade-ins and purchased instruments must be submitted on a one-to-one trade to purchase basis to be considered eligible for the program. Customer must request a trade-in at the time the order for the new instrument is placed.  This promotion is available only for purchases of new eligible instruments.  Trade-in instruments must be returned to Life Technologies within 30 days following installation of the new instrument. Life Technologies will be responsible for shipping costs on the traded-in instrument.  Where necessary, customer agrees to allow a Life Technologies Field Service representative to de-install the instrument from their facilities, and charges for such deinstallation will be paid for by Life Technologies..  Prior to return, Customer will fully clean the Customer’s trade-in unit and decontaminate it of radioactive, biological, toxic, and other dangerous or hazardous materials or substances and, if requested by Life Technologies, Customer agrees to complete and return a Certificate of Decontamination to Life Technologies. Cannot be combined with other discounts or promotions.  Offer void where prohibited by federal, state, provincial, or local laws or regulation, or agency/institutional policy.  Other restrictions may apply.
**Internet connection and Thermo Fisher Cloud account required.
Support
New Capillary Electrophoresis Instruments Support Center
Find tips, troubleshooting help, and resources for support related to your capillary electrophoresis instruments.
Visistat reference component