Complementary technologies for seamless workflows
Cover a wide range of sequencing spectrum and research applications with the Applied Biosystems SeqStudio Genetic Analyzer for Sanger sequencing and the Ion GeneStudio S5 Series for next-generation sequencing.
No matter which sequencing instrument you choose, we offer optimized reagents and consumables, intuitive analysis software, and world-class technical support.
Sequencing platforms SeqStudio System for Sanger sequencing
Applied Biosystems 3500 System for Sanger sequencing
Ion GeneStudio S5
Series for NGS
Description
Easy-to-use platform, combined with trusted Sanger sequencing results
The gold-standard sequencing platform for fast, flexible analysis
Simple, fast, and flexible  benchtop sequencers for cancer, inherited disease, reproductive health, and microbial research
Benefits
Flexible—combine Sanger sequencing and fragment analysis on the same plate
Easy to use—all-in-one cartridge takes setup from hours to minutes
Fast—runs in as little as 30 minutes
Connected—access your data anytime, anywhere*
*With internet connection and Thermo Fisher Cloud account
Accuracy—the ultimate sequencing verification tool
Speed and flexibility—run from 1 to 96 samples typically in <5 hours
Read length— highest read length capability, up to 1,000 bp
Flexibility—3 different instruments and 5 sequencing chips for flexible throughput and turnaround time
Simplicity— automated workflow solutions
Scalability— from 2M to 130M reads per run
Speed—DNA to data in as little as 24 hours with less than 45 min of hands-on time
Sequencing applications
Confirmation of NGS results
Plasmid sequencing
CRISPR-Cas9 genome editing analysis
Human cell line authentication
Applied Biosystems SNaPshot genotyping
Multiplex ligation–dependent analysis of human copy number variation
Confirmation of NGS results
Targeted amplicon resequencing
Bacterial, fungal, and viral identification
Fragment analysis
Microsatellite/STR analysis
Targeted RNA and DNA
Microbial
Transcriptome
Exome
RNA-seq
  Learn more › Learn more › Learn more ›
Visistat reference component
Visistat reference component