Stem Cell Engineering
Manipulating cells for downstream applications
Cellular engineering requires introduction of new genetic content into a cell. One specific type of engineering is reprogramming by conversion of somatic cells into induced pluripotent stem cells. This can be accomplished with Thermo Fisher’s non-integrating technologies by viral delivery or episomal plasmids. Once cell lines have been established, genomic editing and manipulation of iPSCs into other cell types allows for the building of cell models to better answer biological questions.
Request information
Reprogramming
CytoTune-iPS 2.0 Sendai Reprogramming Kit
Highest-efficiency, integration-free system
Epi5 Episomal iPSC Reprogramming Vectors
High efficiency viral-free system
Genome editing
CRISPR Products and Services
Consistently high editing efficiency across all gene targets and cell types.
TALEN-Based Genome Editing
Precise and flexible editing; targeting to any gene in any cell, with all-around freedom.
StemFlex Medium
Supports up to 2-fold faster recovery following gene editing
Services
CellModel Services
Reprogrammed cells provided in as little as 4 to 6 months
GeneArt Genes-to-Cell Lines Services
Allow us to generate high-performance cell lines with stable expression of your gene
Stem cell reprogramming
Reprogramming somatic cells to induced pluripotent stem cells is a critical and potentially time-intensive step in stem cell research. We offer choices in integration-free reprogramming technologies and services to support your research goals.
Learn more about:
Genome editing
From precision genome editing and gene modification technologies to high-efficiency delivery systems, we have developed a broad range of solutions to help you create the modified genes, expression systems, and stable cell lines you need for your research—from culturing cells to modification, then detection and analysis.
Learn more about:
Additional tools for modifying stem cells
In addition to innovative technologies and services for reprogramming and genomic editing of stem cells, we offer tools for cell transfection, transduction, and cloning.
Learn more about:
  • Neon Transfection System—Electroporation technology for highly efficient delivery of nucleic acids into virtually any animal cell type. The system can be used for reprogramming somatic cells into iPSCs.
  • Gateway cloning—Circumvent traditional restriction enzyme–based cloning limitations; Gateway cloning enables access to virtually any expression system.
  • BacMam technology—A baculovirus delivery system for efficient transduction of mammalian cell lines, including primary and stem cells.
  • Lipofectamine 3000—Advanced lipid nanoparticle technology for superior transfection efficiency in a broad spectrum of difficult-to-transfect cell types. This reagent kit can be used for generation and transfection of iPSCs.
Technical resources
Support