TaqMan hPSC Scorecard Panel Instrument Requirements
Several Applied Biosystems real-time PCR platforms are compatible with the TaqMan hPSC Scorecard Panel. If you already have any ofthese instruments installed you just need to download the template file (.edt or .sdt), import it into your instrument's data collection software, and then begin your experiment:
  • QuantStudio 12K Flex
  • ViiA 7
  • StepOnePlus Real-Time PCR System
  • QuantStudio 6/7 Flex Real-Time PCR system
  • 7500 Fast Real-Time PCR System
  • 7900HT Fast Real-Time PCR System
If you are less familiar with the Applied Biosystems real-time PCR platforms, we are happy to help you choose the best instrument for your laboratory’s needs, locate a nearby instrument which you may be able to use, or we can run the hPSC Scorecard assay for you with our CellModel Characterization Services.
Request a demo
Need help? Have a question? Find support resources now ›
QuantStudio 12K FlexReal-Time PCR System
Maximum throughput, flexibility and scalability
The QuantStudio 12K Flex is fully compatible with both the 96-well and 384-well TaqMan hPSC Scorecard panels, allowing the processing of up to 4 samples in a single run.
Learn more ›
StepOnePlus Real-Time PCR System
Easy to use for all levels of experience
The 96-well format is easy to use and ideal for researchers running only a few samples at a time. The 96-well fast cycling block enables data collection in little as 30 mins.
Learn more ›
ViiA 7 Real-Time PCR System
High flexibility and intuitive
design
The ViiA 7 system is fully compatible with both the 96-well and 384-well TaqMan hPSC Scorecard panels, allowing the processing of up to 4 samples in a single run.
Learn more ›
QuantStudio 6 Flex Real-Time PCR system
High flexibility & intuitive
design
The QuantStudio 6 Flex Real-Time PCR System is the ideal instrument for laboratories with project requirements that go beyond a single application or project and need to accommodate the needs of many on a limited budget.
Learn more ›
QuantStudio 7 Flex Real-Time PCR system
High versatility & intuitive
design
The QuantStudio 7 Flex Real-Time PCR System delivers the proven reliability, sensitivity and accuracy of the ViiA 7 System in a new industrial design.
Learn more ›
7500 Fast Real-Time PCR System
Trusted and fast performance & dye – versatility
Applied Biosystems 7500 Real-Time PCR Systems offer you high performance, multicolor real-time PCR wherever your research takes you. Choose from our Fast and standard 7500 systems.
Learn more ›
7500 Fast Real-Time PCR System
Trusted and fast performance & dye – versatility
Applied Biosystems 7500 Real-Time PCR Systems offer you high performance, multicolor real-time PCR wherever your research takes you. Choose from our Fast and standard 7500 systems.
Learn more ›
Cell model services
Access the information that the hPSC Scorecard Panel provides without access to the instrument. Our CellModel Services offer a convenient alternative to obtain the same data, without the need for an instrument.
Set up for hPSC Scorecard data collection in 3 easy steps!
1. Load your template file
Download the corresponding template file below and open it in the instrument software. The template file contains all of the necessary cycling parameters and sample names to save you time and make sure the instrument is set up correctly.
Example of opening new template file on ViiA 7 Real-Time PCR System
Figure 1. Example of opening new template file on ViiA 7 Real-Time PCR System.  Click on browse from the “select experiment template” area and open template file for TaqMan hPSC Scorecard Panel.
2. Insert your plate and Go!
Insert your TaqMan hPSC Scorecard plate into the instrument and start the run.
Inserting a TaqMan hPSC Scorecard plate into the instrument
Figure 2. Inserting a TaqMan hPSC Scorecard plate into the instrument.
3. Save your data and analyze
After the run is complete, just save your data in a desired location, and you are done. All of the data analysis can be performed by the TaqMan hPSC Scorecard Data Analysis software.
Screenshot of TaqMan hPSC Scorecard Data Analysis software
Figure 3. Screenshot of TaqMan hPSC Scorecard Data Analysis software.
Compatible instrument set-up and support
Check out the instrument set-up guides for your instrument for detailed instruction (with screenshots) on how to read the TaqMan hPSC Scorecard panel. Save Zip file to computer and unzip.
Need more information?
Need help choosing a PCR instrument?  See detailed information here.
Contact us with questions about setting up your instrument or supporting the hPSC Scorecard Panel
Request a demo