Oligonucleotides custom-manufactured to your specifications. Choose from dual-labeled Applied Biosystems TaqMan™ probes, unlabeled oligos for use as PCR primers, 5´-labeled fluorescent primers, and primer pairs for use in CE (Sanger) sequencing.
Custom Primers and TaqMan Probes
Applied Biosystems TaqMan MGB (minor groove binder), TAMRA, and QSY custom probes are dual-labeled and can be used in all of your real-time PCR research applications.
Features include:
  • Choice of delivered scales
  • Suitable for use in any real-time PCR application
  • HPLC purification of all TaqMan probes
Which probe is right for your application?
Custom TaqMan MGB probes Best all-around probes for most applications
Custom TaqMan TAMRA probes Good for maintaining consistency in projects in which TAMRA
Custom TaqMan QSY probes Best for multiplexing applications when 3 or more dyes are needed
Order custom probes
TaqMan probes and primers are available together as predesigned TaqMan Assays
Learn more about multiplex real-time PCR
Custom 5´labeled primers are fluorescently labeled oligos with a choice of dye on the 5´ end. These products can be used for fragment analysis, gene expression studies, PCR, and genotyping research.
Features include:
  • Choice of delivered scales
  • All oligos are desalted. Larger scales of labeled primers have an option for HPLC purification
  • IMPORTANT: HPLC purification reduces the yield in half
Catalog # Size HPLC yield 5' Reporter dye Quantity Order
450007 10,000 pmol N/A 6-FAM, TET, VIC, HEX, NED, PET 1 tube Order now
450006 80,000 pmol 40,000 pmol minimum 6-FAM, TET, VIC, HEX, NED, PET 1 tube Order now
450017 300,000 pmol 150,000 pmol minimum 6-FAM, TET, VIC, HEX, NED, PET 1 tube Order now
Specifications
Format Delivered either in solution (either 1X TE or water) or dried down
Purification Desalted; option of HPLC purification with larger scale
Shelf life 12 months from manufacturing date
Shipping condition Room temperature
Storage condition –20°C
Custom 5´labeled primer pairs include a fluorescently labeled oligo with a choice of dye on the 5´ end, along with an unlabeled oligo, in each pair. These products can be used in a variety of research applications.
Features include:
  • Choice of delivered scales
  • For use in fragment analysis, gene expression studies, genotyping and PCR
  • All oligos are desalted
Catalog # Size HPLC yield 5' Reporter dye Quantity Order
450056 10,000 pmol 5´ Labeled/Unlabeled Primer Pairs 6-FAM, TET, VIC, HEX, NED, PET 2 tubes Order now
450059 80,000 pmol 5´ Labeled/Unlabeled Primer Pairs 6-FAM, TET, VIC, HEX, NED, PET 2 tubes Order now
450062 300,000 pmol 5´ Labeled/Unlabeled Primer Pairs 6-FAM, TET, VIC, HEX, NED, PET 2 tubes Order now
4304976 10,000 pmol 5´ Labeled/Unlabeled Di-Repeats 6-FAM, TET, VIC, HEX, NED, PET 2 tubes Order now
4304977 80,000 pmol 5´Labeled/Unlabeled Di-Repeats 6-FAM, TET, VIC, HEX, NED, PET 2 tubes Order now
4304978 300,000 pmol 5´ Labeled/Unlabeled Di-Repeats 6-FAM, TET, VIC, HEX, NED, PET 2 tubes Order now
4304979 10,000 pmol 5´ Labeled/Unlabeled Di-Repeat + Tail 6-FAM, TET, VIC, HEX, NED, PET 2 tubes Order now
4304981 80,000 pmol 5´ Labeled/Unlabeled Di-Repeat + Tail 6-FAM, TET, VIC, HEX, NED, PET 2 tubes Order now
4304982 300,000 pmol 5´ Labeled/Unlabeled Di-Repeat + Tail 6-FAM, TET, VIC, HEX, NED, PET 2 tubes Order now
Specifications
Format Delivered either in solution (either 1X TE or water) or dried down
Purification Desalted; option of HPLC purification with larger scale
Shelf life 12 months from manufacturing date
Shipping condition Room temperature
Storage condition –20°C
Sequence detection primers are unlabeled primers that can be used with TaqMan probes or SYBR Green dye for your real-time PCR research applications.
Features include:
  • Choice of delivered scales
  • For use in any real-time PCR or PCR application
  • All sequence detection primers are desalted
Real-time PCR sequence detection primers are delivered either in solution or dried.
Catalog # Size Quantity Order
4304970 10,000 pmol 1 tube Order now
4304971 80,000 pmol 1 tube Order now
4304972 130,000 pmol 1 tube Order now
Specifications
Format Delivered either in solution (either 1X TE or water) or dried down
Purification Desalted
Shelf life 12 months from manufacturing date
Shipping condition Room temperature
Storage condition –20°C
We believe it is our responsibility to provide our customers with sustainable product options. These products and/or their packaging incorporate one or more environmental green features. Learn more ›
Support
  • Order support
    Contact order & web support, search FAQs, view order history and track your order
  • Technical support
    Contact a technical application scientist, instrument maintenance & repair requests and search product documents & FAQs