Thermo Fisher Scientific's biochemical assay development services are designed to create and validate an assay that meets your discovery or screening needs. Work with our dedicated team of experts to custom-design an assay that fits your workflow.
Send us an inquiry ›
Designing your custom assay
All you need to design your custom biochemical assay is your target of interest and an understanding of what information you wish to gather by studying your target of interest. Our dedicated team of experts will work with you to understand your challenges and suggest potential solutions.
For more information on custom assay development projects, submit an online inquiry.
Assay technologies to address your needs
Technology Benefit Application
LanthaScreen™ Eu Kinase Binding Assay Ideal assay for the study of non-activated, low-activity kinase targets, or kinases with no known substrate due to direct binding readout. Real-time readout makes this assay ideal for analysis of binding kinetics. Primary screening or profiling
LanthaScreen Kinase Activity Assay Ideal primary screening technology due to lack of compound interference Primary screening
Z´-LYTE™ FRET Assay Ideal for secondary screening, hit validation, and profiling because this technology has been validated with >250 kinases; it is the primary technology used in our SelectScreen™ Kinase Profiling Service. Secondary screening, hit validation, and profiling
Adapta™ Universal Kinase Assay Ideal for screening projects when the target of interest does not phosphorylate traditional peptide substrates. Primary or secondary screening
Custom Biology Services team
With many labs facing resources restraints, our Custom Biology team offers institutions a powerful resource to extend their research teams. Wholly dedicated to developing custom tools and assays, the team has the expertise to select an assay technology from the extensive Thermo Fisher Scientific toolbox and use these technologies to craft tools that meet your unique requirements.  
Dedicated project management
From your initial inquiry through every milestone of your project, you can expect exceptional service and frequent, proactive communication from a dedicated project manager. Your project manager will help ensure all parties involved in the project are aligned on project requirements, timelines and budgets.
For more information on our custom cell engineering services, send us an inquiry online. A Thermo Fisher Scientific representative will respond to your inquiry within two business days.