Table 1.8 Amine-reactive xanthene derivatives.
Fluorophore (Abs/Em) * COOH Succinimidyl Ester Other Protein and Nucleic Acid Labeling Kits Notes
Eosin (524/544)     E18 (ITC)5  
  • Useful for DAB photoconversion
  • Phosphorescent
Fluorescein (494/518) C1359 5
C1360 6
C1904 M
C2210 5
C6164 6
C1311 M
F6106 (X)6
F2181 (X)M
F6129 (X)M
F6130 (EX)5
C20050 (C)5
F143 (ITC)5
F1906 (ITC)5
F1907 (ITC)5
D16 (DTA)5
F6433 (F)
F6434 (F)
F10240 (P)
Z25042 (Z)
Z25342 (Z)
  • Most widely used green-fluorescent labeling dye
  • Absorption overlaps the 488 nm spectral line of the argon-ion laser
  • Prone to photobleaching
  • pH-sensitive spectra between pH 5 and pH 8
  • Common donor in FRET applications that utilize tetramethylrhodamine as the acceptor
JOE (520/548)   C6171MP 6    
  • Traditional fluorophore used in automated DNA sequencing
  • pH-insensitive spectra at pH >6
Oregon Green 488 (496/524) O6146 5 O6147 5
O6149 6
O6185 (X)6
O6080 M F6153 (F)
O10241 (P)
A10476 (APEX)
U21659 (U)
  • Photostable fluorescein substitute
  • pH-insensitive spectra at pH >6
  • Recognized by anti-fluorescein antibodies
Oregon Green 514 (511/530) O6138 M O6139 6    
  • Photostable fluorescein substitute
  • pH-insensitive spectra at pH >6
Rhodamine Green (502/527)   R6107 M
R6113 (X)M
  • Photostable fluorescein substitute
  • pH-insensitive spectra
* The numbers in parentheses reflect the absorption (Abs) and fluorescence emission (Em) maxima, in nm, of the goat anti–mouse IgG antibody or dextran conjugates in aqueous buffer.
Superscripts 5, 6 and M correspond to 5-isomer, 6-isomer and a mixture of 5- and 6-isomers, respectively.
(APEX) = APEX Antibody Labeling Kit (Kits for Labeling Proteins and Nucleic Acids—Section 1.2).
(C) = Caged; the probe is nonfluorescent until the caging group is removed by UV illumination.
(COOH) = Carboxylic acid.
(DTA) = Dichlorotriazine.
(EX) = A seven-atom spacer that is more hydrophilic than X.
(F) = FluoReporter Protein Labeling Kit (Kits for Labeling Proteins and Nucleic Acids—Section 1.2).
(ITC) = Isothiocyanate.
(P) = Easy-to-Use Protein Labeling Kit (Kits for Labeling Proteins and Nucleic Acids—Section 1.2).
(U) = ULYSIS Nucleic Acid Labeling Kit (Labeling Oligonucleotides and Nucleic Acids—Section 8.2).
(X) = Aminohexanoyl spacer separating the dye and SE.
(Z) = Zenon Antibody Labeling Kit (Zenon Technology: Versatile Reagents for Immunolabeling—Section 7.3).