Anti-Fluorophore and Anti-Hapten Antibodies—Table 7.8
Learn more in The Molecular Probes® Handbook
Table 7.8 Anti-fluorophore and anti-hapten antibodies.
| Cat. No. | Anti-Fluorophore | Host | Label |
|---|---|---|---|
| Anti-Fluorophore Antibodies and Their Conjugates | |||
| A5760 | Alexa Fluor 405/Cascade Blue dye | Rabbit | None |
| A11094 | Alexa Fluor 488 dye | Rabbit | None |
| A5770 | BODIPY FL dye | Rabbit | None |
| A6398 | Dansyl | Rabbit | None |
| A6421 | Fluorescein/Oregon Green dye | Mouse (clone 4-4-20) | None |
| A889 | Fluorescein/Oregon Green dye | Rabbit | None |
| A6413 | Fluorescein/Oregon Green dye | Rabbit, Fab fragment | None |
| A11095 | Fluorescein/Oregon Green dye | Goat | None |
| A11090 | Fluorescein/Oregon Green dye | Rabbit | Alexa Fluor 488 dye |
| A11096 | Fluorescein/Oregon Green dye | Goat | Alexa Fluor 488 dye |
| A11091 | Fluorescein/Oregon Green dye | Rabbit | Alexa Fluor 594 dye |
| A21250 | Fluorescein/Oregon Green dye | Rabbit | R-phycoerythrin |
| A982 | Fluorescein/Oregon Green dye | Rabbit | Biotin-XX |
| A21253 | Fluorescein/Oregon Green dye | Rabbit | Horseradish peroxidase |
| A5750 | Lucifer yellow dye | Rabbit | None |
| A5751 | Lucifer yellow dye | Rabbit | Biotin-XX |
| A6397 | Tetramethylrhodamine and Rhodamine Red dye | Rabbit | None |
| A6399 | Texas Red and Texas Red-X dye | Rabbit | None |
| Anti-Hapten Antibodies and Their Conjugates | |||
| 033700 | Biotin | Mouse | None |
| A6430 | Dinitrophenyl | Rabbit | None |
| A6435 | Dinitrophenyl | Rabbit | Biotin-XX |
| A11097 | Dinitrophenyl | Rabbit | Alexa Fluor 488 |
| A6423 | Dinitrophenyl | Rabbit | Fluorescein |
| A21285 | Nitrotyrosine | Rabbit | None |
For Research Use Only. Not for use in diagnostic procedures.