Zenon Antibody Labeling Kits—Table 7.7
Learn more in The Molecular Probes® Handbook
Table 7.7 Zenon Antibody Labeling Kits.
| Label | Abs/Em * | Mouse IgG1 Labeling Kit | Mouse IgG2a Labeling Kit | Mouse IgG2b Labeling Kit | Rabbit IgG Labeling Kit | Goat IgG Labeling Kit | Human IgG Labeling Kit |
|---|---|---|---|---|---|---|---|
| Alexa Fluor Dyes | |||||||
| Alexa Fluor 350 | 346/442 | Z25000 | Z25100 | Z25200 | Z25300 | Z25400 | |
| Alexa Fluor 405 | 402/421 | Z25013 | Z25113 | Z25213 | Z25313 | ||
| Alexa Fluor 430 | 434/539 | Z25001 | Z25301 | ||||
| Alexa Fluor 488 | 495/519 | Z25002 Z25090 † |
Z25102 | Z25202 | Z25302 | Z25602 | Z25402 |
| Alexa Fluor 532 | 531/554 | Z25003 | Z25303 | ||||
| Alexa Fluor 546 | 556/573 | Z25004 | Z25104 | Z25204 | Z25304 | ||
| Alexa Fluor 555 | 555/565 | Z25005 | Z25105 | Z25205 | Z25305 | Z25605 | Z25405 |
| Alexa Fluor 568 | 578/603 | Z25006 | Z25106 | Z25206 | Z25306 | Z25606 | |
| Alexa Fluor 594 | 590/617 | Z25007 | Z25107 | Z25207 | Z25307 | Z25607 | Z25407 |
| Alexa Fluor 647 | 650/668 | Z25008 | Z25108 | Z25208 | Z25308 | Z25608 | Z25408 |
| Alexa Fluor 660 | 663/690 | Z25009 | |||||
| Alexa Fluor 680 | 679/702 | Z25010 | Z25110 | Z25210 | Z25310 | ||
| Alexa Fluor 700 | 702/723 | Z25011 | |||||
| Alexa Fluor 750 | 749/775 | Z25312 | |||||
| Classic Dyes | |||||||
| Pacific Blue | 410/455 | Z25041 | Z25156 | Z25341 | |||
| Pacific Green | 411/500 | Z11203 | |||||
| Pacific Orange | 400/551 | Z25256 | Z25257 | ||||
| Fluorescein | 494/518 | Z25042 | Z25342 | ||||
| Texas Red-X | 595/615 | Z25045 | |||||
| Biotin |
|||||||
| Biotin-XX | NA | Z25052 | Z25152 | Z25252 | Z25352 | Z25452 | |
| Phycobiliproteins and Alexa Fluor Dye–Phycobiliprotein Tandem Conjugates | |||||||
| R-Phycoerythrin (R-PE) | 496, 546, 565/578 ‡ | Z25055 | Z25155 | Z25255 | Z25355 | Z25455 | |
| Alexa Fluor 610–R-PE | 496, 546, 565/628 ‡ | Z25020 | |||||
| Alexa Fluor 647–R-PE | 496, 546, 565/668 ‡ | Z25021 | Z25121 | Z25221 | |||
| Alexa Fluor 680–R-PE | 496, 546, 565/702 ‡ | Z25022 | |||||
| Allophycocyanin (APC) | 650/660 | Z25051 | Z25151 | Z25251 | Z25351 | Z25451 | |
| Alexa Fluor 700–APC | 650/723 | Z25030 | |||||
| Alexa Fluor 750–APC | 650/775 | Z25031 | |||||
| Enzymes | |||||||
| Horseradish peroxidase | NA | Z25054 | Z25154 | Z25254 | Z25354 | Z25454 | |
| Alkaline phosphatase | NA | Z25350 | |||||
| Tricolor Labeling Kits | |||||||
| Kit #1: Alexa Fluor 488, Alexa Fluor 555, Alexa Fluor 647 dyes | see above | Z25060 | Z25160 | Z25260 | Z25360 | Z25460 | |
| Kit #2: Alexa Fluor 350, Alexa Fluor 488, Alexa Fluor 594 dyes | see above | Z25070 | Z25170 | Z25270 | Z25370 | Z25470 | |
| Kit #3: Alexa Fluor 488, R-PE, Alexa Fluor 647–R-PE dyes | see above | Z25080 | Z25180 | Z25280 | Z25380 | ||
| * Approximate absorption and emission maxima, in nm. † This Zenon Antibody Labeling Kit is enhanced with TSA technology. ‡ Multiple absorption peaks. NA = Not applicable. | |||||||
For Research Use Only. Not for use in diagnostic procedures.