Molecular Probes Nucleic Acid Labeling Kits—Table 8.6
Learn more in The Molecular Probes® Handbook
Table 8.6 Molecular Probes nucleic acid labeling kits.
| Fluorophore | Ex/Em * | Similar Dyes | ULYSIS Nucleic Acid Labeling Kits | ARES DNA Labeling Kits | FISH Tag DNA Kits † | FISH Tag RNA Kits † | FluoReporter Oligonucleotide Amine Labeling Kits |
|---|---|---|---|---|---|---|---|
| Alexa Fluor 488 | 490/520 | Fluorescein (FITC or FAM), SpectrumGreen | U21650 | A21665 | F32947 | F32952 | A20191 |
| Oregon Green 488 | 495/520 | Fluorescein (FITC or FAM), SpectrumGreen | U21659 | ||||
| Alexa Fluor 532 | 525/550 | Rhodamine 6G | U21651 | ||||
| Alexa Fluor 546 | 555/570 | Cy3 dye, tetramethylrhodamine (TRITC), SpectrumOrange | U21652 | A21667 | |||
| Alexa Fluor 555 | 555/570 | Cy3 dye, tetramethylrhodamine (TRITC), SpectrumOrange | A21677 | F32948 | F32953 | ||
| Alexa Fluor 568 | 575/600 | Lissamine rhodamine B dye | U21653 | ||||
| Alexa Fluor 594 | 590/615 | Texas Red dye, SpectrumRed | U21654 | A21669 | F32949 | F32954 | |
| Alexa Fluor 647 | 650/670 | Cy5 dye | U21660 | A21676 | F32950 | F32955 | A20196 |
| * Excitation (Ex) and Emission (Em) maxima, in nm. † Also available are the FISH Tag DNA Multicolor Kit (F32951) and the FISH Tag RNA Multicolor Kit (F32956), each containing four different amine-reactive Alexa Fluor dyes. | |||||||
For Research Use Only. Not for use in diagnostic procedures.