Our Newest Cellular Analysis Products and Technologies
On this page:
Click-IT® Edu Now Available in Larger Size
Click-iT® EdU offers a simplified method for detecting nascent DNA synthesis and is now available in a 5-gram size. EdU is an alkyne-containing nucleoside analog of thymidine whose detection is based on a click reaction, a copper (I)–catalyzed reaction between an azide and an alkyne. The small size of the detection azide allows the use of mild conditions to access EdU incorporated into DNA. The Click-iT® EdU assay can be completed in less than 90 minutes, without the need for immunoreagents or signal amplification.
Learn More about Click-iT® EdU
Comparison of signal strength in tissues labeled with EdU or BrdU
Comparison of signal strength in tissues labeled with EdU (A) or BrdU (B).
New Mitochondrial Antibodies
Invitrogen™ mitochondrial antibodies can be used to study the role of mitochondria in various cellular pathways and diseases. New antibodies are available for the specific detection of mitochondrial proteins including GAPDH, HSDL2, DCXR, and SOD2. These antibodies are validated for multiple applications including western blots, immunocytochemistry, immunohistochemistry, immunoprecipitation, and ELISA.
Search for Primary Antibodies
Fibroblast cells labeled with L-xylulose reductase (DCXR) monoclonal antibody  
Fibroblast cells labeled with
L-xylulose reductase (DCXR)
monoclonal antibody.
CellLight® Lysosomes-GFP
CellLight® reagents are fluorescent protein–signal peptide fusions that permit specific labeling of cellular structures, including lysosomes, for live-cell imaging or for analyses following formaldehyde-based fixation. CellLight® reagents are powered by BacMam technology, which uses a modified insect cell baculovirus coupled with a mammalian promoter to efficiently deliver and express genes in mammalian cells.
Learn More about CellLight® Reagents
Imaging lysosomes and mitochondria with CellLight® reagents  
Imaging lysosomes (green)
and mitochondria (red) with
CellLight® reagents.
Click-iT® Protein Enrichment Kit
The Click-iT® Protein Enrichment Kit allows proteins that have metabolically or enzymatically incorporated azide-containing sugars, fatty acids, or amino acids to be efficiently captured on a resin of alkyne agarose. Bound protein is removed by standard proteolytic release upstream of protein sequencing. Superior to biotin- or lectin-based approaches, the kit is ideal for LC MS/MS–based biomarker discovery, posttranslational modification analysis, and other proteomics applications.
Learn More about Click-iT® Detection of Protein Synthesis & Posttranslational Modification
Comparison of Click-iT® protein enrichment technology and biotin-streptavidin protocols  
Comparison of Click-iT®
protein enrichment technology
and biotin-streptavidin protocols.
(click to enlarge)
PrestoBlue™ Cell Viability Reagent
The PrestoBlue™ Cell Viability Reagent is a simple add-and-read cell viability assay. Once a plate containing cells and compounds is prepared, the PrestoBlue™ reagent is added and results can be read in as little as 10 minutes. The PrestoBlue™ reagent allows for reliable, high-quality cell viability data and continuous live-cell monitoring and assessment after measuring viability.
Learn More about the PrestoBlue™ Cell Viability Reagent.
 Measure cell viability in a fraction of the time resazurin-based assays take, without loss of sensitivity  
Measure cell viability
in a fraction of the time
resazurin-based assays take, without loss of sensitivity.
Influenza Neuraminidase Assay Kits
Both the Applied Biosystems® NA-XTD™ Influenza Neuraminidase Assay Kit and NA-Fluor™ Influenza Neuraminidase Assay Kit provide chemiluminescent or fluorescent detection reagents for quantitating the sensitivity of influenza virus isolates to neuraminidase inhibitors. The NA-XTD™ assay provides longer-lasting light emission compared to the first-generation NA-Star® assay, enabling flexibility in read time and improved ease of use. The NA-Fluor™ kit provides the classic MUNANA-based assay in an easy-to-use kit format. These assays enable viral quantitation, viral growth assays, and identification of new inhibitors in cells or virus isolates.
Learn More about the NA-XTD™ Influenza Neuraminidase Assay Kit and the NA-Fluor™ Influenza Neuraminidase Assay Kit
Neuraminidase sensitivity determination with NA-Fluor, NA-XTD, and NA-Star assays
Neuraminidase sensitivity determination with NA-Fluor™, NA-XTD™, and NA-Star® assays.
Qubit® 2.0 Fluorometer: The Next Generation of Nucleic Acid and Protein Quantitation
Designed in response to customer feedback, the new Qubit® 2.0 Fluorometer features an improved user interface that makes the instrument more intuitive and easier to use, while providing the same high accuracy and sensitivity as the original Qubit® Fluorometer. The new features include a larger LCD touch screen, step-by-step workflow navigation, and a USB port and automated data logging for efficient data management. See The Qubit® 2.0 Fluorometer article, in this issue.
Learn More about the Qubit® 2.0 Fluorometer
The Qubit® 2.0 Fluorometer  
The Qubit® 2.0 Fluorometer.
New Antibodies for Hyperosmotic Stress Research
FAK phosphorylation at tyrosine 861 has been indicated as a requirement for mammalian cells to survive under hyperosmotic stress. EGFR is also activated in downstream hyperosmotic stress signaling cascades. Invitrogen now offers two new ABfinity™ recombinant rabbit monoclonal antibodies to help elucidate FAK [pY861] and EGFR signaling. These antibodies are validated for western blotting and immunofluorescence applications.
Search for Primary Antibodies
HeLa cells labeled with EGFR ABfinity™ antibody  
HeLa cells labeled with
EGFR ABfinity™ antibody.
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.