Image of a Molecular Probes® product pouch
Two-color assay indicates live and dead sperm cells
This kit enables analysis of sperm viability and fertilizing potential. SYBR® 14 labels live sperm with green fluorescence and propidium iodide labels membrane-compromised or dead sperm with red fluorescence.
This protocol can be used for:
  • Identifying live and dead sperm using a fluorescence microscope
This protocol should not be used for:
You will need the following for this protocol:
Assay protocol
Dilute
1. Dilute semen sample in Live Cell Imaging Solution plus 10% BSA
Add-2
2. Add 900 µL DMSO to SYBR® 14 dye to make a stock solution
Add-2
3. Add 1 µL of SYBR® 14 stock solution and 5 µL of propidium iodide solution to a 1 mL sample of diluted semen to label
Clock-5
4. Incubate for 5–10 minutes at 37°C
Image
5. Image cells
Spectral information and storage
  SYBR® 14 Propidium Iodide
Excitation/Emission (in nm) 488/516 535/617
Standard filter set FITC or GFP TRITC
EVOS® Light Cube GFP TRITC
Storage conditions –20°C –20°C
 Protocol tips
  • Phosphate-containing buffers may interfere with SYBR® 14 dye staining
  • Semen dilutions of 1:10 (goat) to 1:40 (bovine) result in acceptable cell densities
  • Dye stock solutions should be stored frozen
live-dead-sperm-s000002
Bovine sperm labeled with the LIVE/DEAD® Sperm Viability Kit.