CyQUANT™ MTT and XTT Cell Viability Assays
Invitrogen™
CyQUANT™ MTT and XTT Cell Viability Assays
CyQUANT MTT and XTT Cell Viability assays are colorimetric microplate assays for accurate assessment of cell health, proliferation, and cytotoxicity. CyQUANT MTT is a well-established assay, while CyQUANT XTT offers improved performance and continuous measurement without cell lysis or solubilization.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDye TypeEmission
X12223XTT450, 660
V13154MTT570
Catalog number X12223
Price (USD)
424.65
Online Exclusive
433.00
Save 8.35 (2%)
1 kit
Dye Type:
XTT
Emission:
450, 660
Price (USD)
424.65
Online Exclusive
433.00
Save 8.35 (2%)
1 kit
CyQUANT MTT and XTT Cell Viability assays are colorimetric microplate assays used to measure the metabolic activity of cells, which is an indicator of cell viability, proliferation, and cytotoxicity. CyQUANT MTT and XTT function as cell health indicators by using the cellular redox potential of live cells to convert a tetrazolium-based salt reagent, MTT or XTT, to a brightly colored formazan product. The absorbance of the formazan can be measured using a microplate reader and correlates with the number of metabolically active cells to quantitively measure cell viability and proliferation.
Measuring cell viability is essential for assessing cell health, determining genotoxicity, and evaluating anti-cancer drugs. Colorimetric indicators such as MTT and XTT offer rapid, cost-effective, and consistent methods for quantitatively determining mammalian cell viability using microplate readers. The CyQUANT MTT assay is a well-established and popular cell viability assay, while the CyQUANT XTT assay provides a high dynamic range and low variability. In addition to improved performance, the CyQUANT XTT assay offers continuous measurement without the need for cell lysis or solubilization.
Features of the CyQUANT MTT Cell Viability Assay
  • Complete and optimized MTT assay kit—includes all reagents needed for colorimetric detection of viable mammalian cells
  • Trusted MTT reagent—uses the well-established MTT reagent to measure cellular redox potential
  • Configured to perform 1,000 tests using a 96-well microplate
Principle of the CyQUANT MTT Cell Viability Assay
The CyQUANT MTT assay is based on the reduction of the water-soluble, yellow tetrazolium salt MTT (3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide) to insoluble, purple formazan crystals by metabolically active cells. The redox potential in the mitochondria of active mammalian cells reduces MTT to a strongly pigmented formazan product. After solubilization, the absorbance of the formazan can be measured on a microplate reader, which correlates with the number of metabolically active cells.
Protocol for the CyQUANT MTT Cell Viability Assay
Prepare the MTT reagent and incubate cells for a specific period, during which viable cells convert MTT into insoluble purple formazan product. The formazan crystals are then solubilized with the included SDS (sodium dodecyl sulfate) reagent or DMSO, and the absorbance is measured using a microplate reader at 570 nm.
Features of the CyQUANT XTT Cell Viability Assay
  • High reproducibility—absorbance-based viability assay that measures redox potential in live cells with low variability
  • Improved dynamic range and sensitivity—enhanced performance compared to other colorimetric viability assays
  • Easy mix-and-read format—configured for one 96-well plate at a time
  • Non-toxic and continuous—allows multiple time points to be collected without cell lysis
  • Quick and simple—no solubilization step required, making it faster and easier to perform than the MTT assay
  • Complete XTT assay kit—includes all the materials needed for ten 96-well plates
Principle of the CyQUANT XTT Cell Viability Assay
The CyQUANT XTT assay relies on the reduction of the water-soluble, yellow tetrazolium salt XTT (2,3-bis-(2-methoxy-4-nitro-5-sulfophenyl)-2H-tetrazolium-5-carboxanilide) to a water-soluble orange formazan product by metabolically active cells. Unlike MTT, XTT does not form insoluble crystals and produces a soluble formazan product that does not require solubilization. Since no cell lysis or solubilization are required for detection, the XTT assay can be used as a continuous assay, and multiple time points can be analyzed using an absorbance-based microplate reader.
In addition to the XTT Reagent, the CyQUANT XTT Cell Viability Assay kit includes electron coupling reagent to improve the dynamic range of the assay. The sensitivity and consistency of the assay is significantly increased when the electron coupling reagent is used with the CyQUANT XTT assay kit.
Protocol for the CyQUANT XTT Cell Viability Assay
For each 96-well plate, one bottle of XTT reagent and one tube of electron coupling reagent are thawed and mixed to create a stock solution, then added to cells. Cells are incubated for 2–4 hours, and metabolically active cells reduce XTT to form a water-soluble orange formazan dye. The absorbance of the dye is then measured using a microplate reader at 450 nm and 650 nm.
The XTT/electron coupling reagent stock solution should be used promptly after mixing, as extended room temperature storage or freeze/thaw cycles reduce assay performance and sensitivity. To ensure optimal performance and reproducibility, the CyQUANT XTT assay kit consists of separate XTT and electron coupling reagents that are mixed just prior to use.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Cell TypeMammalian Cells
Detection MethodColorimetric
Dye TypeXTT
Format96-well plate
Quantity1 Kit
Shipping ConditionWet Ice
Emission450, 660
For Use With (Application)Viability Assay, Proliferation Assay, Cytotoxicity Assay
For Use With (Equipment)Microplate Reader
Product LineCyQUANT
Product TypeCell Viability Assay
Unit Size1 kit
Contents & Storage
Store at ≤ -20°C and protect from light
Frequently asked questions (FAQs)
If I used XTT (Cat. No. X6493) with an electron coupling reagent, would I get the same results as if I used the CyQUANT XTT Cell Viability Assay kit (Cat. No. X12223)?
Both products use the same reagent, XTT. In theory, the results of any optimized assay using XTT (Cat. No. X6493) may be comparable to using the CyQUANT XTT Cell Viability Assay kit (Cat. No. X12223). Variations may arise due to the type of electron coupling reagent used, the final concentration of XTT, and any assay-specific requirements of the solutions used in resolubilizing the reagents.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
When should I use the XTT reagent alone (Cat No. X6493) versus the CyQUANT XTT Cell Viability Assay (Cat No. X12223)?
For the CyQUANT XTT Cell Viability Assay kit (Cat No. X12223), the amount of reagents, the formulation of the solutions, and the identity of the Electron Coupling Reagent are proprietary information. On the other hand, XTT (Cat. No. X6493) may be resolubilized in either water, buffer, media, or DMSO to a desired stock concentration. We would recommend using XTT, Cat No. X6493 if you need to optimize the final working concentration, if the type of buffer/solvent used to resolubilize the reagent is critical to the study, if you elect not to use an electron coupling reagent, or if you'd like to use a known electron coupling reagent of a defined concentration.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
What is the difference between XTT (Cat No. X6493) and CyQUANT XTT Cell Viability Assay (Cat No. X12223)?
XTT (Cat No. X6493) is 100 mg of XTT in solid form; you must resolubilize this into an aqueous solution or DMSO prior to use.
CyQUANT XTT Cell Viability Assay (Cat No. X12223) is a kit that provides two components: 10 vials each of the XTT Reagent and Electron Coupling Reagent. Both kit components are in solution in a ready-to-use format; mix 1 vial of the XTT Reagent and 1 vial of the Electron Coupling Reagent immediately before use.
Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.
Citations & References (3)
Citations & References
Abstract
Besnoitia besnoiti infection alters both endogenous cholesterol de novo synthesis and exogenous LDL uptake in host endothelial cells.
Authors:Silva LMR, Lütjohann D, Hamid P, Velasquez ZD, Kerner K, Larrazabal C, Failing K, Hermosilla C, Taubert A,
Journal:Sci Rep
PubMed ID:31040348
'Besnoitia besnoiti, an apicomplexan parasite of cattle being considered as emergent in Europe, replicates fast in host endothelial cells during acute infection and is in considerable need for energy, lipids and other building blocks for offspring formation. Apicomplexa are generally considered as defective in cholesterol synthesis and have to scavenge ... More
Mitochondrial Dysfunction in Huntington's Disease; Interplay Between HSF1, p53 and PGC-1a Transcription Factors.
Authors:Intihar TA, Martinez EA, Gomez-Pastor R,
Journal:Front Cell Neurosci
PubMed ID:30941017
Huntington's disease (HD) is a neurodegenerative disease caused by an expanded CAG repeat in the huntingtin (HTT) gene, causing the protein to misfold and aggregate. HD progression is characterized by motor impairment and cognitive decline associated with the preferential loss of striatal medium spiny neurons (MSNs). The mechanisms that determine ... More
Silver/silver chloride (Ag/AgCl) nanoparticles synthesized from Azadirachta indica lalex and its antibiofilm activity against fluconazole resistant Candida tropicalis.
Authors:Al Aboody MS,
Journal:Artif Cells Nanomed Biotechnol
PubMed ID:31137983
In this study, latex of Azadirachta indica was used for the synthesis of silver nanoparticles (AgNP). UV-visible spectroscopy revealed the formation of AgNPs and the absorption band optimized at 442?nm. Fourier transform infrared (FTIR) spectroscopy shows different functional groups (carboxyl, amine and hydroxyl) of biomolecule which are responsible for reduction ... More