protocol
Introduction
The TA Cloning® Kit with pCR®2.1 provides a quick, one-step cloning strategy for the direct insertion of a PCR product into a plasmid vector.
Advantages
Using the TA Cloning® Kit:
  • Eliminates any enzymatic modifications of the PCR product
  • Does not require the use of PCR primers that contain restriction sites
How TA Cloning® Works
Taq polymerase has a nontemplate-dependent activity that adds a single deoxyadenosine (A) to the 3´ ends of PCR products. The linearized vector supplied in this kit has single 3´ deoxythymidine (T) residues. This allows PCR inserts to ligate efficiently with the vector.
Diagram
The diagram below shows the concept behind the TA Cloning® method.
Thermostable polymerases containing extensive 3´ to 5´ exonuclease activity, such as Platinum® Pfx, do not leave 3´ A-overhangs. PCR products generated with Taq polymerase have a high efficiency of cloning in the TA Cloning® system as the 3´ A-overhangs are not removed. However, if you use a proofreading polymerase or wish to clone blunt-ended fragments, you can add 3´ A-overhangs by incubation with Taq at the end of your cycling program. 
Alternatively, you may want to try the Zero Blunt® PCR Cloning Kit (Catalog nos. K2700-20 and K2750-20). This kit offers efficient cloning of blunt-end PCR products generated using thermostable, proofreading polymerases. For more information, visit our website or contact Technical Service.
Materials
Storage Instructions
The TA Cloning® Kits are shipped on dry ice and contain a box of TA Cloning® Reagents (Box 1) and a box of One Shot® Competent Cells (Box 2). Catalog nos. K2020-20 and K2020-40 are not supplied with One Shot® Competent Cells. Store Box 1 at -20°C in a non-frost-free freezer and Box 2 at -80°C.
TA Cloning® Reagents
TA Cloning® Reagents (Box 1) are listed below. Note that the user must supply Taq Polymerase. Forty reaction kits are supplied as two 20 reaction kits. Store Box 1 at -20ºC.
Item
Composition
Amount
pCR ®2.1, linearized
25 ng/µl in 10 mM Tris-HCl, 1 mM EDTA, pH 8
5 x 10 µl
10X PCR Buffer
100 mM Tris-HCl, pH 8.3 (at 42°C)
500 mM KCl
25 mM MgC12
0.01% gelatin
100 µl
10X Ligation Buffer
60 mM Tris-HCl, pH 7.5
60 mM MgCl2
50 mM NaCl
1 mg/ml bovine serum albumin
70 mM b-mercaptoethanol
1 mM ATP
20 mM dithiothreitol
10 mM spermidine
100 µl
50 mM dNTPs
12.5 mM dATP
12.5 mM dCTP
12.5 mM dGTP
12.5 mM dTTP
(adjusted to pH 8.0)
10 µl
T4 DNA Ligase
4.0 Weiss units/µl
25 µl
Water
Deionized, autoclaved water
1 ml
Control DNA Template
0.1 µg/µl in 10 mM Tris-HCl, 1 mM EDTA, pH 8
10 µl
Control PCR Primers
0.1 µg/µl each in 10 mM Tris-HCl, 1 mM EDTA, pH 8
10 µl
One Shot® Reagents
The table below describes the items included in the One Shot® competent cell kit. Catalog nos. K2020-20 and K2020-40 are not supplied with competent cells. Forty reaction kits are supplied as two 20 reaction kits.
The transformation efficiency for TOP10F´ and TOP10 cells is 1 x 109 cfu/µg DNA. The transformation efficiency for INVaF´ is 1 x 108 cfu/µg DNA. Store competent cells at -80ºC.
TOP
Component
Composition
Amount
S.O.C. Medium
(may be stored at room temperature or +4ºC)
2% Tryptone
0.5% Yeast Extract
10 mM NaCl
2.5 mM KCl
10 mM MgCl2
10 mM MgSO4
20 mM glucose (dextrose)
6 ml
INVaF´, TOP10F´, or TOP10 cells
--
21 x 50 µl
pUC19 Control DNA
10 pg/µl in 5 mM Tris-HCl, 0.5 mM EDTA, pH 8
50 µl
Genotype of INVaF´
F´ endA1 recA1 hsdR17 (rk- , mk+ ) supE44 thi-1 gyrA96 relA1 f80lacZD M15 D (lacZYA-argF)U169 l-
Genotype of TOP10F´
F´ [lacIq Tn10 (TetR)] mcrA D (mrr-hsdRMS-mcrBC) F80lacZD M15 D lacC74 recA1 araD139 D(ara-leu)7697 galU galK rpsL (StrR ) endA1 nupG
Genotype of TOP10
F- mcrA D (mrr-hsdRMS-mcrBC) F80lacZDM15 D lacC74 recA1 araD139 D (ara-leu)7697 galU galK rpsL (StrR ) endA1 nupG
TOP
Experimental Outline
Introduction
To clone your gene of interest into pCR® 2.1, you must first generate a PCR product. The PCR product is ligated into pCR® 2.1 and transformed into competent cells. Since the PCR product can ligate into the vector in either orientation, individual recombinant plasmids need to be analyzed to confirm proper orientation. The correct recombinant plasmid is then purified for further subcloning or characterization.
Flow Chart
The table below describes the major steps necessary to clone your gene of interest into pCR® 2.1.
Step
Action
1
Amplify your PCR product using Taq polymerase and your own primers and parameters.
2
Ligate your PCR product into pCR ® 2.1.
3
Transform your ligation into competent E. coli.
4
Select colonies and isolate plasmid DNA. Analyze plasmid DNA for the presence and orientation of the PCR product by restriction enzyme digestion or sequencing.
When using the TA Cloning® Kit for the first time, we recommend that you perform the control reactions to help you evaluate your results.
TOP
Producing PCR Products
Guidelines for PCR
Generally 10-100 ng of DNA is sufficient to use as a template for PCR. If amplifying a pool of cDNA, the amount needed will depend on the relative abundance of the message of interest in your mRNA population. For optimal ligation efficiencies, we recommend using no more than 30 cycles of amplification.
Materials Supplied by the User
  • DNA template and primers for PCR product
  • Taq polymerase and appropriate 10X PCR buffer 
  • Thermocycler
You will need the following reagents and equipment.
Polymerase Mixtures
If you wish to use a mixture containing Taq polymerase and a proofreading polymerase, Taq must be in excess of a 10:1 ratio to ensure the presence of 3’ A-overhangs on the PCR product. We recommend using our Platinum® Taq DNA Polymerase High Fidelity.
If you use polymerase mixtures that do not have enough Taq polymerase or a proofreading polymerase only, you can add 3’ A-overhangs using the method in the Appendix.
Producing PCR Products
Perform the PCR in a 50 µl volume containing:
DNA Template 0-100 ng
10X PCR Buffer 5 µl
50 mM dNTPs 0.5 µl
Primers 1 µM each
Water to a total volume of 49 µl
Taq Polymerase 1 unit
Total Volume 50 µl
Gel Purification
If you do not obtain a single, discrete band from your PCR, you may gel-purify your fragment before proceeding. Take special care to avoid sources of nuclease contamination and long exposure to UV light. Alternatively, you may optimize your PCR to eliminate multiple bands and smearing (Innis et al., 1990). Our PCR Optimizer™ Kit (Catalog no. K1220-01) can help you optimize your PCR. Contact Technical Service for more information.
TOP
Cloning into pCR® 2.1
For optimal ligation efficiencies, we recommend using fresh (less than 1 day old) PCR products. The single 3´ A-overhangs on the PCR products will be degraded over time, reducing ligation efficiency. Take care when handling the pCR® 2.1 vector as loss of the 3´ T-overhangs will cause a blunt-end self-ligation of the vector and subsequent decrease in ligation efficiency.
Calculating Amount of PCR Product to Use
Use the formula below to estimate the amount of PCR product needed to ligate with 50 ng (20 fmoles) of pCR ® 2.1 vector:
X ng PCR product  =    (Y bp PCR product)(50 ng pCR® 2.1 vector)
                                        (size in bp of the pCR ® 2.1 vector: ~3900)
where X ng is the amount of PCR product of Y base pairs to be ligated for a 1:1 (vector:insert) molar ratio.
In general, 0.5 to 1.0 µl of a typical PCR sample with an average insert length (400-700 bp) will give the proper ratio of 1:1 (vector:insert). The ratio of 1:1 (vector:insert) gives the best efficiency of ligation. You may wish to do a second ligation reaction at a ratio of 1:3 (vector:insert), if you are concerned about the accuracy of your DNA concentrations.
Do not use more than 2-3 µl of the PCR sample in the ligation reaction as salts in the PCR sample may inhibit the T4 DNA Ligase.
Ligation at higher or lower temperatures than 14°C may reduce the ligation efficiency.
Procedure
  1. Centrifuge one vial of pCR® 2.1 to collect all the liquid in the bottom of the vial.
  2. Determine the volume of PCR sample needed to reach the required amount of PCR product (see above). Use sterile water to dilute your PCR sample if necessary.
  3. Set up the 10 µl ligation reaction as follows:
  4. Fresh PCR product                                                                            X µl
    10X Ligation Buffer                                                                            1 µl
    pCR ® 2.1 vector (25 ng/µl)                                                                 2 µl
    Water                                                                  to a total volume of 9 µl
    T4 DNA Ligase (4.0 Weiss units)                                                      1 µl
    Final volume                                                                                     10 µl
  5. Incubate the ligation reaction at 14°C for a minimum of 4 hours (preferably overnight). Proceed to Transforming Competent Cells.
Note:
   You may store your ligation reaction at-20°C until you are ready for transformation.
TOP
Transforming Competent Cells
Introduction
Once you have a ligated your insert into pCR ® 2.1, you are ready to transform the construct into competent E. coli. One Shot ® cells are provided to facilitate transformation. A protocol to transform One Shot ®cells is provided in this section. To transform another competent strain, refer to the manufacturer’s instructions.
INVaF´ and TOP10 E. coli do not express the lac repressor. You may express your product from pCR ® 2.1 in the absence of IPTG due to the presence of the lac promoter. IPTG will not have any affect on INVaF´ or TOP10 cells.
TOP10F´ does express the lac repressor (lacIq), which will repress transcription from the lac promoter. To perform blue-white screening for inserts, you must add IPTG to your plates to express LacZ a.
E. coli Host Strain
You may use any recA, endA E. coli strain including TOP10, TOP10F´, INVaF´, DH5a™, or equivalent for transformation. Other strains are suitable. 
If you amplified the PCR product from an ampicillin-resistant plasmid, use kanamycin to select for transformants containing your pCR ® 2.1 construct. Selecting with kanamycin will prevent contamination of the transformation reaction by the original ampicillin-resistant plasmid.
Materials Supplied by the User
In addition to general microbiological supplies (e.g. plates, spreaders), you will need the following reagents and equipment.
  • Chemically competent E. coli suitable for transformation
  • S.O.C. medium (warmed to room temperature)
  • Positive control, optional (e.g. pUC19)
  • LB plates containing 50 µg/ml kanamycin or 100 µg/ml ampicillin (two for each transformation)
  • 42°C water bath
  • 37°C shaking and non-shaking incubator
Preparing for Transformation
  • Equilibrate a water bath to 42°C.
  • Bring the S.O.C. medium to room temperature.
  • If you are using INVaF´ or TOP10 cells, take LB plates containing antibiotic and equilibrate at 37°C for 30 minutes.  Spread each plate with 40 µl of 40 mg/ml X-Gal. Let the liquid soak into the plates.
  • If you are using TOP10F´ cells, take LB plates containing antibiotic and equilibrate at 37°C for 30 minutes. Spread 40 µl each of 100 mM IPTG and 40 mg/ml X-Gal onto the plates. Let the liquid soak into the plates.
One Shot® Transformation Protocol
Follow the protocol below to transform One Shot ® Competent Cells. To transform another strain, refer to the manufacturer’s instructions.
  1. Centrifuge vials containing the ligation reactions briefly and place them on ice.
  2. Thaw on ice one 50 µl vial of frozen One Shot® Competent Cells for each transformation.
  3. Pipette 2 µl of each ligation reaction directly into the vial of competent cells and mix by stirring gently with the pipette tip.
  4. Incubate the vials on ice for 30 minutes. Store the remaining ligation mixtures at -20°C.
  5. Heat shock the cells for 30 seconds at 42°C without shaking. Immediately transfer the vials to ice.
  6. Add 250 µl of room temperature S.O.C. medium to each vial.
  7. Shake the vials horizontally at 37°C for 1 hour at 225 rpm in a shaking incubator.
  8. Spread 10 µl to 200 µl from each transformation vial on LB agar plates containing X-Gal and 50 µg/ml of kanamycin or 100 µg/ml ampicillin. Be sure to also include IPTG if you are using TOP10F´ cells. We recommend plating 10-50 µl for TOP10F´ or TOP10 cells and 50-200 µl for INVaF´ cells. Note: Be sure to plate two different volumes to ensure that at least one plate has well-spaced colonies. For plating small volumes, add 20 µl of S.O.C. to allow even spreading.
Incubate plates overnight at 37ºC. Shift plates to +4°C for 2-3 hours to allow for proper color development.
Transformed INVaF´ cells may appear very small after overnight growth when compared to other E. coli strains. The transformants may need to grow an additional 2-3 hours before selecting colonies for analysis.
Expected Results
For an insert size of 400-700 bp, you should obtain 50-200 colonies per plate depending on the volume plated and of these, approximately 80% should be white on X-Gal plates (INVaF´ and TOP10) or X-Gal/IPTG plates (TOP10F´). Note that ligation efficiency depends on insert size. As insert size increases, the efficiency will decrease.
TOP
Analyzing Transformants
Analyzing Positive Clones
  1. Pick at least 10 white colonies for plasmid isolation and restriction analysis.
  2. Grow colonies overnight in 2-5 ml LB broth containing either 100 µg/ml of ampicillin or 50 µg/ml kanamycin.
  3. Plasmid and analyze by restriction mapping or sequencing for orientation of the insert. We recommend using the PureLink™ HQ Mini Plasmid Purification Kit for purifying your plasmid DNA.
Sequencing Your Insert
If you wish to sequence your insert in pCR® 2.1, you may use the M13 Reverse Primer to sequence into your insert from the lac promoter. To sequence into the insert from the lacZa fragment, you can use either the T7 Promoter Primer or the M13 Forward Primer. Refer to the map of pCR® 2.1 for the primer sequences and location of the primer binding sites. For your convenience, we offer a custom primer synthesis service.
If you have problems obtaining transformants or the correct insert, perform the control reactions as described. These reaction swill help you troubleshoot your experiment. Refer to the Troubleshooting section for additional tips.
Long-Term Storage
Once you have identified the correct clone, be sure purify the colony and make a glycerol stock for long-term storage. We recommend that you store a stock of plasmid DNA at -20°C.
  1.   Streak the original colony on LB plates containing 100 µg/ml ampicillin or 50 µg/ml kanamycin.
  2.   Isolate a single colony and inoculate into 1-2 ml of LB containing 100 µg/ml ampicillin or 50 µg/ml kanamycin.
  3.   Grow until culture reaches stationary phase.
  4.   Mix 0.85 ml of culture with 0.15 ml of sterile glycerol and transfer to a cryovial.
  5.   Store at -80°C. 
TOP
Troubleshooting
If you do not obtain the results you expect, use the following table to troubleshoot your experiment. We recommend performing the control reactions to help you evaluate your results.
Problem
Reason
Solution
No colonies obtained from transformation
Bacteria were not competent.
Use the pUC19 control vector included with the One Shot ® kit to test transformation efficiency.
Incorrect concentration of antibiotic on plates or the plates are too old.
Use 100 µg/ml of ampicillin or 50 µg/ml kanamycin. Use fresh ampicillin plates (< 1 month old).
White colonies do not have insert
Single 3´ T-overhangs on the vector degraded.
Use another tube of vector. Avoid storing the vector for longer than 6 months or subjecting it to repeated freeze/thaw cycles. Check vector by performing the Self-Ligation Reaction.
Only white colonies obtained
No IPTG or X-Gal in plates.
Be sure to include X-Gal for blue/white screening and both IPTG and X-Gal if using TOP10F´.
Majority of colonies are blue or light blue with very few white colonies
The insert does not interrupt the reading frame of the lacZ gene.
If you have a small insert (< 500 bp), you may have light blue colonies. Analyze blue colonies as they may contain insert.
Used a polymerase that does not add 3´ A-overhangs.
Do not use proofreading polymerases such as Platinum ® Pfx as they do not add 3´ A-overhangs. Use Taq polymerase.
PCR products were gel-purified before ligation.
Gel purification can remove the single 3´ A-overhangs. If gel purification is needed, use nuclease-free solutions to purify fragment or optimize your PCR.
The PCR products were stored for a long period of time before performing the ligation reaction.
Use fresh PCR products. Efficiencies are reduced after as little as 1 day of storage.
Too much of the amplification reaction was added to the ligation.
The high salt content of PCR reactions can inhibit ligation. Do not use more than 2-3 µl of the PCR reaction in the ligation reaction.
Incorrect molar ratio of vector:insert used in the ligation reaction.
Estimate the concentration of the PCR product. Set up the ligation reaction with a 1:1 or 1:3 vector:insert molar ratio.
Some colonies have a light blue color or appear white with blue centers
Leaky expression of the lacZ fragment or only a partial disruption of lacZ by the insert.
If you are looking for a smaller size insert, 500 bp or less, analyze these colonies as they may contain insert.
White colonies or blue colonies of normal size are surrounded by smaller, white colonies
The smaller colonies are ampicillin-sensitive satellite colonies. Do not pick the small colonies as they do not contain any plasmid.
Use kanamycin selection. Be sure the stock solution of ampicillin and your plates are both fresh.
White colonies do not grow in liquid culture
Ampicillin-sensitive satellite colonies.
Be sure to pick large white colo­nies. Be sure the ampicillin is fresh. Use kanamycin to eliminate this problem.
No results from sequencing
Accidental use of the amplification primers in the kit for sequencing. These are for generating the control PCR product only.
Use the M13 Forward (-20) and Reverse Primers for sequencing. You may also use the T7 promoter primer to sequence into the insert.
The T7 primer used was not the right sequence.
Check the sequence of your T7 promoter primer and make sure it matches with the priming site on pCR ® 2.1.
An Sp6 primer was used to sequence inserts in pCR ® 2.1.
Do not use an Sp6 primer to sequence pCR ® 2.1. There is no binding site for this primer.
No PCR product
Either the Taq polymerase is inactive or the conditions for your PCR are not optimal.
Perform the control reactions to test the activity of the Taq polymerase. If Taq polymerase is active, you may need to optimize the conditions for your PCR reaction.
Low plasmid yield
Cells do not grow well in LB.
Try using S.O.C. medium with the appropriate antibiotic.
Explanation of Control Reactions
The following table describes the control reactions that can be performed to troubleshoot your TA Cloning® experiment and how to interpret the results from these control reactions.
TOP
Control Reaction
Explanation
Self- Ligation
This control reaction shows if pCR ® 2.1 has lost the 3´ T overhangs. Loss of the T overhangs results in blunt end ligation and disruption of the lacZa reading frame. False white colonies will result. Normally, less than 5% of the colonies should be white.
Transformation Control
Tests the transformation efficiency of the One Shot ® Competent Cells.
Transformation efficiency should be 1 x 10 8 cfu/µg DNA for INVF'and 1 x 10 9 cfu/µg DNA for TOP10 and TOP10F'.
Control PCR Product
Tests the PCR reagents including Taq polymerase.
Control Ligation Reaction
Tests the ligation reagents and pCR ® 2.1. Greater than 80% white colonies are produced and these colonies should contain vector with insert.
TOP
Appendix
Performing the Self-Ligation Reaction
The TA Cloning® vector is stable for six months if not subjected to repeated freeze-thaw cycles. Vector that has been stored for longer periods or repeatedly frozen and thawed will lose the 3´ T-overhangs resulting in "false" white positives. Follow the protocol below to perform the self-ligation reaction and transform One Shot® Competent Cells. If you are using another E. coli strain, follow the manufacturer’s instructions.
Procedure
  1. Set up the 10 µl self-ligation reaction as follows:
  2. ®
  3. Incubate overnight at 14-15°C. Centrifuge the vials containing the ligation reactions briefly and place them on ice.
  4. Thaw on ice one 50 µl vial of frozen One Shot® Competent Cells for each transformation.
  5. Pipette 1 µl of the Control Ligation Reaction from Step 1, above, directly into the vial of competent cells and mix by stirring gently with the pipette tip.
  6. Incubate the vial on ice for 30 minutes. Store the remainder of the ligation mixture at -20°C.
  7. Heat shock cells for 30 seconds at 42°C without shaking. Immediately transfer vials to ice.
  8. Add 250 µl of room temperature S.O.C. medium to the vial.
  9. Shake the vial horizontally at 37°C for 1 hour at 225 rpm in a shaking incubator.
  10. Spread 50 µl from the vial on a labeled LB agar plate containing 50 µg/ml of kanamycin or 100µg/ml ampicillin and X-Gal. Be sure to include IPTG if you are using TOP10F´.
  11. Incubate plates overnight at 37ºC.
    Expected Results
    You should expect about 5-25 blue colonies from the 50 µl plated. There should be less than 5% white colonies which result from supercoiled pCR® 2.1 vector. Over time, the 3´ T-overhangs will degrade, causing a blunt-end self-ligation of the vector. This can cause a frameshift of the lacZ gene, resulting in a "false" white or light blue colony with no insert.
    Performing the Control Reactions
    We recommend performing the control reactions the first time you use the kit to help you evaluate your results. Performing the control reactions involve producing a control PCR product using the reagents included in the kit and using this product in a ligation reaction.
    Producing the Control PCR Product
    TOP
    Use Taq Polymerase and the protocol below to amplify the control PCR product.
      1.   Set up the 50 µl PCR as follows:
    Control DNA Template (100 ng)                        1 µl
    10X PCR Buffer                                                5 µl
    50 mM dNTPs                                                 0.5 µl
    Control PCR Primers                                          1 µl
    Water                                                           41.5 µl
    Taq Polymerase (1 unit/µl)                                1 µl
    Total Volume                                                   50 µl
      2.   Overlay with 70 µl of mineral oil.
      3.   Amplify using the cycling parameters below:
    Step
    Time
    Temperature
    Cycles
    Denaturation
    1 minute
    94°C
    25X
    Annealing
    1 minute
    55°C
    25X
    Extension
    1 minute
    72°C
    25X
    Final Extension
    7 minutes
    72°C
    1X
      4.   Remove 10 µl from the reaction and analyze by agarose gel electrophoresis. A discrete 700 bp band should be visible.
    Control Ligation Reaction
    Using the control PCR product produced, set up the following ligation reaction. In general, 1 µl of the Control PCR Product should be sufficient for ligation. Alternatively, you may use the formula given in Cloning into pCR® 2.1 to estimate the amount of PCR product to ligate with 50 ng of pCR® 2.1.
    1. Set up the 10 µl Control Ligation Reaction as follows:
    2. ®
    3. Incubate the Control Ligation Reaction at 14°C for a minimum of 4 hours (preferably overnight).
    4. Transform 1 µl of the Control Ligation Reaction into one vial of One Shot® Competent Cells or into another suitable competent E. coli strain.
    5. Plate 10-50 µl of each transformation mix on LB agar plates containing 50 µg/ml kanamycin with X-Gal (and IPTG for TOP10F´ cells).
    6. Incubate plates overnight at 37°C.
      Transformation Control
      TA Cloning® Kits supplied with One Shot® Competent Cells will also be supplied with pUC19 plasmid for use as a transformation control. Transform one vial of One Shot® cells with 10 pg of pUC19 using the protocolin Transforming Competent Cells. Plate 10 µl to 50 µl of the transformation mixture on LB plates containing 100 µg/ml ampicillin. Transformation efficiency should be 1 x 109 cfu/µg DNA for TOP10F´ and TOP10 cells and 1 x 108cfu/µg DNA for INVaF´.
      Expected Results
      The Control Ligation Reaction should produce >80% white colonies. Over time, the 3´ T-overhangs will degrade, causing an increase in the number of background white colonies (those without inserts). The number of background colonies should not exceed 10% (see Performing the Self-Ligation Reaction). If this occurs, use another vial of pCR® 2.1 and avoid repeated freeze-thaw cycles.
      Adding 3´ A-Overhangs
      Direct cloning of DNA amplified by proofreading polymerases into pCR® 2.1 is often difficult due to very low cloning efficiencies. These low efficiencies are caused by the 3´ to 5´ exonuclease proofreading activity that removes the 3´ A-overhangs necessary for TA Cloning®. We have developed a simple method to clone these blunt-ended fragments.
      If you routinely clone blunt PCR products, we recommend the Zero Blunt® PCR Cloning Kit (Catalog nos. K2700-20 and K2750-20) for optimal cloning of blunt PCR products.
      Materials Supplied by the User
      You will need the reagents and equipment.
      • Taq polymerase
      • A heat block equilibrated to 72°C
      • Phenol-chloroform
      • 3 M sodium acetate
      • 100% ethanol
      • 80% ethanol
      • TE buffer
      Procedure
      1. After amplification with a proofreading polymerase, place vials on ice and add 0.7-1 unit of Taq polymerase per tube. Mix well. It is not necessary to change the buffer.
      2. Incubate at 72°C for 8-10 minutes (do not cycle).
      3. Extract immediately with an equal volume of phenol-chloroform.
      4. Add 1/10 volume of 3 M sodium acetate and 2X volume of 100% ethanol.
      5. Centrifuge at maximum speed for 5 minutes at room temperature to precipitate the DNA.
      6. Remove the ethanol, rinse the pellet with 80% ethanol, and allow to air dry. 
        TOP
Vector Maps
Map and Features of pCR ® 2.1
The table below describes the features of pCR ® 2.1. All features have been functionally tested.
Feature Benefit
lac promoter Allows bacterial expression of the lacZa fragment for a-complementation (blue-white screening).
lacZa fragment Encodes the first 146 amino acids of ß-galactosidase. Complementation in trans with the W fragment gives active ß-galactosidase for blue-white screening.
Kanamycin resistance gene Allows selection and maintenance in E. coli, useful when cloning products amplified from ampicillin-resistant plasmids.
Ampicillin resistance gene Allows selection and maintenance in E. coli.
pUC origin Allows replication, maintenance, and high copy number in E. coli.
T7 promoter and priming site Allows in vivo or in vitro transcription of anti-sense RNA. Allows sequencing of the insert.
M13 Forward (-20) and M13 Reverse Priming Sites Allows sequencing of the insert.
f1 origin Allows rescue of sense strand for mutagenesis and single-strand sequencing.
TOP
References
General Reference for PCR Technology
  1. Innis, M. A., Gelfand, D. H., Sninsky, J. J., and White, T. J. (eds.) (1990) PCR Protocols: A Guide to Methods and Applications. Academic Press, Inc., San Diego, CA.
General Molecular Biology Techniques
  1. Ausubel, F. M., Brent, R., Kingston, R. E., Moore, D. D., Seidman, J. G., Smith, J. A., Struhl, K., eds. (1990) Current Protocols in Molecular Biology. Greene Publishing Associates and Wiley-Interscience, New York.
  2. Sambrook, J., Fritsch, E. F., and Maniatis, T. (1989) Molecular Cloning: A Laboratory Manual, second edition, Cold Spring Harbor Laboratory Press, Plainview, New York.
TA Cloning®
  1. Andres, D. A., Seabra, M. C., Brown, M. S., Armstrong, S. A., Smeland, T. E., Cremers, F. P. M., and Goldstein, J. L. (1993) cDNA Cloning of Component A of Rab Geranylgeranyl Transferase and Demonstration of Its Role as a Rab Escort Protein. Cell 73: 1091-1099.
  2. Bates, M. D., Olsen, C. L., Becker, B. N., Albers, F. J., Middleton, J. P., Mulheron, J. G., Jin, S.-L. C., Conti, M., and Raymond, J. R. (1993) Elevation of cAMP Is Required for Down-regulation, but not Agonist-induced Desensitization, of Endogenous Dopamine D1 Receptors in Opossum Kidney Cells. J. Biol. Chem. 268: 14757-14763.
  3. Chen, Y., Weeks, J., Mortin, M. A., and Greenleaf, A. L. (1993) Mapping Mutations in Genes Encoding the Two Large Subunits of Drosophila RNA Polymerase II Defines Domains Essential for Basic Transcription Functions and for Proper Expression of Developmental Genes. Molec. Cell. Biol. 13: 4214-4222.
  4. Clark, J. M. (1988) Novel Non-Templated Nucleotide Addition Reactions Catalyzed by Procaryotic and Eucaryotic DNA Polymerases. Nuc. Acids Res. 16: 9677-9686.
  5. Falls, D. L., Rosen, K. M., Corfas, G., Lane, W. S., and Fischbach, G. D. (1993) ARIA, a Protein that Stimulates Acetylcholine Receptor Synthesis, is a Member of the Neu Ligand Family. Cell 72: 801-815.
  6. Hake, L. E. and Hecht, N. B. (1993) Utilization of an Alternative Transcription Initiation Site of Somatic Cytochrome c in the Mouse Produces a Testis-Specific Cytochrome c mRNA. J. Biol. Chem. 268: 4788-4797.
  7. Liang, P. and Pardee, A. B. (1992) Differential Display of Eukaryotic Messenger RNA by Means of the Polymerase Chain Reaction. Science 257: 967-973.
  8. Lin, X.-L., Lin, Y.-Z., Koelsch, G., Gustchina, A., Wlodawer, A., and Tang, J. (1992) Enzymic Activities of Two-chain Pepsinogen, Two-chain Pepsin, and the Amino-terminal Lobe of Pepsinogen. J. Biol. Chem. 267: 17257-17263.
  9. Mahendroo, M. S., Mendelson, C. R., and Simpson, E. R. (1993) Tissue-specific and Hormonally Controlled Alternative Promoters Regulate Aromatase Cytochrome P450 Gene Expression in Human Adipose Tissue. J. Biol. Chem. 268: 19463-19470.
  10. Mallick, C. A., Dudley, E. C., Viney, J. L., Owen, M. J., and Hayday, A. C. (1993) Rearrangement and Diversity of T Cell Receptor b Chain Genes in Thymocytes: A Critical Role for the b Chain in Development. Cell 73: 513-519.
  11. Mead, D. A., Pey, N. K., Herrnstadt, C., Marcil, R. A., and Smith, L. M. (1991) A Universal Method for the Direct Cloning of PCR Amplified Nucleic Acid. Bio/Technology 9: 657-663.
  12. Miller, W. H., Kakizuka, A., Frankel, S. Warrell, R. P., DeBlasio, A., Levine, K., Evans, R. M., and Dmitrovsky, E. (1992) Reverse Transcription Polymerase Chain Reaction for the Rearranged Retinoic Acid Receptor a Clarifies Diagnosis and Detects Minimal Residual Disease in Acute Promyelocytic Leukemia. Proc. Natl. Acad. Sci USA 89: 2694-2698.
TOP
25-0024       Version V  4/7/2004