Introduction
Dynabeads Sheep anti-Rat IgG in combination with any primary rat IgG antibody are ideal for depletion or positive isolation of cells from different species (e.g., mouse) depending on the specificity of the primary antibody. Cells can be directly isolated from any sample such as whole blood, bone marrow, MNC suspensions, or tissue digests.
Principle of Isolation
The primary rat IgG antibody is either added to the cell sample (indirect technique) or pre-coated onto the beads (direct technique) prior to cell isolation. Dynabeads are then mixed with the cell sample in a tube. The Dynabeads will bind to the target cells during a short incubation, and then the bead-bound cells are separated by a magnet.
  • Positive isolation – discard the supernatant and use the bead-bound cells for downstream applications (e.g., molecular analysis, cell culture).
  • Depletion – discard the bead-bound cells and use the remaining, untouched cells for any application.
Description of Materials
Dynabeads Sheep anti-Rat IgG are uniform, superparamagnetic polystyrene beads (4.5 μm diameter) coated with polyclonal Sheep anti-Rat IgG antibodies. Cross-reactivity to mouse antibodies is high and cross-reactivity to human antibodies is minimal.
Materials Supplied
  • 5 ml Dynabeads Sheep anti-Rat IgG 4 x 108 beads/ml in phosphate buffered saline (PBS), pH 7.4, containing 0.1% bovine serum albumin (BSA) and 0.02% sodium azide (NaN3).
  • This product will process up to 2 x 109 cells.
Additional Material Required
  • Magnet: (Dynal MPC™) - MPC-S for 20 μl to 2 ml samples, MPC-L for 1-8 ml samples, MPC-15 for 1-15 ml samples and MPC-50 for 15-50 ml samples.
  • Mixer allowing both tilting and rotation.
  • Buffer 1: PBS (without Ca2+ and Mg2+) w/0.1% BSA and 2 mM EDTA, pH 7.4.
Important Notes:
BSA can be replaced by human serum albumin (HSA) or fetal calf serum (FCS). EDTA can be replaced by sodium citrate. PBS containing Ca2+ or Mg2+ is not recommended.
TOP
Protocols
Dynabeads Washing Procedure
Dynabeads should be washed before use. The washing procedure is facilitated by the use of a magnet.
  1.    Resuspend the Dynabeads in the vial.
  2.    Transfer the desired volume of Dynabeads to a tube.
  3.    Add the same volume of Buffer 1, or at least 1 ml, and mix.
  4.    Place the tube in a magnet for 1 min and discard the supernatant.
  5.    Remove the tube from the magnet and resuspend the washed Dynabeads in the same volume of Buffer 1 as the initial volume of Dynabeads.
Sample Preparation
Cells can be directly isolated from any sample such as whole blood, bone marrow, MNC suspensions, or tissue digests. Please visit www.lifetechnologies.com/samplepreparation for a list of recommended sample preparation procedures.
Critical Steps for Cell Isolation
Use a mixer that provides tilting and rotation of the tubes to ensure Dynabeads do not settle at the bottom of the tube. When incubating Dynabeads and cells, the incubation temperature must be 2–8°C to reduce phagocytic activity and other metabolic processes. Never use less than 25 μl (1 x 107) Dynabeads per ml cell sample and at least 4 Dynabeads per target cell.
Table 1: Volume of Dynabeads added per ml of cell sample. The volumes can be scaled up as required.
  Positive isolation Depletion
Sample volume (1 x 107 cells/ml*) 1 ml Max 2.5 x 106
target cells
1 ml Max 2.5 x 106
target cells
Volume of Dynabeads 25 μl 50 μl
* If the concentration of cells is increased or the target cell concentration exceeds 2.5 x 106, the Dynabeads volume must be increased accordingly. Cell concentration can be up to 1 x 108cells per ml.
Cell Isolation - Indirect Technique
Labeling Cells with Rat IgG Antibodies
Use approximately 1 μg antibody per 106 target cells.
  • Recommended cell concentration: 1 x 107 cells/ml.
  1.    Add rat IgG antibodies to the cell suspension and mix.
  2.    Incubate for 20 min at 2–8°C.
  3.    Wash the cells by adding 2 ml Buffer 1 per 1 x 107 cells and centrifuge at 300 x g for 8 min. Discard the supernatant.
  4.    Resuspend the cells in Buffer 1 at 1 x 107 cells/ ml.
  5.    Proceed to Isolation or Depletion of Cells.
Isolation or Depletion of Cells
  1.    Add Dynabeads to the prepared sample according to Table 1.
  2.    Incubate for 20 min (positive isolation) or 30 min (depletion) at 2–8°C with gentle tilting and rotation.
  3.    Double the volume with Buffer 1 to limit trapping of unbound cells (optional).
  4.    Place the tube in a magnet for 2 min.
  5.    Depletion: Transfer the supernatant containing the unbound cells to a fresh tube for further experiments.
  6.    Positive isolation: Discard the supernatant and gently wash the bead-bound cells 4 times, using the following procedure:
            i) Add 1 ml Buffer 1 per 1 x 107 Dynabeads.
           ii) Place the tube in the magnet for 1 min and discard the supernatant.
  7.    Resuspend the cells in buffer/medium for downstream application.
Cell Isolation – Direct Technique
Pre-coating Dynabeads
  • Use 0.5 – 1.5 μg rat IgG antibodies per 25 μl (1 x 107) Dynabeads. (It is recommended to titrate the amount of antibody.)
  1.    Transfer washed Dynabeads to a tube.
  2.    Add rat IgG antibodies.
  3.    Incubate for ≥ 30 minutes at 2-8°C with gentle tilting and rotation.
  4.    Place the tube in a magnet for 1 min and discard the supernatant.
  5.    Wash the beads twice using 2 ml of Buffer 1.
  6.    Remove the tube from the magnet and resuspend the Dynabeads in the same volume Buffer 1 as the initial volume of Dynabeads.
Isolation or Depletion of Cells
  1.    Add the pre-coated Dynabeads to the cells according to Table 1.
  2.    Incubate for 20 min (positive isolation) or 30 min (depletion) at 2–8°C with gentle tilting and rotation.
  3.    Double the volume with Buffer 1 to limit trapping of unbound cells (optional).
  4.    Place the tube in a magnet for 2 min.
  5.    Depletion: Transfer the supernatant containing the unbound cells to a fresh tube for further experiments.
  6.    Positive isolation: Discard the supernatant and gently wash the bead-bound cells 4 times, using the following procedure:
            i) Add 1 ml Buffer 1 per 1 x 107 Dynabeads.
           ii) Place the tube in the magnet for 1 min and discard the supernatant.
  7.    Resuspend the cells in buffer/medium for downstream application.
TOP
Technical Recommendations
Indirect versus Direct Technique
Use the indirect technique when:
  • A cocktail of rat IgG antibodies is used.
  • Very high depletion efficiency is needed.
  • The affinities of rat IgG antibodies are low.
  • The cells express low number of target antigens.
  • The direct technique gives unsatisfactory purity.
The direct technique may be used if:
  • The affinity of the primary antibody is high.
  • The cells express a high number of target antigens.
  • A stock preparation of primary coated Dynabeads is desired.
Antibody Selection
The choice of primary antibody is the most important factor for successful cell isolation. Note that some antibodies may show reduced antigen-binding efficiency when coated onto beads (direct technique), even though the antibody shows good results in other immunological assays.
Labeling Cells with Rat IgG Antibodies
  • To avoid non-specific binding of cells (e.g., monocytes, B cells), add aggregated IgG to block Fc receptors prior to adding the primary antibodies.
  • Excess antibody must be removed by washing before cell isolation.
Isolation and Depletion of Target Cells
  • Remove density gradient media (e.g., Ficoll): Wash cells prior to adding rat IgG antibodies or Dynabeads.
  • Remove soluble factors in serum: Serum may contain soluble factors (e.g., antibodies or cell surface antigens), which can interfere with the cell isolation protocol. Washing the cells once may reduce this interference.
TOP
General Information
Invitrogen Dynal AS complies with the Quality System Standards ISO 9001:2000 and ISO 13485:2003.
Storage/Stability
This product is stable until the expiry date stated on the label when stored unopened at 2-8°C. Store opened vials at 2-8°C and avoid bacterial contamination. Keep Dynabeads in liquid suspension during storage and all handling steps, as drying will result in reduced performance. Resuspend well before use.
Warnings And Limitations
This product is for research use only. Not intended for any animal or human therapeutic or diagnostic use unless otherwise stated. Follow appropriate laboratory guidelines. This product contains 0.02% sodium azide as a preservative, which is cytotoxic.
Avoid pipetting by mouth!
Sodium azide may react with lead and copper plumbing to form highly explosive metal azides. When disposing through plumbing drains, flush with large volumes of water to prevent azide build up. Certificate of Analysis (CoA) is available upon request. Material Safety Data Sheet (MSDS) is available at .
TOP
References
  1. Philippe D et al. (2003) Anti-inflammatory properties of the m opioid receptor support its use in the treatment of colon inflammation. J.Clin.Invest. 111:1329-1338.
  2. Feng B et al. (2000) Development of mouse dendritic cells from lineage-negative c-kitlow pluripotent hemopoieitic stem cells in vitro. Stem Cells. 18:53-60.
  3. Zhou P et al. (2004) Expression of dual TCR on DO11.10 T cells allows for ovalbumin-induced oral tolerance to prevent T cell-mediated colitis directed against unrelated enteric antigens. J. Immunol. 172:1515-1523.
  4. Yeh JRJ et al. (2000) The anti-angiogenic agent TNP-470 requires P53 and P21CIP/WAF for endothelial cell growth arrest. PNAS 97(23): 12782-12787.
  5. Na Nakorn T et al. (2002) Characterization of mouse clonogenic megakaryocyte progenitors. PNAS 100(1):205-210.
  6. Steptoe RJ et al. (2003) Transfer of hematopoietic stem cells encoding auto-antigen prevents autoimmune diabetes. J.Clin.Invest. 111:1357–1363.
  7. Tanchot C et al. (2004) Immune regulation by self-reactive T cells is antigen-specific. J. Immunol. 172:4285-4291.
  8. Weber M et al. (2004) Minor histocompatibility antigens on canine hemopoietic progenitor cells. J. Immunol. 170:5861-5868.
  9. Metwali A et al. (2004) Cutting Edge: Hemokinin has Substance P-like function and expression in inflammation. J. Immunol. 172: 6528-6532.
  10. Tajima F et al. (2000) CD34 expression by murine hematopoietic stem cells mobilized by granulocyte colony-stimulating factor. Blood 96: 1989-1993.
  11. El-Sukkari D et al. (2003) The protease inhibitor Cystatin C is differentially expressed among dendritic cell populations, but does not control antigen presentation. J. Immunol. 171:5003-5011.
  12. Foussat A et al. (2003) A comparative study between T Regulatory Type I and CD4+CD25+ T cells in the control of inflammation. J. Immunol. 171: 5018-5026.
  13. Joffre O et al. (2004) Induction of antigen-specific tolerance to bone marrow allografts with CD4+CD25+  lymphocytes. Blood. 103: 4216-4221.
  14. Hausl C et al. (2004) Preventing restimulation of memory B cells in hemophilia A: a potential new strategy for the treatment of antibody-dependent immune disorders. Blood. 104: 115-122.
TOP
110.35.indd   Rev 003     5-May-2006