Introduction
This protocol is intended for magnetic isolation or depletion of CD2+cells directly from whole blood, buffy coat or MNC. For rapid and consistent results in protein or gene expression analysis, lyse the CD2+ cells while they are still attached to the beads and proceed with the lysate for further molecular analysis. If bead-free T cells are required, please see list of recommended products.
Principle of Isolation
Dynabeads are mixed with the sample in a tube. The Dynabeads will bind to the target cells during a short incubation, and then the bead-bound cells are separated by a magnet.
  • Positive isolation – discard the supernatant and use the bead-bound cells for downstream molecular applications (e.g. protein or gene expression analysis).
  • Depletion – discard the bead-bound cells and use the remaining, untouched cells for any application.
Sample Preparation
Isolate T cells directly from whole blood, buffy coat or MNC. Please visit Pan Mouse IgG - Depletion or Positive Isolation of Cells from Different Species and follow our QuickLinks for recommended sample preparation procedures.
Additional Materials Required
Materials that are not included, but are needed to perform the entire protocol:
• Mixer allowing both tilting and rotation
• Magnet: See Dynabeads® mRNA Purification Kit for mRNA Purification from Total RNA preps for magnet recommendations
• Isolation buffer: Ca2+ and Mg2+ free phosphate buffered saline (PBS) (e.g. Gibco cat.no. 14190-094) supplemented with 0.1% BSA and 2mM EDTA
Note:
BSA can be replaced by human serum albumin (HSA) or 2% FBS/FCS. EDTA can be replaced by 0.6% sodium citrate.
Critical notes
  • Use a mixer that provides tilting and rotation of the tubes to ensure that Dynabeads do not settle at the bottom of the tube.
  • Follow the recommended volumes and incubation times.
  • Avoid air bubbles during pipetting.
TOP
Protocols
This protocol describes magnetic labeling and isolation of CD2+ T cells from whole blood, buffy coat or MNC.
Table 1: Volume of Dynabeads added per ml of sample.
Positive isolation Depletion
Sample volume
  • up to 1x107 MNC/ml*
  • washed whole blood
  • washed and diluted (1:1) buffy coat*
1 ml 1 ml
Volume of Dynabeads 50 μl 100 μl
Total no. of cells processed per product 1x109 5x108
* When working with lower cell concentrations, use the same volumes as indicated. When working with higher cell concentrations, scale up all reagent volumes and total volumes accordingly.
Dynabeads Washing Procedure
Dynabeads should be washed before use.
  • Resuspend the Dynabeads in the vial.
  • Transfer the desired volume of Dynabeads to a tube.
  • Add the same volume of Isolation buffer, or at least 1 ml, and mix.
  • Place the tube in a magnet for 1 min and discard the supernatant.
  • Remove the tube from the magnet and resuspend the washed Dynabeads in the same volume of Isolation buffer as the initial volume of Dynabeads (bullet point 2).
Depletion or Positive Isolation of CD2+ cells
  1. Add the appropriate volume of Dynabeads to the prepared sample according to table 1.
  2. Incubate for 20 min (positive isolation) or 30 min (depletion) at 2 - 8°C with gentle tilting and rotation.
  3. Place the tube in a magnet for 2 min.
  4. For depletion, transfer supernatant to a new tube for further use.
  5. For positive isolation, discard the supernatant and wash the beadbound cells 3 times by resuspending in Isolation Buffer to the original sample volume, and separate using a magnet.
For molecular studies, lyse cells while still attached to the beads and transfer supernatant (lysate) to a new tube for further processing.
TOP
General Information
Invitrogen Dynal AS complies with the Quality System Standards ISO 9001:2000 and ISO 13485:2003.
Storage/Stability
This product is stable until the expiry date stated on the label when stored unopened at 2-8°C. Store opened vials at 2-8°C and avoid bacterial contamination. Keep Dynabeads in liquid suspension during storage and all handling steps, as drying will result in reduced performance. Resuspend well before use.
Warnings And Limitations
This product is for research use only. Not intended for any animal or human therapeutic or diagnostic use unless otherwise stated. Follow appropriate laboratory guidelines. This product contains 0.02% sodium azide as a preservative, which is cytotoxic.
Avoid pipetting by mouth!
Sodium azide may react with lead and copper plumbing to form highly explosive metal azides. When disposing through plumbing drains, flush with large volumes of water to prevent azide build up. Certificate of Analysis (CoA) is available upon request. Material Safety Data Sheet (MSDS) is available at .
TOP
References
  1. Colucci S et al (2004) T cells support osteoclastogenesis in an in vitro model derived from human multiple myleloma bone disease: the role of the OPG/TRAIL interaction. Blood 104: 3722-3730.
  2. Decker T et al (2003) Rapamycin-induced G1 arrest in cycling B-CLL cells is associated with reduced expression of cyclin D3, cyclin E, cyclin A and surviving. Blood 101:278-285.
  3. Ma W et al (2004) Dexamethasone inhibits IL-12p40 production in lipo-polysaccharide- stimulated human monocytic cells by down-regulating the activity of c- Jun N-terminal kinase, the activation protein- 1 and NF-kB transcription factors. J. Immunol. 172:318-330.
  4. Sato K et al (2003) Modified myeloid dendritic cells act as regulatory dendritic cells to induce anergic and regulatory T cells. Blood 101:3581-3589.
  5. Slager EH et al (2004) Identification of multiple HLA-DR-restricted epitopes of the tumor associated antigen CAMEL by CD4+ Th1/Th2 lymphocytes. J. Immunol. 172:5095-5102.
  6. Viglietta V et al (2004) Loss of functional suppression by CD4+CD25+ regulatory T cells in patients with multiple sclerosis. J.Exp.Med. 199: 971-979. 
TOP
111.59D.indd  Rev 002    5-May-2007