| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Inorganic Salts | |||
| Potassium Phosphate monobasic (KH2PO4) | 136.0 | 144.0 | 1.0588236 |
| Sodium Chloride (NaCl) | 58.0 | 9000.0 | 155.17241 |
| Sodium Phosphate dibasic (Na2HPO4-7H2O) | 268.0 | 795.0 | 2.966418 |
Reference:
Related Products:
20012 - PBS, pH 7.2
Cat No:
20012019
,
20012027
,
20012043
,
20012050
,
20012068
70013 - PBS, 10x, pH 7.2
Cat No:
70013016
,
70013032
,
70013065
,
70013073
70011 - PBS,10x, pH 7.4
Cat No:
70011036
,
70011044
,
70011051
,
70011069