Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  2  3  4  5  6  7  8  9  10  11  12  13  14  15  16  17  18  19  20  21  22  23  24  25  26  27  28  29  30  40  50  60  67 
421 - 450 of 2000 products displayed
  • c-Myc Tag Antibody, Alexa Fluor® 647 (Monoclonal, 9E10)
  • Host Mouse
  • Target Species Human
  • Conjugate Alexa Fluor® 647
  • Catalog # MA1980A647
  • Size 50 µl
Applications
Immunocytochemistry (1:25-1:100), Immunofluorescence (1:25-1:100), Western Blot (1:500)
Alternate names:
Transcription factor p64; Proto-oncogene c-Myc; Class E basic helix-loop-helix protein 39
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1980A647 219.00
Check your price
  • Mouse anti-Human IgG1 Fc, HRP
  • Host Mouse
  • Target Species Human
  • Conjugate HRP
  • Catalog # A10648
  • Size 200 µg
Applications
ELISA (1:500 to 1:2,000), Immunohistochemistry (1:500 to 1:2,000), Western Blot (1:500 to 1:2,000)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A10648 149.00
Check your price
  • TIMP1 Antibody, Biotin (Monoclonal, 102D1)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # MA513685
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1-2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Tissue inhibitor of metalloproteinases 1; Fibroblast collagenase inhibitor; Erythroid-potentiating activity
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA513685 399.00
Check your price
  • TRAC Antibody (Monoclonal, 3A8)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # TCR1145
  • Size 200 ug
Applications
Flow Cytometry (1-2 µg/test), Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1:10-1:100), Immunoprecipitation (Assay Dependent)
Alternate names:
T-cell antigen receptor, alpha polypeptide; T-cell receptor, alpha (V,D,J,C)
Catalog # Size Price (AUD) Your Price (AUD) Qty
TCR1145 309.00
Check your price
  • Complement C3b Antibody (Monoclonal, 10C7)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA170054
  • Size 250 ug
Applications
ELISA (Assay dependent), Flow Cytometry (1 ug/1 million cells)
Alternate names:
HSE-MSF; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA170054 399.00
Check your price
  • Digoxigenin Antibody
  • Host Sheep
  • Target Species Chemical
  • Conjugate Unconjugated
  • Catalog # PA185378
  • Size 1 mL
Applications
ELISA (1:100-1:5000), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
DIG, C23H34O5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA185378 399.00
Check your price
  • CUX1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536355
  • Size 100 µl
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Homeobox protein cux-1; CCAAT displacement protein; CDP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536355 309.00
Check your price
  • Human Serum Albumin Antibody, FITC
  • Host Chicken
  • Target Species Human
  • Conjugate FITC
  • Catalog # PA172056
  • Size 500 µg
Applications
Immunohistochemistry (Assay Dependent)
Alternate names:
Albumin, HSA, Serum albumin precursor, ALB
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA172056 199.00
Check your price
  • Human PKM2 Recombinant Protein
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP75744
  • Size 1 mg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Tumor Type M2 Pyruvate Kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75744 2
Check your price
  • Human IL-10 Recombinant Protein
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RIL1025
  • Size 25 ug
Applications
Control (Assay Dependent), ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Cytokine synthesis inhibitory factor
Catalog # Size Price (AUD) Your Price (AUD) Qty
RIL1025 608.00
Check your price
  • Rhodopsin Antibody (Monoclonal, 1D4)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1722
  • Size 100 µL
Applications
Immunocytochemistry (1:1,000), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay dependent), Western Blot (1:100-1:1000)
Alternate names:
Opsin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1722 375.00
Check your price
  • HOXA13 Antibody
  • Host Rabbit
  • Target Species Dog, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA540459
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Homeobox protein Hox-1.10; Homeobox protein Hox-1J
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540459 309.00
Check your price
  • CD56 Antibody, APC (Monoclonal, MEM-188)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate APC
  • Catalog # MA119462
  • Size 100 tests
Applications
Flow Cytometry (10µl/10^6 cells)
Alternate names:
NCAM1, NCAM, CD-56, MSK39, neural cell adhesion molecule 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA119462 309.00
Check your price
  • Donkey anti-Rabbit IgG (H+L) Highly Cross-Adsorbed, Alexa Fluor® 555
  • Host Donkey
  • Target Species Rabbit
  • Conjugate Alexa Fluor® 555
  • Catalog # A31572
  • Size 500 µL
Applications
Flow Cytometry (1-10 µg/mL), Immunocytochemistry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A31572 209.00
Check your price
  • Human IFN gamma Recombinant Protein
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RIFNG50
  • Size 50 ug
Applications
Control (Assay Dependent), ELISA (Assay Dependent), Immunoprecipitation (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Immune interferon
Catalog # Size Price (AUD) Your Price (AUD) Qty
RIFNG50 207.00
Check your price
  • P-Selectin Antibody, PE (Monoclonal, Psel.KO.2.7)
  • Host Mouse
  • Target Species Bovine, Horse, Cat, Goat, Human, Mouse, Sheep, Rat
  • Conjugate PE
  • Catalog # MA181849
  • Size 100 Tests
Applications
Flow Cytometry (Neat-1/5)
Alternate names:
Platelet activation dependent granule-external membrane protein; Leukocyte-endothelial cell adhesion molecule 3; Granule membrane protein 140; CD62 antigen-like family member P; CD62P antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA181849 375.00
Check your price
  • PRMT6 Antibody (Monoclonal, 4G2)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA515584
  • Size 100 µL
Applications
Western Blot (1/500-1/2000)
Alternate names:
Histone-arginine N-methyltransferase PRMT6; Heterogeneous nuclear ribonucleoprotein methyltransferase-like protein 6
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515584 349.00
Check your price
  • Phospho-p21 (Ser130) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA512644
  • Size 400 µL
Applications
Dot blot (1:500)
Alternate names:
p21; Melanoma differentiation-associated protein 6; CDK-interacting protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA512644 349.00
Check your price
  • Cytokeratin 5/8 Antibody (Monoclonal, RCK102)
  • Host Mouse
  • Target Species Dog, Cat, Hamster, Human, Mouse, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA106314
  • Size 100 µL
Applications
Flow Cytometry (1:100-1:200), Immunocytochemistry (Assay dependent), Immunohistochemistry (Paraffin, Frozen) (1:100-1:200), Western Blot (1:100-1:1000)
Alternate names:
Type-II keratin Kb8; Keratin-8; Cytokeratin-8; Type-II keratin Kb5; Keratin-5; Cytokeratin-5; 58 kDa cytokeratin; Cytokeratin endo A
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106314 349.00
Check your price
  • N-cadherin Antibody (Monoclonal, NCD-2)
  • Host Rat
  • Target Species Chicken, Xenopus
  • Conjugate Unconjugated
  • Catalog # 132100
  • Size 100 µg
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Immunoprecipitation (Assay Dependent), Inhibition Assays (100 ug/ml), Western Blot (1-10 ug/ml)
Alternate names:
Neural cadherin
Catalog # Size Price (AUD) Your Price (AUD) Qty
132100 324.00
Check your price
  • Goat anti-Mouse IgG2b Cross-Adsorbed, Alexa Fluor® 350
  • Host Goat
  • Target Species Mouse
  • Conjugate Alexa Fluor® 350
  • Catalog # A21140
  • Size 250 µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A21140 149.00
Check your price
  • Phospho-Tau (Thr212) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 44740G
  • Size 10 blots
Applications
Flow Cytometry (3-5 µg/million cells), Immunocytochemistry (1µg/mL), Immunofluorescence (1µg/mL), Western Blot (1:500-1:2000)
Alternate names:
Paired helical filament-tau; Neurofibrillary tangle protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
44740G 306.00
Check your price
  • Acetyl-CBP/p300 (Lys1499, Lys1535) Antibody
  • Host Rabbit
  • Target Species Non-human primate, Rat, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA516185
  • Size 100 µL
Applications
ChIP assay (1:25), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
E1A-associated protein p300
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516185 349.00
Check your price
  • CaMKII alpha Antibody (Monoclonal, 6G9)
  • Host Mouse
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1048
  • Size 100 µL
Applications
Immunofluorescence (1:100), Immunohistochemistry (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (0.5-1 µg/mL)
Alternate names:
Calcium/calmodulin-dependent protein kinase type 2, CaMKII
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1048 375.00
Check your price
  • Human PKM2 Recombinant Protein
  • Host E. Coli
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP75742
  • Size 10 µg
Applications
Control (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Tumor Type M2 Pyruvate Kinase
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP75742 225.00
Check your price
  • Rabies Virus Antibody (Monoclonal, 1C5)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA516456
  • Size 200 ug
Applications
ELISA (Assay Dependent), Functional Assay (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516456 399.00
Check your price
  • Donkey anti-Goat IgG (H+L) Cross-Adsorbed, Alexa Fluor® 546
  • Host Donkey
  • Target Species Goat
  • Conjugate Alexa Fluor® 546
  • Catalog # A11056
  • Size 500 µL
Applications
Flow Cytometry (1-10 µg/mL), Immunofluorescence (1-10 µg/mL)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
A11056 209.00
Check your price
  • Human alpha-Thrombin (DFP) Native Protein
  • Host Human
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # RP43101
  • Size 100 µg
Applications
Control (Assay Dependent)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
RP43101 102.00
Check your price
  • Glucocorticoid Receptor Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Reptile, Rat
  • Conjugate Unconjugated
  • Catalog # PA1510A
  • Size 200 µL
Applications
Gel Shift (Assay dependent), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (1:250), Immunoprecipitation (Assay dependent), Western Blot (1:100)
Alternate names:
Nuclear receptor subfamily 3 group C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1510A 375.00
Check your price
  • Chikungunya Virus Antibody (Monoclonal, A54Q)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA518181
  • Size 100 µg
Applications
ELISA (1:20-1:200), Immunofluorescence (1:10-1:50)
Alternate names:
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518181 287.00
Check your price