Sort by: Popularity  Favorites  Price  Brand  Title 
Page: 1  10  19  20  21  22  23  24  25  26  27  28  29  30  31  32  33  34  35  36  37  38  39  40  41  42  43  44  45  46  47  48  50  60  67 
991 - 1020 of 2000 products displayed
  • AspRS Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530666
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Aspartyl-tRNA synthetase
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530666 349.00
Check your price
  • ARFGAP1/ARFGAP3 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA528707
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
ARF1-directed GTPase-activating protein; ADP-ribosylation factor 1 GTPase-activating protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA528707 349.00
Check your price
  • ARD1A Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA532236
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Immunoprecipitation (Assay-Dependent), Western Blot (1:1000-1:10,000)
Alternate names:
NatA catalytic subunit Naa10; N-terminal acetyltransferase complex ARD1 subunit homolog A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532236 349.00
Check your price
  • Annexin A3 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534829
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Placental anticoagulant protein III; Lipocortin III; Inositol 1,2-cyclic phosphate 2-phosphohydrolase; Annexin-3; Annexin III; 35-alpha calcimedin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534829 349.00
Check your price
  • AMIGO1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530696
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Alivin-2; AMIGO-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530696 349.00
Check your price
  • AKAP1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527736
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Spermatid A-kinase anchor protein 84; Protein kinase A-anchoring protein 1; Dual specificity A-kinase-anchoring protein 1; A-kinase anchor protein 149 kDa
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527736 349.00
Check your price
  • AGPS Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527401
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Alkylglycerone-phosphate synthase; Aging-associated gene 5 protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527401 349.00
Check your price
  • Adenylate Kinase 7 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529375
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
ATP-AMP transphosphorylase 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529375 349.00
Check your price
  • XBP1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520218
  • Size 100 µg
Applications
Immunocytochemistry (10 µg/ml), Western Blot (0.5-2 µg/ml)
Alternate names:
Tax-responsive element-binding protein 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520218 349.00
Check your price
  • SMAD7 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Sheep, Rat
  • Conjugate Unconjugated
  • Catalog # PA523118
  • Size 100 µg
Applications
Western Blot (2-5 µg/ml)
Alternate names:
SMAD family member 7; Mothers against decapentaplegic homolog 8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523118 349.00
Check your price
  • PSD93 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1043
  • Size 100 µg
Applications
Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay-dependent), Immunohistochemistry (Frozen) (5 µg/ml), Western Blot (3 µg/ml)
Alternate names:
Postsynaptic density protein PSD-93; Channel-associated protein of synapse-110
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1043 349.00
Check your price
  • PRC1 Antibody (Monoclonal, 16F2)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA1846
  • Size 100 µg
Applications
Immunocytochemistry (1-2 µg/ml), Immunofluorescence (1-2 µg/ml), Immunoprecipitation (Assay dependent), Western Blot (1-3 µg/ml)
Alternate names:
Protein regulator of cytokinesis 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1846 349.00
Check your price
  • PHD2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA116524
  • Size 100 µg
Applications
Flow Cytometry (3 µg/ml), Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Paraffin) (1:10-1:500), Immunoprecipitation (Assay-Dependent), Western Blot (1:500-1:2500)
Alternate names:
SM-20; Prolyl hydroxylase domain-containing protein 2; Hypoxia-inducible factor prolyl hydroxylase 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116524 349.00
Check your price
  • PD-1 Antibody (Monoclonal, 7A11B1)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA515780
  • Size 100 µg
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
CD279, PD-1, PD1, SLEB2, hPD-1, hPD-l, programmed cell death protein 1, protein PD-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515780 349.00
Check your price
  • PCQAP Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA519081
  • Size 100 µg
Applications
Western Blot (1-3 ug/ml)
Alternate names:
Trinucleotide repeat-containing gene 7 protein; TPA-inducible gene 1 protein; Positive cofactor 2 glutamine/Q-rich-associated protein; Mediator complex subunit 15; CTG repeat protein 7a; Activator-recruited cofactor 105 kDa component
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519081 349.00
Check your price
  • P-cadherin Antibody (Monoclonal, PCD-1)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # 132000Z
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Inhibition Assays (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
Placental cadherin
Catalog # Size Price (AUD) Your Price (AUD) Qty
132000Z 347.00
Check your price
  • OTUD5 Antibody
  • Host Rabbit
  • Target Species Rat, Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520611
  • Size 100 µg
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (2.5 µg/ml), Western Blot (1-2 ug/ml)
Alternate names:
Deubiquitinating enzyme A
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520611 349.00
Check your price
  • Nogo-C Antibody
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547773
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (1 µg/ml)
Alternate names:
Reticulon-5; RTN-x; Neuroendocrine-specific protein C homolog; Neuroendocrine-specific protein; Neurite outgrowth inhibitor; Foocen
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547773 349.00
Check your price
  • NMDAR1 Antibody
  • Host Rabbit
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA534599
  • Size 100 µg
Applications
Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (PFA fixed) (Assay-Dependent), Western Blot (0.5-2 µg/ml)
Alternate names:
N-methyl-D-aspartate receptor subunit NR1; Glutamate [NMDA] receptor subunit zeta-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534599 349.00
Check your price
  • NFATC4 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA1021
  • Size 100 µg
Applications
Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (1:3000), Immunoprecipitation (3 µg), Western Blot (1:1000)
Alternate names:
T-cell transcription factor NFAT3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1021 349.00
Check your price
  • MRP4 Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA519596
  • Size 100 µg
Applications
Western Blot (1 µg/ml)
Alternate names:
Multi-specific organic anion transporter B; MRP/cMOAT-related ABC transporter; ATP-binding cassette sub-family C member 4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519596 349.00
Check your price
  • MEP1A Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547467
  • Size 100 µg
Applications
Flow Cytometry (0.25 µg/10^6 cells), Immunohistochemistry (5-15 µg/ml), Immunoprecipitation (25 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
PPH alpha; PABA peptide hydrolase; N-benzoyl-L-tyrosyl-P-amino-benzoic acid hydrolase subunit alpha; Endopeptidase-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547467 349.00
Check your price
  • MECP2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA1888
  • Size 100 µg
Applications
Western Blot (0.5 µg/ml)
Alternate names:
MeCP2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1888 349.00
Check your price
  • MDA5 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA520337
  • Size 100 µg
Applications
Immunocytochemistry (20 ug/ml), Immunofluorescence (20 ug/ml), Immunohistochemistry (5 µg/ml), Western Blot (1-4 µg/ml)
Alternate names:
RNA helicase-DEAD box protein 116; RIG-I-like receptor 2; Murabutide down-regulated protein; Melanoma differentiation-associated protein 5; Interferon-induced with helicase C domain protein 1; Helicase with 2 CARD domains; Clinically amyopathic dermatomyositis autoantigen 140 kDa; Interferon induced with helicase C domain protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA520337 349.00
Check your price
  • LIGHT Antibody
  • Host Goat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # PA547203
  • Size 100 µg
Applications
Western Blot (0.1 µg/ml)
Alternate names:
TNFSF14, tumor necrosis factor (ligand) superfamily, member 14, CD258, HVEML, LIGHT, LTg, TR2, delta transmembrane LIGHT, herpesvirus entry mediator A, ligand for herpesvirus entry mediator, tumor necrosis factor ligand superfamily, member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547203 349.00
Check your price
  • Ku80 Antibody (Monoclonal, 9403)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA123314
  • Size 100 µg
Applications
Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Western Blot (1:1000-1:5000)
Alternate names:
X-ray repair complementing defective repair in Chinese hamster cells 5 (double-strand-break rejoining); Thyroid-lupus autoantigen; Nuclear factor IV; Lupus Ku autoantigen protein p86; Ku86; Ku80; DNA repair protein XRCC5; CTC box-binding factor 85 kDa subunit; ATP-dependent DNA helicase II 80 kDa subunit; ATP-dependent DNA helicase 2 subunit 2; 86 kDa subunit of Ku antigen
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA123314 349.00
Check your price
  • ITGB1BP1 Antibody
  • Host Sheep
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547878
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (5-15 µg/ml), Western Blot (2 µg/ml)
Alternate names:
Integrin cytoplasmic domain-associated protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547878 349.00
Check your price
  • IL1RL2 Antibody
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA547770
  • Size 100 µg
Applications
Western Blot (0.1 µg/ml)
Alternate names:
Interleukin-1 receptor-related protein 2; IL-36 receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547770 349.00
Check your price
  • IL-12 p40 Antibody
  • Host Goat
  • Target Species Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518741
  • Size 100 µg
Applications
Western Blot (1-3 ug/ml)
Alternate names:
IL-12 subunit p40; Cytotoxic lymphocyte maturation factor 40 kDa subunit
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518741 349.00
Check your price
  • IFNGR2 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA547938
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells), Neutralization (0.5-2.5 µg/ml), Western Blot (0.1 µg/ml)
Alternate names:
Interferon gamma transducer 1; Interferon gamma receptor accessory factor 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA547938 349.00
Check your price