1 - 30 of 30 products displayed
  • ABCA1 Antibody
  • Host Rabbit
  • Target Species Hamster, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA132129
  • Size 100 µL
Applications
Immunocytochemistry (Assay dependent), Immunofluorescence (Assay dependent), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (Assay dependent), Western Blot (1:500)
Alternate names:
Cholesterol efflux regulatory protein; ATP-binding cassette transporter 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA132129 425.00
Check your price
  • Abca1 Antibody (Monoclonal, 5A1-1422.11)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA116937
  • Size 100 µL
Applications
Flow Cytometry (1:1000-1:3000)
Alternate names:
ATP-binding cassette transporter 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116937 375.00
Check your price
  • Abca1 Antibody (Monoclonal, 3A1.891.3)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA116936
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1:1000-1:3000)
Alternate names:
ATP-binding cassette transporter 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116936 375.00
Check your price
  • ABCA1 Antibody (Monoclonal, AB1.G6)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA191401
  • Size 100 µg
Applications
ELISA (3-6 µg/ml), Immunohistochemistry (Frozen) (Assay dependent), Immunoprecipitation (Assay dependent), Western Blot (1:1000-1:2000)
Alternate names:
Cholesterol efflux regulatory protein; ATP-binding cassette transporter 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA191401 399.00
Check your price
  • ABCA7 Antibody (Monoclonal, 7A1-144)
  • Host Rat
  • Target Species Mouse
  • Conjugate
  • Catalog # MA116935
  • Size 100µL
Applications
Flow Cytometry (1:500-1:4000), Western Blot (1:2000)
Alternate names:
ABC 1 antibody, ABC Transporter 1 antibody, ABC1 antibody, ATP-binding Cassette 1 antibody, ATP-binding Cassette Transporter 1 antibody, CERP antibody
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA116935 309.00
Check your price
  • ABCA1 Antibody, DyLight® 488
  • Host Rabbit
  • Target Species Dog, Chicken, Hamster, Human, Mustelid, Mouse, Pig, Rat
  • Conjugate DyLight® 488
  • Catalog # PA522908
  • Size 100 µL
Applications
Flow Cytometry (1:10-1:1000), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (1:10-1:500), Western Blot (1:500)
Alternate names:
Cholesterol efflux regulatory protein; ATP-binding cassette transporter 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522908 375.00
Check your price
  • ABCA1 Antibody
  • Host Rabbit
  • Target Species Dog, Chicken, Hamster, Human, Mustelid, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # PA116789
  • Size 100 µL
Applications
ChIP assay (Assay-Dependent), ELISA (Assay-Dependent), Flow Cytometry (1:400), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Frozen) (Assay-Dependent), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (1:10-1:500), Western Blot (1:500)
Alternate names:
Cholesterol efflux regulatory protein; ATP-binding cassette transporter 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA116789 375.00
Check your price
  • ABCA1 Antibody, Biotin
  • Host Rabbit
  • Target Species Dog, Chicken, Hamster, Human, Mustelid, Mouse, Pig, Rat
  • Conjugate Biotin
  • Catalog # PA522906
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (1:10-1:500), Western Blot (1:500)
Alternate names:
Cholesterol efflux regulatory protein; ATP-binding cassette transporter 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522906 375.00
Check your price
  • ABCA1 Antibody, HRP
  • Host Rabbit
  • Target Species Dog, Chicken, Hamster, Human, Mustelid, Mouse, Pig, Rat
  • Conjugate HRP
  • Catalog # PA522909
  • Size 100 µL
Applications
Immunoprecipitation (1:10-1:500), Western Blot (1:500)
Alternate names:
Cholesterol efflux regulatory protein; ATP-binding cassette transporter 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522909 375.00
Check your price
  • ABCA1 Antibody (Monoclonal, HJ1)
  • Host Mouse
  • Target Species Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA516026
  • Size 100 µL
Applications
Flow Cytometry (1 µg/10^6 cells), Immunohistochemistry (Paraffin) (1:400), Western Blot (1:2000)
Alternate names:
Cholesterol efflux regulatory protein; ATP-binding cassette transporter 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516026 349.00
Check your price
  • ABCA1 Antibody, DyLight® 650
  • Host Rabbit
  • Target Species Dog, Chicken, Hamster, Human, Mustelid, Mouse, Pig, Rat
  • Conjugate DyLight® 650
  • Catalog # PA522907
  • Size 100 µL
Applications
Flow Cytometry (1:10-1:1000), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (1:10-1:500), Western Blot (1:500)
Alternate names:
Cholesterol efflux regulatory protein; ATP-binding cassette transporter 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522907 375.00
Check your price
  • ABCA1 Antibody, DyLight® 550
  • Host Rabbit
  • Target Species Dog, Chicken, Hamster, Human, Mustelid, Mouse, Pig, Rat
  • Conjugate DyLight® 550
  • Catalog # PA522910
  • Size 100 µL
Applications
Flow Cytometry (1:10-1:1000), Immunocytochemistry (1:100), Immunofluorescence (1:100), Immunohistochemistry (Paraffin) (1:200), Immunoprecipitation (1:10-1:500), Western Blot (1:500)
Alternate names:
Cholesterol efflux regulatory protein; ATP-binding cassette transporter 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA522910 375.00
Check your price
  • ABCA9 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518953
  • Size 100 ug
Applications
Western Blot (0.1-0.3 ug/ml)
Alternate names:
ABCA9; ATP-binding cassette, sub-family A (ABC1), member 9; DKFZp686F2450; EST640918; MGC75415; ATP-binding cassette A9; ATP-binding cassette, sub-family A, member 9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518953 349.00
Check your price
  • ABCA5 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA542402
  • Size 100 µl
Applications
Immunohistochemistry (5 µg/ml), Western Blot (1 µg/ml)
Alternate names:
ATP-binding cassette, sub-family A (ABC1) member 5, ABC13, DKFZp451F117, DKFZp779N2435, EST90625, FLJ16381
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542402 309.00
Check your price
  • CABC1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513906
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
aarF domain-containing protein kinase 3; Chaperone activity of bc1 complex-like
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513906 349.00
Check your price
  • ABCA7 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA534396
  • Size 100 ug
Applications
Immunocytochemistry (5 ug/mL), Immunofluorescence (5 ug/mL), Western Blot (1-2 ug/mL)
Alternate names:
Macrophage ABC transporter; Autoantigen SS-N; ABCA-SSN
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534396 349.00
Check your price
  • ABCA5 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537131
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
ATP-binding cassette sub-family A member 5, KIAA1888
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537131 309.00
Check your price
  • ABCA8 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536429
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
ATP-binding cassette sub-family A member 8, KIAA0822
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536429 309.00
Check your price
  • CABC1 Antibody (Monoclonal, 216CT7.5.1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA518109
  • Size 400 µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:100-1:250)
Alternate names:
aarF domain-containing protein kinase 3; Chaperone activity of bc1 complex-like
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA518109 349.00
Check your price
  • CABC1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA529125
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
aarF domain-containing protein kinase 3; Chaperone activity of bc1 complex-like
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529125 349.00
Check your price
  • ABCA13 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA542406
  • Size 100 µl
Applications
Western Blot (1 µg/ml )
Alternate names:
ATP-binding cassette, sub-family A (ABC1) member 13
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542406 309.00
Check your price
  • ABCA8 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA539040
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
ATP-binding cassette sub-family A member 8, KIAA0822
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539040 309.00
Check your price
  • POLR2E Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530123
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
XAP4; RPB5 homolog; DNA-directed RNA polymerase II subunit E; DNA-directed RNA polymerase II 23 kDa polypeptide
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530123 349.00
Check your price
  • ABCA4 Antibody (Monoclonal, 3F4)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Xenopus
  • Conjugate Unconjugated
  • Catalog # P21933
  • Size 100µL
Applications
Immunohistochemistry (1:100), Western Blot (1:1,000)
Alternate names:
RIM ABC transporter; ATP-binding cassette sub-family A member 4; Stargardt disease protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
P21933 299.00
Check your price
  • ABCA12 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA542400
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
ATP-binding cassette transporter 12
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542400 Not Orderable Online
Check your price
  • ABCA10 Antibody (Monoclonal, 8F4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA521013
  • Size 100 µg
Applications
ELISA (Assay-Dependent), Western Blot (1-5 µg/ml)
Alternate names:
FKHL6, FREAC2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA521013 309.00
Check your price
  • MRP1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513543
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Leukotriene C(4) transporter; ATP-binding cassette sub-family C member 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513543 349.00
Check your price
  • ABCD2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA526632
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Adrenoleukodystrophy-related protein; Adrenoleukodystrophy-like 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526632 349.00
Check your price
  • ABCF1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA526286
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
TNF-alpha-stimulated ABC protein; ATP-binding cassette 50
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526286 349.00
Check your price
  • ABCB7 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513108
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
ATP-binding cassette transporter 7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513108 349.00
Check your price