1 - 8 of 8 products displayed
  • Adenovirus Type 5 Hexon Antibody, FITC
  • Host Goat
  • Target Species Virus
  • Conjugate FITC
  • Catalog # PA173053
  • Size 1 mL
Applications
Immunofluorescence (1:10-1:50)
Alternate names:
HADV, AdV, Adeno Virus, HADV-5, AdV-5, Adeno Virus 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173053 375.00
Check your price
  • Adenovirus Type 5 Hexon Antibody, HRP
  • Host Goat
  • Target Species Virus
  • Conjugate HRP
  • Catalog # PA173054
  • Size 1 mL
Applications
ELISA (1:200-1:1,000)
Alternate names:
HADV, AdV, Adeno Virus, HADV-5, AdV-5, Adeno Virus 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173054 375.00
Check your price
  • Adenovirus Type 5 Hexon Antibody (Monoclonal, 65H6)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # LFMA0177
  • Size 100 µL
Applications
Immunocytochemistry (1:500), Western Blot (1:5000)
Alternate names:
HADV, AdV, Adeno Virus, HADV-5, AdV-5, Adeno Virus 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0177 309.00
Check your price
  • Adenovirus Type 5 Hexon Antibody
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA128357
  • Size 100 µL
Applications
ELISA (Assay dependent), Immunocytochemistry (Assay-dependent), Immunofluorescence (Assay dependent)
Alternate names:
HADV, AdV, Adeno Virus, HADV-5, AdV-5, Adeno Virus 5
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA128357 275.00
Check your price
  • Adenovirus Hexon Antibody, FITC (Monoclonal, D36J)
  • Host Mouse
  • Target Species Virus
  • Conjugate FITC
  • Catalog # MA17329
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunofluorescence (1:5)
Alternate names:
HADV, AdV, Adeno Virus, HAdVC_gp08
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17329 349.00
Check your price
  • Adenovirus Hexon Antibody, FITC (Monoclonal, E28H)
  • Host Mouse
  • Target Species Virus
  • Conjugate FITC
  • Catalog # MA17330
  • Size 100 µg
Applications
ELISA (Assay Dependent), Immunofluorescence (1:5)
Alternate names:
HADV, AdV, Adeno Virus, HAdVC_gp08
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17330 349.00
Check your price
  • Adenovirus Type 2 Hexon Antibody, Biotin
  • Host Goat
  • Target Species Virus
  • Conjugate Biotin
  • Catalog # PA173055
  • Size 1 mL
Applications
ELISA (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
HADV, AdV, Adeno Virus, HADV-2, AdV-2, Adeno Virus 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA173055 349.00
Check your price
  • Adenovirus Type 2 Antibody
  • Host Goat
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # PA17201
  • Size 1 mL
Applications
ELISA (Assay dependent), Immunofluorescence (Assay dependent)
Alternate names:
HADV, AdV, Adeno Virus, HADV-2, AdV-2, Adeno Virus 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA17201 309.00
Check your price