1 - 4 of 4 products displayed
  • NGAL Antibody (Monoclonal, 15)
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # ABS0391502
  • Size 200 µL
Applications
ELISA (Assay Dependent)
Alternate names:
p25; Siderocalin LCN2; Lipocalin-2; Alpha-2U globulin-related protein; Alpha-2-microglobulin-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0391502 650.00
Check your price
  • NGAL Antibody, Biotin (Monoclonal)
  • Host Mouse
  • Target Species Rat
  • Conjugate Biotin
  • Catalog # ABS03932B005
  • Size 50 µL
Applications
ELISA (1:4000)
Alternate names:
p25; Siderocalin LCN2; Lipocalin-2; Alpha-2U globulin-related protein; Alpha-2-microglobulin-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS03932B005 475.00
Check your price
  • GLP1R Antibody (Monoclonal, 03)
  • Host Mouse
  • Target Species Mammal
  • Conjugate Unconjugated
  • Catalog # ABS0460302
  • Size 200 µL
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
p25; Siderocalin LCN2; SV-40-induced 24P3 protein; Lipocalin-2; Alpha-2U globulin-related protein; Alpha-2-microglobulin-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
ABS0460302 650.00
Check your price
  • NGAL Antibody
  • Host Goat
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # PA546938
  • Size 100 µg
Applications
ELISA (0.1-0.4 µg/ml (Detection), 0.2-0.8 µg/ml (Capture)), Immunoprecipitation (25 µg/ml), Western Blot (1 µg/ml)
Alternate names:
p25; Siderocalin LCN2; Lipocalin-2; Alpha-2U globulin-related protein; Alpha-2-microglobulin-related protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA546938 375.00
Check your price