1 - 30 of 30 products displayed
  • Smooth Muscle Actin Antibody (Monoclonal, 1A4)
  • Host Mouse
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # MA106110
  • Size 50 µg
Applications
ELISA (Assay dependent), Immunocytochemistry (Assay dependent), Immunofluorescence (1:10-1:100), Immunohistochemistry (Paraffin, Frozen) (1:10-1:100), Immunomicroscopy (Assay dependent), Western Blot (1:10000-1:50000)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA106110 375.00
Check your price
  • Smooth Muscle Actin Antibody (Monoclonal, 0.N.5)
  • Host Mouse
  • Target Species Bovine, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA126017
  • Size 250 µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Frozen) (0.5-1 µg/ml), Immunohistochemistry (Paraffin) (0.5-1 µg/ml)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126017 425.00
Check your price
  • Smooth Muscle Actin Antibody (Monoclonal, 1A4 (asm-1))
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA511547
  • Size 500 µL
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:800), Western Blot (1:100-1:500)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511547 299.00
Check your price
  • Smooth Muscle Actin Antibody
  • Host Rabbit
  • Target Species Bovine, Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA516697
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:25)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516697 299.00
Check your price
  • Smooth Muscle Actin Antibody, Biotin (Monoclonal, 1A4 (asm-1))
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # MA511544
  • Size 500 µL
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:800), Western Blot (1:100-1:500)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511544 349.00
Check your price
  • Smooth Muscle Actin Antibody
  • Host Rabbit
  • Target Species Horse, Human, Mouse, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA519465
  • Size 100 µg
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (1:1000), Immunohistochemistry (Paraffin) (Assay-Dependent), Immunohistochemistry (PFA fixed) (1:200-1:1000), Western Blot (1 µg/ml)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519465 349.00
Check your price
  • Smooth Muscle Actin Antibody (Oligoclonal, 17HCLC)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 710487
  • Size 100 µg
Applications
Flow Cytometry (2-4µg x 10^6 cells), Immunocytochemistry (2-4µg), Immunofluorescence (2-4µg), Immunohistochemistry (1-2 ug/ml), Western Blot (1-3µg/ml)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
710487 304.00
Check your price
  • Smooth Muscle Actin Antibody (Monoclonal, 17H19L35)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 701457
  • Size 100 µg
Applications
Flow Cytometry (2-4µg x 10^6 cells), Immunocytochemistry (2-4µg), Immunofluorescence (2-4µg), Immunohistochemistry (Paraffin) (1:50-1:500), Western Blot (1-3µg/ml)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
701457 306.00
Check your price
  • Actin Antibody (Monoclonal, HHF35)
  • Host Mouse
  • Target Species Dog, Chicken, Cat, Human, Mouse, Non-human primate, Rabbit, Rat, Shrew
  • Conjugate Unconjugated
  • Catalog # MA182599
  • Size 100µL
Applications
Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (1/10-1/40)
Alternate names:
Smooth muscle gamma-actin; Gamma-2-actin; Alpha-actin-3; Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-cardiac actin; Alpha-actin; Alpha-actin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182599 199.00
Check your price
  • Skeletal Muscle Actin Antibody (Monoclonal, 5C5.F8.C7 (alpha-Sr-1))
  • Host Mouse
  • Target Species Amphibian, Bovine, Guinea pig, Human, Mouse, Sheep, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA512542
  • Size 500 µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Alpha-actin-1; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512542 275.00
Check your price
  • Smooth Muscle Actin Antibody (Monoclonal, 1A4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA137027
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA137027 375.00
Check your price
  • Smooth Muscle Actin Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137024
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137024 375.00
Check your price
  • Smooth Muscle Actin Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532587
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532587 349.00
Check your price
  • Smooth Muscle Actin Antibody (Monoclonal, 1A4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA137028
  • Size 7 mL
Applications
Immunohistochemistry (Paraffin) (Ready-to-use)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA137028 225.00
Check your price
  • Smooth Muscle Actin Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137023
  • Size 7 mL
Applications
Immunohistochemistry (Paraffin) (Ready-to-use)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137023 225.00
Check your price
  • Smooth Muscle Actin Antibody (Monoclonal, a-SM1)
  • Host Mouse
  • Target Species Chicken, Human, Rat
  • Conjugate Unconjugated
  • Catalog # MA112772
  • Size 50 µg
Applications
ELISA (Assay Dependent), Immunocytochemistry (Assay Dependent), Immunohistochemistry (1:1000-1:10000), Immunomicroscopy (Assay Dependent), Western Blot (1:10000-1:50000)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112772 425.00
Check your price
  • Actin Antibody, Biotin (Monoclonal, ACTN05 (C4))
  • Host Mouse
  • Target Species Bovine, Dog, Chicken, Human, Mouse, Non-human primate, Plant, Pig, Protozoa, Rabbit, Rat
  • Conjugate Biotin
  • Catalog # MA511866
  • Size 500 µL
Applications
Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (1:200-1:400), Immunoprecipitation (2 ug), Western Blot (1:5000-1:20,000)
Alternate names:
Smooth muscle gamma-actin; Gamma-2-actin; Alpha-actin-3; Beta-actin; Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin-1; Alpha-cardiac actin; Alpha-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511866 399.00
Check your price
  • Sarcomeric alpha Actin Antibody (Monoclonal, 5C5)
  • Host Mouse
  • Target Species Bovine, Fish, Guinea pig, Human, Mouse, Sheep, Pig, Rabbit, Reptile, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA126928
  • Size 500 µL
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (1:25-1:50), Immunohistochemistry (Paraffin) (1:25-1:50)
Alternate names:
Alpha-cardiac actin; Actin alpha 1; Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126928 399.00
Check your price
  • Smooth Muscle Actin Antibody
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518292
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (2-4 µg/ml), Western Blot (0.3-1 µg/ml)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518292 349.00
Check your price
  • Smooth Muscle Actin Antibody (Monoclonal, 1H8)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515805
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515805 349.00
Check your price
  • Smooth Muscle Actin Antibody (Monoclonal, 4A4)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515871
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515871 349.00
Check your price
  • Sarcomeric alpha Actin Antibody (Monoclonal, 5C5)
  • Host Mouse
  • Target Species Bovine, Fish, Guinea pig, Human, Sheep, Pig, Rabbit, Reptile, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA121597
  • Size 6 mL
Applications
Immunohistochemistry (Frozen) (Neat), Immunohistochemistry (Paraffin) (Neat)
Alternate names:
Alpha-cardiac actin; Actin alpha 1; Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121597 349.00
Check your price
  • Actin Antibody (Monoclonal, mAbGEa)
  • Host Mouse
  • Target Species Bovine, Fruit fly, Human, Mouse, Sheep, Plant, Rat, Xenopus, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA1744
  • Size 100 µL
Applications
ELISA (Assay dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (1:100), Western Blot (1:1,000)
Alternate names:
Beta-actin; Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin-1; Cytoplasmic beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1744 375.00
Check your price
  • Actin Antibody (Monoclonal, ACTN05 (C4))
  • Host Mouse
  • Target Species Bovine, Dog, Chicken, Human, Mouse, Non-human primate, Plant, Pig, Protozoa, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA511869
  • Size 500 µL
Applications
Immunocytochemistry (1:10-1:200), Immunofluorescence (1:10-1:200), Immunohistochemistry (Paraffin) (1:50), Immunoprecipitation (2 ug), Western Blot (1:2500-1:10,000)
Alternate names:
Alpha-actin-1; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511869 309.00
Check your price
  • Skeletal Muscle Actin Antibody (Monoclonal, 5C5.F8.C7)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA137021
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:50)
Alternate names:
Alpha-actin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA137021 399.00
Check your price
  • Skeletal Muscle Actin Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137020
  • Size 1 mL
Applications
Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Alpha-actin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137020 275.00
Check your price
  • Skeletal Muscle Actin Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA137019
  • Size 7 mL
Applications
Immunohistochemistry (Paraffin) (Ready-to-use)
Alternate names:
Alpha-actin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA137019 199.00
Check your price
  • Smooth Muscle Actin Antibody (Monoclonal, 4F4)
  • Host Mouse
  • Target Species Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA515806
  • Size 100 µL
Applications
Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515806 349.00
Check your price
  • Actin Antibody (Monoclonal, HHF35)
  • Host Mouse
  • Target Species Dog, Chicken, Cat, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA110046
  • Size 100 ug
Applications
Immunohistochemistry (Frozen) (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent), Western Blot (1 ug/ml)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Gamma-actin; Alpha-cardiac actin; Alpha-actin; Alpha-actin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA110046 249.00
Check your price
  • Actin Antibody, FITC (Monoclonal, AC-40)
  • Host Mouse
  • Target Species Bovine, Dog, Fish, Guinea pig, Goat, Hamster, Human, Mouse, Sheep, Pig, Rabbit, Rat, Xenopus
  • Conjugate FITC
  • Catalog # MA124908
  • Size 50 µL
Applications
Immunocytochemistry (1:50), Immunofluorescence (1:50), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA124908 349.00
Check your price