1 - 4 of 4 products displayed
  • Adiponectin Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA184881
  • Size 100 µg
Applications
Flow Cytometry (Assay dependent), Immunocytochemistry (10 ug/ml), Immunohistochemistry (Paraffin) (10 ug/ml), Western Blot (1-2 ug/ml)
Alternate names:
Gelatin-binding protein; Adipose most abundant gene transcript 1 protein; Adipocyte, C1q and collagen domain-containing protein; Adipocyte complement-related 30 kDa protein; 30 kDa adipocyte complement-related protein; Adipocyte-specific protein AdipoQ
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA184881 375.00
Check your price
  • CD93 Antibody, APC (Monoclonal, 273107)
  • Host Mouse
  • Target Species Human
  • Conjugate APC
  • Catalog # MA523615
  • Size 100 Tests
Applications
Flow Cytometry (10 µl/10^6 cells)
Alternate names:
Matrix-remodeling-associated protein 4; Complement component 1 q subcomponent receptor 1; CDw93; C1q/MBL/SPA receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA523615 299.00
Check your price
  • Complement C3b Antibody (Monoclonal, 10C7)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA170054
  • Size 250 ug
Applications
ELISA (Assay dependent), Flow Cytometry (1 ug/1 million cells)
Alternate names:
HSE-MSF; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA170054 399.00
Check your price
  • Calreticulin Antibody
  • Host Rabbit
  • Target Species Dog, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA3900
  • Size 100 µL
Applications
Flow Cytometry (Assay Dependent), Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunoprecipitation (Assay Dependent), Western Blot (1:1000)
Alternate names:
HACBP; Endoplasmic reticulum resident protein 60; Calregulin; CRP55; Calcium-binding protein 3; CALBP; grp60
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA3900 375.00
Check your price