1 - 30 of 30 products displayed
  • Actin Antibody (Monoclonal, mAbGEa)
  • Host Mouse
  • Target Species Bovine, Fruit fly, Human, Mouse, Sheep, Plant, Rat, Xenopus, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # MA1744
  • Size 100 µL
Applications
ELISA (Assay dependent), Immunocytochemistry (1:10-1:100), Immunofluorescence (1:10-1:100), Immunohistochemistry (1:100), Western Blot (1:1,000)
Alternate names:
Beta-actin; Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin-1; Cytoplasmic beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1744 375.00
Check your price
  • beta Actin Antibody (Monoclonal, 8H10D10)
  • Host Mouse
  • Target Species Hamster, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515452
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1:200-1:1000), Immunofluorescence (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515452 349.00
Check your price
  • beta Actin Antibody (Monoclonal, 15G5A11/E2)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA1140
  • Size 100 ug
Applications
Immunocytochemistry (1:1000-1:4000), Immunofluorescence (1:1000-1:4000), Immunoprecipitation (2ug), Western Blot (1:5000-1:20,000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA1140 219.00
Check your price
  • beta Actin Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA1183
  • Size 100 µg
Applications
Immunocytochemistry (1:50-1:200), Immunofluorescence (1:50-1:200), Immunoprecipitation (2 µg), Western Blot (1:500-1:2000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1183 225.00
Check your price
  • beta Actin Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Chicken, Fruit fly, Cat, Guinea pig, Human, Leech, Mouse, Sheep, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA516914
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (2-4 µg/ml)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516914 349.00
Check your price
  • beta Actin Antibody
  • Host Rabbit
  • Target Species Many
  • Conjugate Unconjugated
  • Catalog # PA121167
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100), Western Blot (Assay dependent)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA121167 399.00
Check your price
  • beta Actin Antibody
  • Host Rabbit
  • Target Species Avian, Fish, Hamster, Human, Mouse, Non-human primate, Pig, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA146296
  • Size 50 µL
Applications
Blocking Assay (Assay-Dependent), ELISA (Assay-Dependent), Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Paraffin) (1:100), Immunoprecipitation (1:10-1:500), Neutralization (Assay-Dependent), Western Blot (1:100-1:2000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA146296 375.00
Check your price
  • beta Actin Antibody (Monoclonal, AC-15)
  • Host Mouse
  • Target Species Bovine, Dog, Chicken, Fruit fly, Cat, Guinea pig, Human, Sheep, Pig, Rabbit
  • Conjugate Unconjugated
  • Catalog # AM4302
  • Size 100 µg
Applications
Immunocytochemistry (Assay Dependent), Immunofluorescence (Assay Dependent)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
AM4302 268.00
Check your price
  • Actin Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA511570
  • Size 400 µL
Applications
Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (1:10-1:50), Western Blot (1:1000)
Alternate names:
Beta-actin; Alpha-cardiac actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA511570 349.00
Check your price
  • Sarcomeric alpha Actin Antibody (Monoclonal, 5C5)
  • Host Mouse
  • Target Species Bovine, Fish, Guinea pig, Human, Mouse, Sheep, Pig, Rabbit, Reptile, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA126928
  • Size 500 µL
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (1:25-1:50), Immunohistochemistry (Paraffin) (1:25-1:50)
Alternate names:
Alpha-cardiac actin; Actin alpha 1; Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA126928 399.00
Check your price
  • Smooth Muscle Actin Antibody
  • Host Rabbit
  • Target Species Horse, Human, Mouse, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA519465
  • Size 100 µg
Applications
Immunocytochemistry (Assay-Dependent), Immunofluorescence (Assay-Dependent), Immunohistochemistry (Frozen) (1:1000), Immunohistochemistry (Paraffin) (Assay-Dependent), Immunohistochemistry (PFA fixed) (1:200-1:1000), Western Blot (1 µg/ml)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519465 349.00
Check your price
  • Smooth Muscle Actin Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA532587
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:100)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532587 349.00
Check your price
  • beta Actin Antibody (Monoclonal, SP124)
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA516410
  • Size 100 µL
Applications
Flow Cytometry (1:100), Immunohistochemistry (Paraffin) (1:200), Western Blot (1:25)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA516410 225.00
Check your price
  • Smooth Muscle Actin Antibody (Monoclonal, 1A4 (asm-1))
  • Host Mouse
  • Target Species Bovine, Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA511547
  • Size 500 µL
Applications
Immunocytochemistry (1:100-1:500), Immunofluorescence (1:100-1:500), Immunohistochemistry (Paraffin) (1:800), Western Blot (1:100-1:500)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511547 299.00
Check your price
  • Smooth Muscle Actin Antibody
  • Host Rabbit
  • Target Species Bovine, Chicken, Human, Mouse, Non-human primate, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # PA516697
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:25)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA516697 299.00
Check your price
  • Smooth Muscle Actin Antibody (Oligoclonal, 17HCLC)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 710487
  • Size 100 µg
Applications
Flow Cytometry (2-4µg x 10^6 cells), Immunocytochemistry (2-4µg), Immunofluorescence (2-4µg), Immunohistochemistry (1-2 ug/ml), Western Blot (1-3µg/ml)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
710487 304.00
Check your price
  • Smooth Muscle Actin Antibody (Monoclonal, 17H19L35)
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # 701457
  • Size 100 µg
Applications
Flow Cytometry (2-4µg x 10^6 cells), Immunocytochemistry (2-4µg), Immunofluorescence (2-4µg), Immunohistochemistry (Paraffin) (1:50-1:500), Western Blot (1-3µg/ml)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
701457 306.00
Check your price
  • beta Actin Loading Control Antibody (Monoclonal, BA3R)
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA5157391MG
  • Size 1 mg
Applications
ELISA (Assay dependent), Flow Cytometry (1-2 ug/test), Western Blot (1:1000-1:10,000)
Alternate names:
Beta-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA5157391MG 950.00
Check your price
  • Smooth Muscle Actin Antibody
  • Host Goat
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA518292
  • Size 100 µg
Applications
Immunohistochemistry (Paraffin) (2-4 µg/ml), Western Blot (0.3-1 µg/ml)
Alternate names:
Cell growth-inhibiting gene 46 protein; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518292 349.00
Check your price
  • gamma Actin Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513467
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunocytochemistry (1:10-1:50), Immunofluorescence (1:10-1:50), Western Blot (1:1000)
Alternate names:
Gamma-actin
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513467 349.00
Check your price
  • Sarcomeric alpha Actin Antibody (Monoclonal, 5C5)
  • Host Mouse
  • Target Species Bovine, Fish, Guinea pig, Human, Sheep, Pig, Rabbit, Reptile, Rat, Xenopus
  • Conjugate Unconjugated
  • Catalog # MA121597
  • Size 6 mL
Applications
Immunohistochemistry (Frozen) (Neat), Immunohistochemistry (Paraffin) (Neat)
Alternate names:
Alpha-cardiac actin; Actin alpha 1; Cell growth-inhibiting gene 46 protein; Alpha-actin-2; Alpha-actin-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA121597 349.00
Check your price
  • Skeletal Muscle Actin Antibody (Monoclonal, 5C5.F8.C7 (alpha-Sr-1))
  • Host Mouse
  • Target Species Amphibian, Bovine, Guinea pig, Human, Mouse, Sheep, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA512542
  • Size 500 µL
Applications
Flow Cytometry (3-5 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (1:200)
Alternate names:
Alpha-actin-1; Alpha-actin-2
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512542 275.00
Check your price
  • Phospho-Filamin A (Ser2152) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA540239
  • Size 100 µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Non-muscle filamin; Filamin-1; Endothelial actin-binding protein; Alpha-filamin; Actin-binding protein 280
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540239 309.00
Check your price
  • ACTR3B Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA544774
  • Size 100 µl
Applications
Western Blot (0.2-1 µg/ml)
Alternate names:
Actin-like protein 3B; ARP3-beta; Actin-related protein ARP4
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA544774 309.00
Check your price
  • MTSS1 Antibody
  • Host Rabbit
  • Target Species Dog, Chicken, Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # PA523200
  • Size 100 ug
Applications
Western Blot (2-4 µg/ml)
Alternate names:
Missing in metastasis protein; Metastasis suppressor YGL-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA523200 349.00
Check your price
  • Gelsolin Antibody (Monoclonal, 35B2)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFMA0176
  • Size 100 µL
Applications
Immunoprecipitation (0.5 µl), Western Blot (1:2000)
Alternate names:
Brevin; Actin-depolymerizing factor; AGEL
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0176 309.00
Check your price
  • Gelsolin Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA519848
  • Size 100 µg
Applications
Immunocytochemistry (1 µg/ml), Immunofluorescence (1 µg/ml), Immunohistochemistry (Paraffin) (Assay-Dependent), Western Blot (1 µg/ml)
Alternate names:
Brevin; Actin-depolymerizing factor; AGEL
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519848 349.00
Check your price
  • Arp2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Xenopus
  • Conjugate Unconjugated
  • Catalog # PA519760
  • Size 100 µg
Applications
Immunofluorescence (1 µg/ml), Immunoprecipitation (Assay-Dependent), Western Blot (1 µg/ml)
Alternate names:
Actin-like protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA519760 349.00
Check your price
  • CAPZA1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA514399
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunofluorescence (1:10-1:50), Immunohistochemistry (Paraffin) (0.05 mg/ml), Western Blot (1:1000)
Alternate names:
CapZ alpha-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA514399 349.00
Check your price
  • Filamin A Antibody (Monoclonal, FLMN01 (PM6/317))
  • Host Mouse
  • Target Species Chicken, Guinea pig, Human, Mouse, Rabbit, Rat
  • Conjugate Unconjugated
  • Catalog # MA511705
  • Size 500 µL
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (1-2 µg/ml), Western Blot (1:100-1:500)
Alternate names:
Non-muscle filamin; Filamin-1; Endothelial actin-binding protein; Alpha-filamin; Actin-binding protein 280
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511705 309.00
Check your price