1 - 4 of 4 products displayed
  • Influenza B NP Antibody (Monoclonal, 1B6)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # HYB1160302
  • Size 200 µl
Applications
ELISA (Assay dependent), Immunocytochemistry (1:1000), Immunofluorescence (1:1,000), Immunohistochemistry (Assay dependent), Western Blot (1:40)
Alternate names:
Influenza B Nuclear Protein, Influenza Nuclear Protein, Influenza NP
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1160302 550.00
Check your price
  • Influenza B NP Antibody (Monoclonal, 3E9)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # HYB1160202
  • Size 200 µl
Applications
ELISA (Assay dependent), Immunocytochemistry (1:1000), Immunofluorescence (1:1,000), Western Blot (1:20)
Alternate names:
Influenza B Nuclear Protein, Influenza Nuclear Protein, Influenza NP
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1160202 600.00
Check your price
  • Influenza B NP Antibody (Monoclonal, 1D8)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # HYB1160102
  • Size 200 µl
Applications
ELISA (1:200), Immunocytochemistry (1:1000), Immunofluorescence (1:1000), Western Blot (Assay dependent)
Alternate names:
Influenza B Nuclear Protein, Influenza Nuclear Protein, Influenza NP
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB1160102 600.00
Check your price
  • Influenza A NP Antibody, FITC (Monoclonal, D67J)
  • Host Mouse
  • Target Species Virus
  • Conjugate FITC
  • Catalog # MA17322
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent)
Alternate names:
Influenza nucleoprotein
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA17322 375.00
Check your price