1 - 30 of 30 products displayed
  • NEMF Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536308
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Serologically defined colon cancer antigen 1 homolog; Serologically defined colon cancer antigen 1; Antigen NY-CO-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536308 309.00
Check your price
  • NEMF Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA538539
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Serologically defined colon cancer antigen 1 homolog; Serologically defined colon cancer antigen 1; Antigen NY-CO-1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538539 309.00
Check your price
  • SDCCAG3 Isoform 1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA521723
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Antigen NY-CO-3
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA521723 349.00
Check your price
  • STARD10 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536947
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
StAR-related lipid transfer protein 10; Serologically defined colon cancer antigen 28 homolog; START domain-containing protein 10; Serologically defined colon cancer antigen 28; Antigen NY-CO-28
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536947 309.00
Check your price
  • STUB1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA1015
  • Size 100 µg
Applications
Western Blot (1 ug/ml)
Alternate names:
STIP1 homology and U box-containing protein 1; Carboxy terminus of Hsp70-interacting protein; CLL-associated antigen KW-8; Antigen NY-CO-7; E3 ubiquitin-protein ligase CHIP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA1015 375.00
Check your price
  • HSPH1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536059
  • Size 100 µl
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Heat shock 110 kDa protein; Antigen NY-CO-25; Heat shock-related 100 kDa protein E7I; 42 degrees C-HSP
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536059 309.00
Check your price
  • STARD10 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA539579
  • Size 100 µg
Applications
Western Blot (1:500-1:3000)
Alternate names:
StAR-related lipid transfer protein 10; Serologically defined colon cancer antigen 28 homolog; START domain-containing protein 10; Serologically defined colon cancer antigen 28; Antigen NY-CO-28
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA539579 309.00
Check your price
  • STUB1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA532046
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:5000-1:20,000)
Alternate names:
STIP1 homology and U box-containing protein 1; Carboxy terminus of Hsp70-interacting protein; CLL-associated antigen KW-8; Antigen NY-CO-7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA532046 349.00
Check your price
  • SDCCAG8 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA530408
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Centrosomal colon cancer autoantigen protein; Antigen NY-CO-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530408 349.00
Check your price
  • SDCCAG8 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535971
  • Size 100 µL
Applications
Immunocytochemistry (1:1000-1:10,000), Immunofluorescence (1:1000-1:10,000)
Alternate names:
Centrosomal colon cancer autoantigen protein; Antigen NY-CO-8
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535971 309.00
Check your price
  • Phospho-HDAC5 (Ser498) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA536647
  • Size 100 µl
Applications
Immunofluorescence (1:50-1:200), Immunohistochemistry (Paraffin) (1:50-1:200), Western Blot (1:500-1:1000)
Alternate names:
Antigen NY-CO-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA536647 349.00
Check your price
  • WNK2 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537108
  • Size 100 µl
Applications
Western Blot (1:500-1:1000)
Alternate names:
Serologically defined colon cancer antigen 43; Protein kinase with no lysine 2; Protein kinase lysine-deficient 2; Antigen NY-CO-43
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537108 309.00
Check your price
  • Phospho-HDAC4/HDAC5/HDAC9 (Ser246, Ser259, Ser220) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA537835
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Antigen NY-CO-9; MEF2-interacting transcription repressor MITR; Histone deacetylase-related protein; Histone deacetylase 7B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537835 309.00
Check your price
  • TSHZ1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA527647
  • Size 100 µL
Applications
Western Blot (1:500-1:3000)
Alternate names:
Serologically defined colon cancer antigen 33; Antigen NY-CO-33
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA527647 349.00
Check your price
  • Phospho-HDAC5/HDAC9 (Ser259, Ser220) Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA540215
  • Size 100 µl
Applications
Western Blot (1:500-1:2000)
Alternate names:
Antigen NY-CO-9; MEF2-interacting transcription repressor MITR; Histone deacetylase-related protein; Histone deacetylase 7B
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA540215 309.00
Check your price
  • UTP14A Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538778
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Serologically defined colon cancer antigen 16; Antigen NY-CO-16; Juvenile spermatogonial depletion-like X-linked protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538778 309.00
Check your price
  • TSHZ1 Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA534342
  • Size 100 ug
Applications
Immunocytochemistry (2.5 ug/mL), Immunofluorescence (2.5 ug/mL), Western Blot (1 ug/mL)
Alternate names:
Serologically defined colon cancer antigen 33; Antigen NY-CO-33; Serologically defined colon cancer antigen 3 homolog
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA534342 349.00
Check your price
  • Phospho-HDAC5 (Ser259) Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA538116
  • Size 100 µg
Applications
Western Blot (1:500-1:1000)
Alternate names:
Antigen NY-CO-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA538116 309.00
Check your price
  • Phospho-HDAC5 (Ser498) Antibody
  • Host Rabbit
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA537779
  • Size 100 µl
Applications
Immunohistochemistry (Paraffin) (1:50-1:100), Western Blot (1:500-1:1000)
Alternate names:
Antigen NY-CO-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA537779 309.00
Check your price
  • Phospho-HDAC4/HDAC5/HDAC7 (Ser246, Ser259, Ser155) Antibody (Monoclonal, B.544.8)
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA514776
  • Size 100 µL
Applications
ChIP assay (1-3 ul), Immunoprecipitation (1:50), Western Blot (1:1000)
Alternate names:
Histone deacetylase 7A; Antigen NY-CO-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA514776 309.00
Check your price
  • STUB1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA529024
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
STIP1 homology and U box-containing protein 1; Carboxy terminus of Hsp70-interacting protein; CLL-associated antigen KW-8; Antigen NY-CO-7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA529024 349.00
Check your price
  • STUB1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA513419
  • Size 400 µL
Applications
Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
STIP1 homology and U box-containing protein 1; Carboxy terminus of Hsp70-interacting protein; CLL-associated antigen KW-8; Antigen NY-CO-7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA513419 349.00
Check your price
  • USH1C Antibody
  • Host Goat
  • Target Species Bovine, Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # PA131347
  • Size 50 µg
Applications
ELISA (1:32,000), Immunohistochemistry (Paraffin) (Assay-dependent), Western Blot (1-3 µg/ml)
Alternate names:
Usher syndrome type-1C protein homolog; Usher syndrome type-1C protein; Renal carcinoma antigen NY-REN-3; Protein PDZ-73; Autoimmune enteropathy-related antigen AIE-75; Antigen NY-CO-38/NY-CO-37; PDZ domain-containing protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA131347 475.00
Check your price
  • p53 Antibody (Monoclonal, DO-1)
  • Host Mouse
  • Target Species Human, Rhesus monkey
  • Conjugate Unconjugated
  • Catalog # MA512571
  • Size 500 µL
Applications
ChIP assay (1-3 µg x 10^6 cells), Immunocytochemistry (2µg/ml), Immunofluorescence (2µg/ml), Immunohistochemistry (Paraffin) (1-2 µg/ml), Western Blot (1-2 µg/ml)
Alternate names:
Tumor suppressor p53; Phosphoprotein p53; Antigen NY-CO-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512571 309.00
Check your price
  • HSPH1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA524619
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Immunohistochemistry (1:50-1:100), Western Blot (1:1000)
Alternate names:
Heat shock 110 kDa protein; Antigen NY-CO-25
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA524619 349.00
Check your price
  • HDAC5 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA141117
  • Size 100 µg
Applications
ChIP assay (1:10-1:500), Immunoprecipitation (1:10-1:500), Western Blot (1-3 µg/ml)
Alternate names:
Antigen NY-CO-9
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA141117 375.00
Check your price
  • STUB1 Antibody
  • Host Goat
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA518011
  • Size 100 ug
Applications
Western Blot (0.3-1 ug/ml)
Alternate names:
STIP1 homology and U box-containing protein 1; Carboxy terminus of Hsp70-interacting protein; CLL-associated antigen KW-8; Antigen NY-CO-7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA518011 349.00
Check your price
  • STUB1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA535110
  • Size 400 µL
Applications
Flow Cytometry (1:10-1:50), Western Blot (1:1000)
Alternate names:
STIP1 homology and U box-containing protein 1; Carboxy terminus of Hsp70-interacting protein; CLL-associated antigen KW-8; Antigen NY-CO-7
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535110 349.00
Check your price
  • NOXA1 Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA526572
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
p51-nox; P67phox-like factor; NCF2-like protein; Antigen NY-CO-31
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA526572 349.00
Check your price
  • p53 Antibody (Monoclonal, DO-1)
  • Host Mouse
  • Target Species Human, Rhesus monkey
  • Conjugate Unconjugated
  • Catalog # AHO0152
  • Size 500 µl
Applications
ChIP assay (1-3 µg x 10^6 cells), Immunohistochemistry (Paraffin) (Assay-Dependent), Immunoprecipitation (Assay-Dependent), Western Blot (Assay-Dependent)
Alternate names:
Tumor suppressor p53; Phosphoprotein p53; Antigen NY-CO-13
Catalog # Size Price (AUD) Your Price (AUD) Qty
AHO0152 304.00
Check your price