1 - 2 of 2 products displayed
  • BEGAIN Antibody
  • Host Rabbit
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # PA531568
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
brain-enriched guanylate kinase-associated protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA531568 349.00
Check your price
  • Fyn Antibody (Monoclonal, 1S)
  • Host Mouse
  • Target Species Human, Mouse, Rat
  • Conjugate Unconjugated
  • Catalog # MA112607
  • Size 100 µL
Applications
Immunohistochemistry (Paraffin) (1:50), Western Blot (Assay Dependent)
Alternate names:
p59-Fyn; Src-like kinase; Proto-oncogene c-Fyn; Proto-oncogene Syn
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA112607 499.00
Check your price