1 - 30 of 30 products displayed
  • C3a Receptor Antibody (Monoclonal, 534625)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA524235
  • Size 100 µg
Applications
Flow Cytometry (2.5 µg/10^6 cells)
Alternate names:
C3AR1, AZ3B, C3AR, C3R1, C3a receptor, C3a-R, HNFAG09, Anaphylatoxin c3a receptor, C3a anaphylatoxin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA524235 275.00
Check your price
  • Complement C3a Antibody, Biotin (Monoclonal)
  • Host Mouse
  • Target Species Human
  • Conjugate Biotin
  • Catalog # GAU01701B01
  • Size 100 µL
Applications
ELISA (1:40,000)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
GAU01701B01 700.00
Check your price
  • Complement C3a Antibody (Monoclonal, K13/16-5.7)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # GAU0131602
  • Size 200 µL
Applications
ELISA (1:10,000), Western Blot (Assay dependent)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
GAU0131602 449.00
Check your price
  • Complement C3a Antibody (Monoclonal, D17/1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # GAU0170102
  • Size 200 µL
Applications
ELISA (Assay dependent), Western Blot (Assay dependent)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
GAU0170102 399.00
Check your price
  • Complement C3a Antibody (Monoclonal, 2991)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA140157
  • Size 100 µg
Applications
ELISA (Assay Dependent), Western Blot (1:50)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140157 475.00
Check your price
  • Complement C3a Antibody (Monoclonal, 474)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA140156
  • Size 100 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent.)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140156 425.00
Check your price
  • C3a Receptor Antibody, Biotin (Monoclonal, 74)
  • Host Mouse
  • Target Species Rat
  • Conjugate Biotin
  • Catalog # MA517474
  • Size 100 ug
Applications
Flow Cytometry (2.0 ug/10^6 cells), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
C3AR1, AZ3B, C3AR, C3R1, C3a receptor, C3a-R, HNFAG09, Anaphylatoxin c3a receptor, C3a anaphylatoxin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517474 159.00
Check your price
  • C3a Receptor Antibody, FITC (Monoclonal, 74)
  • Host Mouse
  • Target Species Rat
  • Conjugate FITC
  • Catalog # MA517475
  • Size 100 ug
Applications
Flow Cytometry (2.0 ug/10^6 cells), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
C3AR1, AZ3B, C3AR, C3R1, C3a receptor, C3a-R, HNFAG09, Anaphylatoxin c3a receptor, C3a anaphylatoxin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517475 159.00
Check your price
  • C3a Receptor Antibody, PE (Monoclonal, 74)
  • Host Mouse
  • Target Species Rat
  • Conjugate PE
  • Catalog # MA517476
  • Size 50 ug
Applications
Flow Cytometry (2.0 ug/10^6 cells), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
C3AR1, AZ3B, C3AR, C3R1, C3a receptor, C3a-R, HNFAG09, Anaphylatoxin c3a receptor, C3a anaphylatoxin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517476 159.00
Check your price
  • C3a Receptor Antibody (Monoclonal, 74)
  • Host Mouse
  • Target Species Rat
  • Conjugate Unconjugated
  • Catalog # MA517473
  • Size 250 ug
Applications
Flow Cytometry (2.0 ug/10^6 cells), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
C3AR1, AZ3B, C3AR, C3R1, C3a receptor, C3a-R, HNFAG09, Anaphylatoxin c3a receptor, C3a anaphylatoxin receptor
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA517473 159.00
Check your price
  • Complement C3b Antibody (Monoclonal, 6C9)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA170053
  • Size 250 ug
Applications
ELISA (Assay dependent), Flow Cytometry (1 ug/1 million cells)
Alternate names:
HSE-MSF; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA170053 375.00
Check your price
  • Complement C3b Antibody (Monoclonal, 10C7)
  • Host Mouse
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # MA170054
  • Size 250 ug
Applications
ELISA (Assay dependent), Flow Cytometry (1 ug/1 million cells)
Alternate names:
HSE-MSF; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA170054 399.00
Check your price
  • Complement C3f Antibody (Monoclonal, 8G4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HAV0040102
  • Size 200 µl
Applications
ELISA (1:15,000), Western Blot (Assay dependent)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
HAV0040102 425.00
Check your price
  • Complement C3 Antibody (Monoclonal, 2D8)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB0050102
  • Size 200 µL
Applications
ELISA (1:30,000), Western Blot (Assay-Dependent)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB0050102 399.00
Check your price
  • Complement C3a Antibody (Monoclonal, 2898)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA140158
  • Size 100 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent.)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140158 449.00
Check your price
  • Complement C3b Antibody (Monoclonal, 755)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA140155
  • Size 100 µg
Applications
ELISA (Assay Dependent), Western Blot (Assay Dependent.)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140155 425.00
Check your price
  • Complement C3d Antibody (Monoclonal, 7C10)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HAV0030502
  • Size 200 µl
Applications
ELISA (1:16,000), Western Blot (Assay dependent)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
HAV0030502 425.00
Check your price
  • Complement C3d Antibody (Monoclonal, 6F6)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # HYB0300802
  • Size 200 µl
Applications
ELISA (1:30,000), Western Blot (Assay dependent)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
HYB0300802 449.00
Check your price
  • Complement C3d Antibody (Monoclonal, 053A-1149.3.1.4 (1003))
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA183965
  • Size 100 µL
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (1/5000)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183965 449.00
Check your price
  • Complement C3b Antibody (Monoclonal, 013III-1.16)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA182814
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182814 600.00
Check your price
  • Complement C3d Antibody (Monoclonal, 053A-514.3.1.4)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA183987
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunohistochemistry (Frozen) (Assay Dependent)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA183987 600.00
Check your price
  • Complement C3 Antibody (Monoclonal, 28A1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFMA0132
  • Size 100 µL
Applications
ELISA (Assay dependent), Western Blot (2 ug/mL)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0132 309.00
Check your price
  • Complement C3 Antibody (Monoclonal, 10A1)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # LFMA0131
  • Size 100 µL
Applications
ELISA (Assay dependent), Western Blot (0.5 µg/ml)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
LFMA0131 375.00
Check your price
  • Complement C3b Antibody (Monoclonal, H11)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA182934
  • Size 50 µg
Applications
Immunohistochemistry (Frozen) (1/10), Western Blot (Assay Dependent)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182934 700.00
Check your price
  • Complement C3 Antibody (Monoclonal, KT17)
  • Host Mouse
  • Target Species Human
  • Conjugate Unconjugated
  • Catalog # MA172513
  • Size 100 µg
Applications
ELISA (1 µg/ml)
Alternate names:
C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA172513 275.00
Check your price
  • Complement C3 Antibody (Monoclonal, 11H9)
  • Host Rat
  • Target Species Mouse
  • Conjugate Unconjugated
  • Catalog # MA140046
  • Size 100 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent.), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent.), Immunoprecipitation (Assay Dependent.), Western Blot (Assay Dependent.)
Alternate names:
HSE-MSF
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA140046 600.00
Check your price
  • Prohibitin Antibody (Monoclonal, 5H7)
  • Host Mouse
  • Target Species Human, Mouse, Non-human primate, Rat
  • Conjugate Unconjugated
  • Catalog # MA515899
  • Size 100 µL
Applications
Flow Cytometry (1/200-1/400), Immunocytochemistry (1/200-1/1000), Immunofluorescence (1:200-1:1000), Immunohistochemistry (1/200-1/1000), Western Blot (1/500-1/2000)
Alternate names:
B-cell receptor-associated protein 32
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA515899 349.00
Check your price
  • Prohibitin Antibody, Biotin (Monoclonal, II-14-10)
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Pig, Rat
  • Conjugate Biotin
  • Catalog # MA512855
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (2-4 µg/ml), Western Blot (0.5-1.0 µg/ml)
Alternate names:
Repressor of estrogen receptor activity; D-prohibitin; B-cell receptor-associated protein BAP37; B-cell receptor-associated protein 32
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512855 399.00
Check your price
  • Prohibitin Antibody (Monoclonal, II-14-10)
  • Host Mouse
  • Target Species Chicken, Human, Mouse, Pig, Rat
  • Conjugate Unconjugated
  • Catalog # MA512858
  • Size 500 µL
Applications
Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin) (2-4 µg/ml), Western Blot (0.5-1.0 µg/ml)
Alternate names:
Repressor of estrogen receptor activity; D-prohibitin; B-cell receptor-associated protein BAP37; B-cell receptor-associated protein 32
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA512858 349.00
Check your price
  • TPSAB1 Antibody (Monoclonal, AA1)
  • Host Mouse
  • Target Species Dog, Cat, Human, Non-human primate
  • Conjugate Unconjugated
  • Catalog # MA511711
  • Size 500 µL
Applications
Immunohistochemistry (Paraffin) (1:2000), Western Blot (1:50)
Alternate names:
Tryptase alpha-1; Tryptase I
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA511711 309.00
Check your price