1 - 3 of 3 products displayed
  • SLC25A25 Antibody
  • Host Rabbit
  • Target Species Human, Rat
  • Conjugate Unconjugated
  • Catalog # PA535293
  • Size 400 µL
Applications
Western Blot (1:1000)
Alternate names:
Solute carrier family 25 member 25; Small calcium-binding mitochondrial carrier protein 2; Mitochondrial Ca(2+)-dependent solute carrier protein 3; Mitochondrial ATP-Mg/Pi carrier protein 3; Peroxisomal Ca(2+)-dependent solute carrier-like protein; Mitochondrial ATP-Mg/Pi carrier protein
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA535293 349.00
Check your price
  • SLC25A23 Antibody
  • Host Rabbit
  • Target Species Human, Mouse
  • Conjugate Unconjugated
  • Catalog # PA530702
  • Size 100 µL
Applications
Immunocytochemistry (1:100-1:1000), Immunofluorescence (1:100-1:1000), Immunohistochemistry (Paraffin) (1:100-1:1000), Western Blot (1:500-1:3000)
Alternate names:
Solute carrier family 25 member 23; Small calcium-binding mitochondrial carrier protein 3; Mitochondrial Ca(2+)-dependent solute carrier protein 2; Mitochondrial ATP-Mg/Pi carrier protein 2
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA530702 349.00
Check your price
  • SLC25A24 Antibody
  • Host Rabbit
  • Target Species Bovine, Dog, Horse, Guinea pig, Human, Mouse, Rabbit, Rat, Zebrafish
  • Conjugate Unconjugated
  • Catalog # PA542459
  • Size 100 µl
Applications
Western Blot (2.5 µg/ml)
Alternate names:
Solute carrier family 25 member 24; Small calcium-binding mitochondrial carrier protein 1; Peroxisomal Ca(2+)-dependent solute carrier; Mitochondrial Ca(2+)-dependent solute carrier protein 1; Mitochondrial ATP-Mg/Pi carrier protein 1
Catalog # Size Price (AUD) Your Price (AUD) Qty
PA542459 309.00
Check your price