1 - 2 of 2 products displayed
  • Coronavirus pan Antibody (Monoclonal, FIPV3-70)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA182189
  • Size 250 µg
Applications
ELISA (Assay Dependent), Flow Cytometry (Assay Dependent), Immunofluorescence (Assay Dependent), Immunohistochemistry (Paraffin, Frozen) (Assay Dependent), Western Blot (Assay Dependent)
Alternate names:
canine coronavirus, CCV, pig coronavirus, swine coronavirus, transmissible gastroenteritis virus, TGEV, ferret coronavirus, bovine coronavirus, BCV, coronavirus nucleocapsid
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182189 499.00
Check your price
  • Canine/Feline Parvovirus Antibody (Monoclonal, CPVI-2AI)
  • Host Mouse
  • Target Species Virus
  • Conjugate Unconjugated
  • Catalog # MA182094
  • Size 250 µg
Applications
ELISA (Assay Dependent), Immunohistochemistry (Paraffin) (Assay Dependent)
Alternate names:
CPV, parvo, FPV
Catalog # Size Price (AUD) Your Price (AUD) Qty
MA182094 399.00
Check your price